Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44004185-44004681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf223
Entrez Gene ID
221416
Gene Name
chromosome 6 open reading frame 223
Gene Aliases
-
UniGene
Hs.531926
Gene Chromosome Location
Chr.6: 43999515-44007615 on Build GRCh38
Transcripts
NM_001171992.2 NM_001324369.1 NM_153246.5 XM_017010425.1 NR_125864.1

Gene Symbol
LOC101929705
Entrez Gene ID
101929705
Gene Name
uncharacterized LOC101929705
Gene Aliases
-
UniGene
Hs.670378
Gene Chromosome Location
Chr.6: 43995723-44074652 on Build GRCh38
Transcripts
NM_001171992.2 NM_001324369.1 NM_153246.5 XM_017010425.1 NR_125864.1

SNP Detail

SNP ID Chromosome Location
rs570228173 Chr.6: 44004248-44004248 on Build GRCh38
rs576143713 Chr.6: 44004649-44004649 on Build GRCh38
rs767784720 Chr.6: 44004424-44004424 on Build GRCh38
rs59069773 Chr.6: 44004489-44004527 on Build GRCh38
rs150976540 Chr.6: 44004498-44004498 on Build GRCh38
rs996556851 Chr.6: 44004553-44004553 on Build GRCh38
rs571835331 Chr.6: 44004423-44004423 on Build GRCh38
rs115059600 Chr.6: 44004555-44004555 on Build GRCh38
rs886560035 Chr.6: 44004531-44004531 on Build GRCh38
rs756403002 Chr.6: 44004496-44004496 on Build GRCh38
rs963537925 Chr.6: 44004526-44004526 on Build GRCh38
rs905085331 Chr.6: 44004493-44004493 on Build GRCh38
rs1002451755 Chr.6: 44004213-44004213 on Build GRCh38
rs993292502 Chr.6: 44004605-44004605 on Build GRCh38
rs73422480 Chr.6: 44004361-44004361 on Build GRCh38
rs987743408 Chr.6: 44004382-44004382 on Build GRCh38
rs555602722 Chr.6: 44004292-44004292 on Build GRCh38
rs954586234 Chr.6: 44004326-44004326 on Build GRCh38
rs533451835 Chr.6: 44004211-44004211 on Build GRCh38
rs999268462 Chr.6: 44004210-44004210 on Build GRCh38
rs896662251 Chr.6: 44004228-44004228 on Build GRCh38
rs142708210 Chr.6: 44004490-44004490 on Build GRCh38
rs969213346 Chr.6: 44004284-44004284 on Build GRCh38
rs1043193709 Chr.6: 44004466-44004466 on Build GRCh38
rs139724125 Chr.6: 44004489-44004489 on Build GRCh38
rs978460686 Chr.6: 44004365-44004365 on Build GRCh38
rs895802353 Chr.6: 44004231-44004231 on Build GRCh38
rs577689647 Chr.6: 44004241-44004241 on Build GRCh38
rs9394977 Chr.6: 44004316-44004316 on Build GRCh38
rs910781790 Chr.6: 44004387-44004387 on Build GRCh38
rs1030271323 Chr.6: 44004557-44004557 on Build GRCh38
rs1016092503 Chr.6: 44004511-44004511 on Build GRCh38
rs943529112 Chr.6: 44004399-44004399 on Build GRCh38
rs900523289 Chr.6: 44004543-44004543 on Build GRCh38
rs1055395579 Chr.6: 44004436-44004436 on Build GRCh38
rs934801299 Chr.6: 44004441-44004441 on Build GRCh38
rs954773364 Chr.6: 44004600-44004600 on Build GRCh38
rs35957677 Chr.6: 44004381-44004382 on Build GRCh38
rs987103737 Chr.6: 44004644-44004644 on Build GRCh38
rs540218639 Chr.6: 44004319-44004319 on Build GRCh38
rs1053605560 Chr.6: 44004444-44004444 on Build GRCh38
rs375124848 Chr.6: 44004525-44004525 on Build GRCh38
rs1006259933 Chr.6: 44004532-44004532 on Build GRCh38
rs145385774 Chr.6: 44004338-44004338 on Build GRCh38
rs35312564 Chr.6: 44004450-44004451 on Build GRCh38
rs936971286 Chr.6: 44004419-44004419 on Build GRCh38
rs543354075 Chr.6: 44004650-44004650 on Build GRCh38
rs896708922 Chr.6: 44004449-44004449 on Build GRCh38
rs371724833 Chr.6: 44004470-44004470 on Build GRCh38
rs1024861778 Chr.6: 44004491-44004491 on Build GRCh38
rs1047873263 Chr.6: 44004482-44004482 on Build GRCh38
rs1015976467 Chr.6: 44004318-44004318 on Build GRCh38
rs568391002 Chr.6: 44004217-44004217 on Build GRCh38
rs904607822 Chr.6: 44004439-44004439 on Build GRCh38
rs932970081 Chr.6: 44004372-44004372 on Build GRCh38
rs1024830412 Chr.6: 44004243-44004243 on Build GRCh38
rs545877893 Chr.6: 44004478-44004478 on Build GRCh38
rs980544598 Chr.6: 44004296-44004296 on Build GRCh38
rs1006207653 Chr.6: 44004273-44004273 on Build GRCh38
rs537384540 Chr.6: 44004263-44004263 on Build GRCh38
rs1026462112 Chr.6: 44004606-44004606 on Build GRCh38
rs754323437 Chr.6: 44004607-44004607 on Build GRCh38
rs1016262992 Chr.6: 44004535-44004535 on Build GRCh38

View Full Product Details