Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44300375-44300883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARS2
Entrez Gene ID
57505
Gene Name
alanyl-tRNA synthetase 2, mitochondrial
Gene Aliases
AARSL, COXPD8, LKENP, MT-ALARS, MTALARS
UniGene
Hs.158381
Gene Chromosome Location
Chr.6: 44297850-44313358 on Build GRCh38
Transcripts
NM_020745.3 XM_005249245.3 XM_011514764.2 XM_017011112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7306382 c.2800A>T p.M934L Chr.6: 44300705-44300705 on Build GRCh38
4401428 c.2898C>T p.S966S Chr.6: 44300607-44300607 on Build GRCh38
1181161 c.2797G>T p.A933S Chr.6: 44300708-44300708 on Build GRCh38
4153432 c.2946C>T p.L982L Chr.6: 44300559-44300559 on Build GRCh38
4625812 c.2917G>A p.A973T Chr.6: 44300588-44300588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775346092 Chr.6: 44300676-44300676 on Build GRCh38
rs952023504 Chr.6: 44300642-44300642 on Build GRCh38
rs879047268 Chr.6: 44300549-44300549 on Build GRCh38
rs772455600 Chr.6: 44300538-44300538 on Build GRCh38
rs369880654 Chr.6: 44300662-44300662 on Build GRCh38
rs977417091 Chr.6: 44300419-44300421 on Build GRCh38
rs960553899 Chr.6: 44300623-44300623 on Build GRCh38
rs551591793 Chr.6: 44300767-44300767 on Build GRCh38
rs763868546 Chr.6: 44300693-44300693 on Build GRCh38
rs934103959 Chr.6: 44300747-44300747 on Build GRCh38
rs545604368 Chr.6: 44300570-44300570 on Build GRCh38
rs774868654 Chr.6: 44300527-44300527 on Build GRCh38
rs113776635 Chr.6: 44300851-44300851 on Build GRCh38
rs767183912 Chr.6: 44300730-44300730 on Build GRCh38
rs570015024 Chr.6: 44300768-44300768 on Build GRCh38
rs564075656 Chr.6: 44300571-44300571 on Build GRCh38
rs940070914 Chr.6: 44300497-44300497 on Build GRCh38
rs769894604 Chr.6: 44300671-44300671 on Build GRCh38
rs371151506 Chr.6: 44300613-44300613 on Build GRCh38
rs373584667 Chr.6: 44300576-44300576 on Build GRCh38
rs199689375 Chr.6: 44300736-44300736 on Build GRCh38
rs771293152 Chr.6: 44300645-44300645 on Build GRCh38
rs758172170 Chr.6: 44300507-44300507 on Build GRCh38
rs771391213 Chr.6: 44300750-44300750 on Build GRCh38
rs939021954 Chr.6: 44300566-44300566 on Build GRCh38
rs990173391 Chr.6: 44300476-44300476 on Build GRCh38
rs772379787 Chr.6: 44300635-44300635 on Build GRCh38
rs746073297 Chr.6: 44300754-44300754 on Build GRCh38
rs766633645 Chr.6: 44300561-44300561 on Build GRCh38
rs916107096 Chr.6: 44300479-44300479 on Build GRCh38
rs572087334 Chr.6: 44300569-44300569 on Build GRCh38
rs77287454 Chr.6: 44300854-44300854 on Build GRCh38
rs756815754 Chr.6: 44300513-44300513 on Build GRCh38
rs751235289 Chr.6: 44300602-44300602 on Build GRCh38
rs756500207 Chr.6: 44300743-44300744 on Build GRCh38
rs755585135 Chr.6: 44300632-44300632 on Build GRCh38
rs946947871 Chr.6: 44300823-44300823 on Build GRCh38
rs773357868 Chr.6: 44300539-44300539 on Build GRCh38
rs909884318 Chr.6: 44300721-44300721 on Build GRCh38
rs924709551 Chr.6: 44300425-44300425 on Build GRCh38
rs325008 Chr.6: 44300634-44300634 on Build GRCh38
rs188305976 Chr.6: 44300781-44300781 on Build GRCh38
rs901342700 Chr.6: 44300500-44300500 on Build GRCh38
rs374497675 Chr.6: 44300735-44300735 on Build GRCh38
rs200778121 Chr.6: 44300568-44300568 on Build GRCh38
rs761544667 Chr.6: 44300728-44300728 on Build GRCh38
rs936061779 Chr.6: 44300437-44300437 on Build GRCh38
rs749953058 Chr.6: 44300625-44300625 on Build GRCh38
rs150039184 Chr.6: 44300600-44300600 on Build GRCh38
rs934176885 Chr.6: 44300518-44300518 on Build GRCh38
rs762941467 Chr.6: 44300685-44300685 on Build GRCh38
rs751367061 Chr.6: 44300714-44300714 on Build GRCh38
rs930596673 Chr.6: 44300410-44300410 on Build GRCh38
rs879701126 Chr.6: 44300778-44300778 on Build GRCh38
rs886061485 Chr.6: 44300711-44300711 on Build GRCh38
rs533429230 Chr.6: 44300746-44300746 on Build GRCh38
rs779332260 Chr.6: 44300633-44300633 on Build GRCh38
rs545868940 Chr.6: 44300738-44300738 on Build GRCh38
rs769161306 Chr.6: 44300526-44300526 on Build GRCh38
rs759729436 Chr.6: 44300660-44300660 on Build GRCh38
rs574430282 Chr.6: 44300537-44300537 on Build GRCh38
rs745307412 Chr.6: 44300525-44300525 on Build GRCh38
rs775641737 Chr.6: 44300758-44300758 on Build GRCh38
rs765200517 Chr.6: 44300496-44300496 on Build GRCh38
rs889961617 Chr.6: 44300816-44300816 on Build GRCh38
rs369397523 Chr.6: 44300512-44300512 on Build GRCh38
rs781613745 Chr.6: 44300753-44300753 on Build GRCh38
rs532546227 Chr.6: 44300743-44300743 on Build GRCh38
rs776944506 Chr.6: 44300656-44300656 on Build GRCh38
rs325009 Chr.6: 44300398-44300398 on Build GRCh38
rs772184730 Chr.6: 44300546-44300546 on Build GRCh38
rs966445611 Chr.6: 44300399-44300399 on Build GRCh38
rs780673226 Chr.6: 44300523-44300523 on Build GRCh38
rs916510673 Chr.6: 44300402-44300402 on Build GRCh38
rs760967545 Chr.6: 44300553-44300553 on Build GRCh38
rs543267101 Chr.6: 44300612-44300612 on Build GRCh38
rs377662340 Chr.6: 44300586-44300586 on Build GRCh38
rs984059361 Chr.6: 44300460-44300460 on Build GRCh38
rs535223487 Chr.6: 44300567-44300567 on Build GRCh38
rs565086376 Chr.6: 44300737-44300737 on Build GRCh38
rs907411309 Chr.6: 44300556-44300556 on Build GRCh38
rs752448574 Chr.6: 44300498-44300498 on Build GRCh38
rs747312867 Chr.6: 44300644-44300644 on Build GRCh38
rs142694525 Chr.6: 44300643-44300643 on Build GRCh38

View Full Product Details