Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53497611-53498156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCLC
Entrez Gene ID
2729
Gene Name
glutamate-cysteine ligase catalytic subunit
Gene Aliases
GCL, GCS, GLCL, GLCLC
UniGene
Hs.654465
Gene Chromosome Location
Chr.6: 53497341-53545129 on Build GRCh38
Transcripts
NM_001197115.1 NM_001498.3 XM_017010749.1

SNP Detail

SNP ID Chromosome Location
rs987644155 Chr.6: 53497981-53497981 on Build GRCh38
rs930586254 Chr.6: 53498035-53498035 on Build GRCh38
rs185495129 Chr.6: 53497816-53497816 on Build GRCh38
rs373507825 Chr.6: 53497658-53497658 on Build GRCh38
rs569060768 Chr.6: 53497888-53497888 on Build GRCh38
rs753065787 Chr.6: 53497868-53497869 on Build GRCh38
rs3830796 Chr.6: 53497650-53497651 on Build GRCh38
rs766894010 Chr.6: 53497789-53497789 on Build GRCh38
rs191159682 Chr.6: 53497671-53497671 on Build GRCh38
rs866647119 Chr.6: 53497919-53497919 on Build GRCh38
rs11298134 Chr.6: 53498005-53498005 on Build GRCh38
rs980685633 Chr.6: 53498084-53498084 on Build GRCh38
rs960647832 Chr.6: 53498000-53498000 on Build GRCh38
rs182188630 Chr.6: 53497763-53497763 on Build GRCh38
rs925351758 Chr.6: 53497908-53497908 on Build GRCh38
rs916785989 Chr.6: 53497881-53497881 on Build GRCh38
rs948213294 Chr.6: 53497889-53497889 on Build GRCh38
rs930544529 Chr.6: 53497800-53497800 on Build GRCh38
rs781756729 Chr.6: 53497649-53497650 on Build GRCh38
rs537426065 Chr.6: 53497920-53497920 on Build GRCh38
rs940481376 Chr.6: 53497722-53497722 on Build GRCh38
rs17883943 Chr.6: 53498092-53498092 on Build GRCh38
rs939329515 Chr.6: 53498097-53498097 on Build GRCh38
rs1018666776 Chr.6: 53497984-53497984 on Build GRCh38
rs1052420071 Chr.6: 53497912-53497912 on Build GRCh38
rs920408356 Chr.6: 53498016-53498016 on Build GRCh38
rs1045496978 Chr.6: 53497841-53497841 on Build GRCh38
rs774830884 Chr.6: 53497877-53497877 on Build GRCh38
rs762694641 Chr.6: 53497715-53497715 on Build GRCh38
rs143549488 Chr.6: 53497690-53497690 on Build GRCh38
rs971977095 Chr.6: 53497751-53497751 on Build GRCh38
rs17880473 Chr.6: 53497923-53497923 on Build GRCh38
rs538865533 Chr.6: 53497996-53497996 on Build GRCh38
rs398084940 Chr.6: 53498014-53498014 on Build GRCh38
rs9382214 Chr.6: 53497672-53497672 on Build GRCh38
rs925370908 Chr.6: 53498098-53498098 on Build GRCh38
rs190204915 Chr.6: 53497924-53497924 on Build GRCh38
rs754126523 Chr.6: 53498071-53498071 on Build GRCh38
rs756318386 Chr.6: 53498115-53498115 on Build GRCh38
rs1014613406 Chr.6: 53497994-53497994 on Build GRCh38
rs546373726 Chr.6: 53497693-53497693 on Build GRCh38
rs1027821253 Chr.6: 53497979-53497979 on Build GRCh38
rs550717773 Chr.6: 53497921-53497921 on Build GRCh38
rs1029102926 Chr.6: 53497974-53497974 on Build GRCh38
rs979546257 Chr.6: 53498094-53498094 on Build GRCh38
rs995978770 Chr.6: 53497938-53497938 on Build GRCh38
rs553598191 Chr.6: 53497988-53497988 on Build GRCh38
rs760137759 Chr.6: 53497910-53497910 on Build GRCh38
rs796873822 Chr.6: 53498015-53498015 on Build GRCh38
rs961880261 Chr.6: 53497986-53497988 on Build GRCh38
rs920545977 Chr.6: 53497765-53497765 on Build GRCh38
rs552407868 Chr.6: 53497855-53497855 on Build GRCh38
rs17881548 Chr.6: 53497989-53497989 on Build GRCh38
rs1043795400 Chr.6: 53497901-53497901 on Build GRCh38
rs764066428 Chr.6: 53498032-53498032 on Build GRCh38
rs916797194 Chr.6: 53498090-53498090 on Build GRCh38
rs17881167 Chr.6: 53497658-53497658 on Build GRCh38
rs397689937 Chr.6: 53497657-53497658 on Build GRCh38
rs369628691 Chr.6: 53498114-53498114 on Build GRCh38

View Full Product Details