Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 56954804-56955301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND6
Entrez Gene ID
221336
Gene Name
BEN domain containing 6
Gene Aliases
C6orf65
UniGene
Hs.582993
Gene Chromosome Location
Chr.6: 56954975-57027652 on Build GRCh38
Transcripts
NM_001318539.1 NM_152731.2 XM_011514346.2 XM_017010408.1

SNP Detail

SNP ID Chromosome Location
rs987515200 Chr.6: 56955098-56955098 on Build GRCh38
rs954839625 Chr.6: 56955261-56955261 on Build GRCh38
rs968498947 Chr.6: 56955226-56955226 on Build GRCh38
rs954842573 Chr.6: 56955088-56955088 on Build GRCh38
rs988633968 Chr.6: 56954888-56954888 on Build GRCh38
rs886705759 Chr.6: 56954945-56954945 on Build GRCh38
rs991600637 Chr.6: 56955159-56955159 on Build GRCh38
rs557561290 Chr.6: 56954926-56954926 on Build GRCh38
rs1035631703 Chr.6: 56954872-56954873 on Build GRCh38
rs1005067804 Chr.6: 56954846-56954846 on Build GRCh38
rs947512919 Chr.6: 56954908-56954908 on Build GRCh38
rs1044173129 Chr.6: 56954970-56954970 on Build GRCh38
rs1021498929 Chr.6: 56955212-56955212 on Build GRCh38
rs1056839988 Chr.6: 56955200-56955200 on Build GRCh38
rs1037895202 Chr.6: 56954958-56954958 on Build GRCh38
rs544240118 Chr.6: 56955120-56955120 on Build GRCh38
rs1044140865 Chr.6: 56955166-56955166 on Build GRCh38
rs886566066 Chr.6: 56954835-56954835 on Build GRCh38
rs573982192 Chr.6: 56955101-56955101 on Build GRCh38
rs914641461 Chr.6: 56954890-56954890 on Build GRCh38
rs560684233 Chr.6: 56955253-56955253 on Build GRCh38
rs1039034102 Chr.6: 56954823-56954823 on Build GRCh38
rs866812311 Chr.6: 56954864-56954864 on Build GRCh38
rs922057229 Chr.6: 56955086-56955086 on Build GRCh38
rs920723935 Chr.6: 56955149-56955149 on Build GRCh38
rs933600945 Chr.6: 56954829-56954829 on Build GRCh38
rs956006200 Chr.6: 56955066-56955066 on Build GRCh38
rs1000326993 Chr.6: 56954856-56954856 on Build GRCh38
rs561807560 Chr.6: 56955011-56955011 on Build GRCh38
rs974706949 Chr.6: 56954924-56954924 on Build GRCh38
rs1055278433 Chr.6: 56955252-56955252 on Build GRCh38
rs1022673939 Chr.6: 56954854-56954854 on Build GRCh38
rs1037968753 Chr.6: 56955130-56955130 on Build GRCh38
rs1052029722 Chr.6: 56954830-56954830 on Build GRCh38
rs891697917 Chr.6: 56955206-56955206 on Build GRCh38
rs995223612 Chr.6: 56954964-56954964 on Build GRCh38
rs1004063350 Chr.6: 56955231-56955231 on Build GRCh38
rs969759556 Chr.6: 56954855-56954855 on Build GRCh38
rs1010073324 Chr.6: 56955210-56955210 on Build GRCh38
rs898275516 Chr.6: 56954853-56954853 on Build GRCh38
rs995977453 Chr.6: 56955235-56955235 on Build GRCh38
rs533493489 Chr.6: 56955209-56955209 on Build GRCh38
rs562861910 Chr.6: 56955121-56955121 on Build GRCh38
rs540538892 Chr.6: 56954981-56954981 on Build GRCh38
rs981727398 Chr.6: 56954865-56954865 on Build GRCh38
rs1010022487 Chr.6: 56955067-56955067 on Build GRCh38
rs1052422048 Chr.6: 56954943-56954943 on Build GRCh38
rs974758943 Chr.6: 56955083-56955083 on Build GRCh38
rs898242933 Chr.6: 56954963-56954963 on Build GRCh38
rs1022471083 Chr.6: 56955279-56955279 on Build GRCh38
rs940839483 Chr.6: 56954954-56954954 on Build GRCh38
rs905504459 Chr.6: 56954998-56954998 on Build GRCh38
rs956039559 Chr.6: 56954885-56954885 on Build GRCh38
rs551888601 Chr.6: 56955247-56955247 on Build GRCh38
rs908100108 Chr.6: 56955115-56955115 on Build GRCh38
rs1036005148 Chr.6: 56955026-56955026 on Build GRCh38
rs933338408 Chr.6: 56954931-56954931 on Build GRCh38
rs948222018 Chr.6: 56955153-56955153 on Build GRCh38
rs905641038 Chr.6: 56955178-56955178 on Build GRCh38
rs1015321238 Chr.6: 56954851-56954851 on Build GRCh38
rs968529168 Chr.6: 56955071-56955072 on Build GRCh38
rs1002935880 Chr.6: 56955190-56955190 on Build GRCh38
rs572688845 Chr.6: 56954959-56954959 on Build GRCh38
rs895283702 Chr.6: 56955271-56955271 on Build GRCh38
rs1011045444 Chr.6: 56955272-56955272 on Build GRCh38
rs900760628 Chr.6: 56954826-56954826 on Build GRCh38
rs1021447925 Chr.6: 56955071-56955071 on Build GRCh38

View Full Product Details