Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73129653-73130180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNQ5
Entrez Gene ID
56479
Gene Name
potassium voltage-gated channel subfamily Q member 5
Gene Aliases
Kv7.5
UniGene
Hs.445324
Gene Chromosome Location
Chr.6: 72621843-73198851 on Build GRCh38
Transcripts
NM_001160130.1 NM_001160132.1 NM_001160133.1 NM_001160134.1 NM_019842.3 XM_017011058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6298717 c.1277+1G>T p.? Chr.6: 73129850-73129850 on Build GRCh38
5931844 c.1221-5C>T p.? Chr.6: 73129788-73129788 on Build GRCh38
6396495 c.1247T>C p.L416P Chr.6: 73129819-73129819 on Build GRCh38
7417574 c.1257G>A p.M419I Chr.6: 73129829-73129829 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748398140 Chr.6: 73129829-73129829 on Build GRCh38
rs1020104316 Chr.6: 73130150-73130150 on Build GRCh38
rs747059317 Chr.6: 73129772-73129772 on Build GRCh38
rs149059252 Chr.6: 73129741-73129741 on Build GRCh38
rs574476649 Chr.6: 73129854-73129854 on Build GRCh38
rs757602633 Chr.6: 73129748-73129748 on Build GRCh38
rs910431068 Chr.6: 73130007-73130008 on Build GRCh38
rs73753264 Chr.6: 73130052-73130052 on Build GRCh38
rs761456542 Chr.6: 73129863-73129863 on Build GRCh38
rs748100395 Chr.6: 73130157-73130157 on Build GRCh38
rs937386544 Chr.6: 73130017-73130017 on Build GRCh38
rs753429116 Chr.6: 73129862-73129862 on Build GRCh38
rs7748968 Chr.6: 73129733-73129733 on Build GRCh38
rs560013119 Chr.6: 73129975-73129975 on Build GRCh38
rs779345740 Chr.6: 73129749-73129749 on Build GRCh38
rs1029444086 Chr.6: 73130098-73130098 on Build GRCh38
rs946083831 Chr.6: 73129961-73129961 on Build GRCh38
rs758972019 Chr.6: 73129760-73129760 on Build GRCh38
rs60176558 Chr.6: 73130023-73130023 on Build GRCh38
rs767467020 Chr.6: 73129846-73129846 on Build GRCh38
rs199842840 Chr.6: 73129734-73129734 on Build GRCh38
rs911175925 Chr.6: 73129769-73129769 on Build GRCh38
rs1024394698 Chr.6: 73129761-73129761 on Build GRCh38
rs558682495 Chr.6: 73129721-73129721 on Build GRCh38
rs573060445 Chr.6: 73130146-73130146 on Build GRCh38
rs75503919 Chr.6: 73129991-73129991 on Build GRCh38
rs770566706 Chr.6: 73129836-73129836 on Build GRCh38
rs779944477 Chr.6: 73129768-73129768 on Build GRCh38
rs763703581 Chr.6: 73129743-73129743 on Build GRCh38
rs928606369 Chr.6: 73130000-73130000 on Build GRCh38
rs180676586 Chr.6: 73129791-73129791 on Build GRCh38
rs984741269 Chr.6: 73129973-73129973 on Build GRCh38
rs943225757 Chr.6: 73130015-73130015 on Build GRCh38
rs890778458 Chr.6: 73130111-73130111 on Build GRCh38
rs1015051615 Chr.6: 73129729-73129729 on Build GRCh38
rs994305512 Chr.6: 73130071-73130071 on Build GRCh38
rs913078463 Chr.6: 73129955-73129955 on Build GRCh38
rs541941734 Chr.6: 73129867-73129867 on Build GRCh38
rs887928349 Chr.6: 73129697-73129697 on Build GRCh38
rs746290611 Chr.6: 73129757-73129757 on Build GRCh38
rs957240413 Chr.6: 73130141-73130141 on Build GRCh38
rs774233700 Chr.6: 73129840-73129840 on Build GRCh38
rs1042035716 Chr.6: 73129719-73129719 on Build GRCh38
rs550055496 Chr.6: 73130093-73130093 on Build GRCh38
rs74455017 Chr.6: 73130153-73130153 on Build GRCh38
rs753553461 Chr.6: 73129745-73129745 on Build GRCh38
rs898018410 Chr.6: 73130060-73130060 on Build GRCh38
rs1000916374 Chr.6: 73130069-73130069 on Build GRCh38
rs187420713 Chr.6: 73130090-73130090 on Build GRCh38
rs765474698 Chr.6: 73129866-73129866 on Build GRCh38
rs74688588 Chr.6: 73130144-73130144 on Build GRCh38
rs6910765 Chr.6: 73129692-73129692 on Build GRCh38
rs1006377614 Chr.6: 73129717-73129717 on Build GRCh38
rs1029720863 Chr.6: 73129756-73129756 on Build GRCh38
rs533766989 Chr.6: 73129699-73129699 on Build GRCh38
rs1006597008 Chr.6: 73130121-73130121 on Build GRCh38
rs775329502 Chr.6: 73129849-73129849 on Build GRCh38
rs760173206 Chr.6: 73129853-73129853 on Build GRCh38
rs759343110 Chr.6: 73129844-73129844 on Build GRCh38
rs758733957 Chr.6: 73129893-73129893 on Build GRCh38
rs778779998 Chr.6: 73130091-73130091 on Build GRCh38
rs572072773 Chr.6: 73129987-73129987 on Build GRCh38
rs60813738 Chr.6: 73130024-73130024 on Build GRCh38
rs776983150 Chr.6: 73129808-73129808 on Build GRCh38
rs553154007 Chr.6: 73130139-73130139 on Build GRCh38
rs754491093 Chr.6: 73130034-73130034 on Build GRCh38
rs989942851 Chr.6: 73129913-73129913 on Build GRCh38
rs1042066992 Chr.6: 73130035-73130035 on Build GRCh38
rs1056073145 Chr.6: 73129687-73129687 on Build GRCh38

View Full Product Details