Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73352984-73353481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPPA5
Entrez Gene ID
340168
Gene Name
developmental pluripotency associated 5
Gene Aliases
ESG1
UniGene
Hs.125331
Gene Chromosome Location
Chr.6: 73353062-73354276 on Build GRCh38
Transcripts
NM_001025290.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1634920 c.311A>G p.E104G Chr.6: 73353360-73353360 on Build GRCh38
1672558 c.310G>A p.E104K Chr.6: 73353361-73353361 on Build GRCh38
4419096 c.324C>G p.A108A Chr.6: 73353347-73353347 on Build GRCh38
1081514 c.328G>A p.E110K Chr.6: 73353343-73353343 on Build GRCh38
3697961 c.309C>T p.A103A Chr.6: 73353362-73353362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759476774 Chr.6: 73353008-73353008 on Build GRCh38
rs756449448 Chr.6: 73353401-73353401 on Build GRCh38
rs768683714 Chr.6: 73353404-73353404 on Build GRCh38
rs1041652773 Chr.6: 73353134-73353134 on Build GRCh38
rs777330080 Chr.6: 73353344-73353344 on Build GRCh38
rs76770796 Chr.6: 73353067-73353067 on Build GRCh38
rs920326044 Chr.6: 73353225-73353225 on Build GRCh38
rs756401898 Chr.6: 73353349-73353349 on Build GRCh38
rs1047527915 Chr.6: 73353232-73353232 on Build GRCh38
rs1035925295 Chr.6: 73353226-73353226 on Build GRCh38
rs1002738363 Chr.6: 73353195-73353195 on Build GRCh38
rs35696509 Chr.6: 73353379-73353380 on Build GRCh38
rs755037198 Chr.6: 73353335-73353335 on Build GRCh38
rs775969448 Chr.6: 73353416-73353416 on Build GRCh38
rs770628324 Chr.6: 73353308-73353309 on Build GRCh38
rs202225803 Chr.6: 73353352-73353352 on Build GRCh38
rs919472611 Chr.6: 73353007-73353007 on Build GRCh38
rs942420830 Chr.6: 73353053-73353053 on Build GRCh38
rs761345763 Chr.6: 73353305-73353305 on Build GRCh38
rs1038759637 Chr.6: 73353210-73353210 on Build GRCh38
rs757900680 Chr.6: 73353310-73353310 on Build GRCh38
rs12200929 Chr.6: 73353369-73353369 on Build GRCh38
rs769347147 Chr.6: 73353400-73353401 on Build GRCh38
rs987694371 Chr.6: 73353432-73353432 on Build GRCh38
rs200196148 Chr.6: 73353308-73353308 on Build GRCh38
rs371129787 Chr.6: 73353109-73353109 on Build GRCh38
rs772000557 Chr.6: 73353378-73353378 on Build GRCh38
rs1052347317 Chr.6: 73353038-73353038 on Build GRCh38
rs886250322 Chr.6: 73353244-73353244 on Build GRCh38
rs780776613 Chr.6: 73353333-73353333 on Build GRCh38
rs912345645 Chr.6: 73353051-73353051 on Build GRCh38
rs759070060 Chr.6: 73353277-73353277 on Build GRCh38
rs776293984 Chr.6: 73353292-73353292 on Build GRCh38
rs143768205 Chr.6: 73353049-73353049 on Build GRCh38
rs911486295 Chr.6: 73353114-73353114 on Build GRCh38
rs766813622 Chr.6: 73353319-73353319 on Build GRCh38
rs943023060 Chr.6: 73353180-73353180 on Build GRCh38
rs987021354 Chr.6: 73353455-73353455 on Build GRCh38
rs753298805 Chr.6: 73353012-73353012 on Build GRCh38
rs569604085 Chr.6: 73353391-73353391 on Build GRCh38
rs910709167 Chr.6: 73353450-73353450 on Build GRCh38
rs765257977 Chr.6: 73353075-73353075 on Build GRCh38
rs955693368 Chr.6: 73353382-73353382 on Build GRCh38
rs879200675 Chr.6: 73353268-73353268 on Build GRCh38
rs560801490 Chr.6: 73353186-73353186 on Build GRCh38
rs745603346 Chr.6: 73353377-73353377 on Build GRCh38
rs563289316 Chr.6: 73353105-73353105 on Build GRCh38
rs369093519 Chr.6: 73353128-73353128 on Build GRCh38
rs930410366 Chr.6: 73353231-73353231 on Build GRCh38
rs1010248664 Chr.6: 73353348-73353348 on Build GRCh38
rs745506457 Chr.6: 73353388-73353388 on Build GRCh38
rs761451170 Chr.6: 73353419-73353419 on Build GRCh38
rs371684858 Chr.6: 73353400-73353400 on Build GRCh38
rs374958598 Chr.6: 73353421-73353421 on Build GRCh38
rs376865248 Chr.6: 73353104-73353105 on Build GRCh38
rs73754622 Chr.6: 73353347-73353347 on Build GRCh38
rs772864348 Chr.6: 73353429-73353429 on Build GRCh38
rs367701006 Chr.6: 73353094-73353094 on Build GRCh38
rs200904292 Chr.6: 73353284-73353284 on Build GRCh38
rs564883537 Chr.6: 73353334-73353334 on Build GRCh38
rs758819995 Chr.6: 73353312-73353312 on Build GRCh38

View Full Product Details