Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 76002838-76003338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPG1
Entrez Gene ID
3617
Gene Name
interphotoreceptor matrix proteoglycan 1
Gene Aliases
GP147, IPM150, SPACR, VMD4
UniGene
Hs.590893
Gene Chromosome Location
Chr.6: 75921114-76072678 on Build GRCh38
Transcripts
NM_001282368.1 NM_001563.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3949483 c.1270G>T p.G424* Chr.6: 76002939-76002939 on Build GRCh38
1446024 c.1244T>A p.V415D Chr.6: 76002965-76002965 on Build GRCh38
743081 c.1274C>A p.A425E Chr.6: 76002935-76002935 on Build GRCh38
4854933 c.1272A>G p.G424G Chr.6: 76002937-76002937 on Build GRCh38
5615729 c.1268A>T p.D423V Chr.6: 76002941-76002941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988106417 Chr.6: 76003164-76003164 on Build GRCh38
rs767759914 Chr.6: 76003046-76003046 on Build GRCh38
rs78195046 Chr.6: 76002938-76002938 on Build GRCh38
rs150541950 Chr.6: 76002942-76002942 on Build GRCh38
rs1012168212 Chr.6: 76003099-76003099 on Build GRCh38
rs868584856 Chr.6: 76002956-76002956 on Build GRCh38
rs781158485 Chr.6: 76002872-76002872 on Build GRCh38
rs749562634 Chr.6: 76002977-76002977 on Build GRCh38
rs931682322 Chr.6: 76003202-76003202 on Build GRCh38
rs138431617 Chr.6: 76002981-76002981 on Build GRCh38
rs545137699 Chr.6: 76003090-76003090 on Build GRCh38
rs980894177 Chr.6: 76003186-76003186 on Build GRCh38
rs1028842880 Chr.6: 76003065-76003065 on Build GRCh38
rs370915172 Chr.6: 76002952-76002952 on Build GRCh38
rs777997248 Chr.6: 76002976-76002976 on Build GRCh38
rs967858041 Chr.6: 76003123-76003123 on Build GRCh38
rs939617510 Chr.6: 76003205-76003205 on Build GRCh38
rs766017144 Chr.6: 76003040-76003040 on Build GRCh38
rs373282933 Chr.6: 76002876-76002876 on Build GRCh38
rs377510349 Chr.6: 76002894-76002894 on Build GRCh38
rs368892463 Chr.6: 76002933-76002933 on Build GRCh38
rs940298851 Chr.6: 76003213-76003213 on Build GRCh38
rs968118609 Chr.6: 76003179-76003179 on Build GRCh38
rs910163708 Chr.6: 76003223-76003223 on Build GRCh38
rs576611684 Chr.6: 76003260-76003260 on Build GRCh38
rs772674590 Chr.6: 76003033-76003033 on Build GRCh38
rs776569999 Chr.6: 76003017-76003017 on Build GRCh38
rs908801838 Chr.6: 76003211-76003211 on Build GRCh38
rs536935278 Chr.6: 76002969-76002969 on Build GRCh38
rs772059757 Chr.6: 76002898-76002898 on Build GRCh38
rs577767810 Chr.6: 76003087-76003087 on Build GRCh38
rs762619388 Chr.6: 76003039-76003039 on Build GRCh38
rs551239189 Chr.6: 76002897-76002897 on Build GRCh38
rs953306416 Chr.6: 76003079-76003079 on Build GRCh38
rs867940475 Chr.6: 76003166-76003166 on Build GRCh38
rs921618992 Chr.6: 76003175-76003175 on Build GRCh38
rs888984693 Chr.6: 76003266-76003266 on Build GRCh38
rs1036047480 Chr.6: 76003214-76003214 on Build GRCh38
rs182414904 Chr.6: 76003269-76003269 on Build GRCh38
rs534568869 Chr.6: 76003064-76003064 on Build GRCh38
rs753319298 Chr.6: 76002923-76002923 on Build GRCh38
rs758038128 Chr.6: 76002940-76002940 on Build GRCh38
rs761720817 Chr.6: 76003023-76003023 on Build GRCh38
rs41269327 Chr.6: 76002974-76002974 on Build GRCh38
rs750131334 Chr.6: 76002939-76002939 on Build GRCh38
rs187315709 Chr.6: 76003085-76003085 on Build GRCh38
rs760577375 Chr.6: 76002909-76002909 on Build GRCh38
rs775167831 Chr.6: 76002984-76002984 on Build GRCh38
rs755474162 Chr.6: 76002949-76002949 on Build GRCh38
rs775325166 Chr.6: 76002901-76002901 on Build GRCh38
rs768481753 Chr.6: 76002990-76002990 on Build GRCh38
rs751062564 Chr.6: 76003124-76003124 on Build GRCh38
rs1057152859 Chr.6: 76003210-76003210 on Build GRCh38
rs192372880 Chr.6: 76003248-76003248 on Build GRCh38
rs764041223 Chr.6: 76002922-76002922 on Build GRCh38
rs149526718 Chr.6: 76003029-76003029 on Build GRCh38
rs770148586 Chr.6: 76002879-76002879 on Build GRCh38
rs898885561 Chr.6: 76003240-76003240 on Build GRCh38
rs1022255143 Chr.6: 76003108-76003108 on Build GRCh38
rs932927833 Chr.6: 76002883-76002883 on Build GRCh38
rs769188306 Chr.6: 76002868-76002868 on Build GRCh38
rs746778199 Chr.6: 76002985-76002985 on Build GRCh38
rs751878384 Chr.6: 76003044-76003044 on Build GRCh38
rs1018941659 Chr.6: 76003128-76003128 on Build GRCh38
rs534225874 Chr.6: 76003133-76003133 on Build GRCh38
rs867747143 Chr.6: 76002926-76002926 on Build GRCh38
rs926868749 Chr.6: 76002929-76002929 on Build GRCh38
rs761369009 Chr.6: 76003189-76003189 on Build GRCh38
rs781678292 Chr.6: 76002968-76002968 on Build GRCh38
rs1019241067 Chr.6: 76003300-76003300 on Build GRCh38
rs755315333 Chr.6: 76003045-76003045 on Build GRCh38
rs763772888 Chr.6: 76003055-76003055 on Build GRCh38

View Full Product Details