Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 82171345-82171880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IBTK
Entrez Gene ID
25998
Gene Name
inhibitor of Bruton tyrosine kinase
Gene Aliases
BTBD26, BTKI
UniGene
Hs.306425
Gene Chromosome Location
Chr.6: 82169987-82248294 on Build GRCh38
Transcripts
NM_001300906.1 NM_015525.3 XM_006715453.2 XM_006715454.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586944 c.4017G>A p.P1339P Chr.6: 82171470-82171470 on Build GRCh38
7207743 c.4048A>T p.K1350* Chr.6: 82171439-82171439 on Build GRCh38
6369278 c.3997G>A p.V1333I Chr.6: 82171490-82171490 on Build GRCh38
6414300 c.3946A>G p.I1316V Chr.6: 82171541-82171541 on Build GRCh38
6579338 c.3982_3993del12 p.N1328_E1331delNPEE Chr.6: 82171494-82171505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780404182 Chr.6: 82171412-82171412 on Build GRCh38
rs370973651 Chr.6: 82171396-82171396 on Build GRCh38
rs772091877 Chr.6: 82171469-82171469 on Build GRCh38
rs763196082 Chr.6: 82171513-82171513 on Build GRCh38
rs971525215 Chr.6: 82171418-82171418 on Build GRCh38
rs778114810 Chr.6: 82171563-82171563 on Build GRCh38
rs201175318 Chr.6: 82171519-82171519 on Build GRCh38
rs368471443 Chr.6: 82171389-82171389 on Build GRCh38
rs779848043 Chr.6: 82171588-82171588 on Build GRCh38
rs376800097 Chr.6: 82171560-82171560 on Build GRCh38
rs1039835509 Chr.6: 82171659-82171659 on Build GRCh38
rs781287639 Chr.6: 82171432-82171432 on Build GRCh38
rs759002757 Chr.6: 82171399-82171399 on Build GRCh38
rs1014979111 Chr.6: 82171812-82171812 on Build GRCh38
rs757859436 Chr.6: 82171386-82171386 on Build GRCh38
rs367708288 Chr.6: 82171583-82171583 on Build GRCh38
rs899989117 Chr.6: 82171686-82171686 on Build GRCh38
rs752817893 Chr.6: 82171559-82171559 on Build GRCh38
rs1027237209 Chr.6: 82171774-82171774 on Build GRCh38
rs745641913 Chr.6: 82171465-82171465 on Build GRCh38
rs945990109 Chr.6: 82171574-82171574 on Build GRCh38
rs140917980 Chr.6: 82171614-82171614 on Build GRCh38
rs367607944 Chr.6: 82171484-82171484 on Build GRCh38
rs868737509 Chr.6: 82171393-82171393 on Build GRCh38
rs774352439 Chr.6: 82171463-82171463 on Build GRCh38
rs770072853 Chr.6: 82171454-82171454 on Build GRCh38
rs923424844 Chr.6: 82171678-82171678 on Build GRCh38
rs762643621 Chr.6: 82171512-82171512 on Build GRCh38
rs201304103 Chr.6: 82171472-82171472 on Build GRCh38
rs764858543 Chr.6: 82171502-82171502 on Build GRCh38
rs145073081 Chr.6: 82171717-82171717 on Build GRCh38
rs565792598 Chr.6: 82171470-82171470 on Build GRCh38
rs559321501 Chr.6: 82171789-82171789 on Build GRCh38
rs951681071 Chr.6: 82171786-82171786 on Build GRCh38
rs746932197 Chr.6: 82171601-82171601 on Build GRCh38
rs1006147086 Chr.6: 82171820-82171820 on Build GRCh38
rs755458338 Chr.6: 82171535-82171535 on Build GRCh38
rs375781712 Chr.6: 82171845-82171845 on Build GRCh38
rs111734593 Chr.6: 82171376-82171376 on Build GRCh38
rs760619638 Chr.6: 82171471-82171471 on Build GRCh38
rs556109702 Chr.6: 82171637-82171637 on Build GRCh38
rs765592139 Chr.6: 82171384-82171384 on Build GRCh38
rs61742744 Chr.6: 82171430-82171430 on Build GRCh38
rs141043970 Chr.6: 82171529-82171529 on Build GRCh38
rs747156865 Chr.6: 82171415-82171415 on Build GRCh38
rs371750912 Chr.6: 82171495-82171495 on Build GRCh38
rs1042078881 Chr.6: 82171575-82171575 on Build GRCh38
rs748398171 Chr.6: 82171448-82171448 on Build GRCh38
rs1008707285 Chr.6: 82171633-82171633 on Build GRCh38
rs542243154 Chr.6: 82171722-82171722 on Build GRCh38
rs905802929 Chr.6: 82171848-82171848 on Build GRCh38
rs781428088 Chr.6: 82171554-82171554 on Build GRCh38
rs138107539 Chr.6: 82171428-82171428 on Build GRCh38
rs1004481795 Chr.6: 82171791-82171791 on Build GRCh38
rs907542110 Chr.6: 82171370-82171370 on Build GRCh38
rs745693309 Chr.6: 82171570-82171570 on Build GRCh38
rs776060160 Chr.6: 82171487-82171487 on Build GRCh38
rs768169135 Chr.6: 82171596-82171596 on Build GRCh38
rs760525198 Chr.6: 82171584-82171585 on Build GRCh38
rs891609528 Chr.6: 82171677-82171697 on Build GRCh38

View Full Product Details