Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89562615-89563111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD6
Entrez Gene ID
22881
Gene Name
ankyrin repeat domain 6
Gene Aliases
-
UniGene
Hs.702213
Gene Chromosome Location
Chr.6: 89433178-89633834 on Build GRCh38
Transcripts
NM_001242809.1 NM_001242811.1 NM_001242813.1 NM_014942.4 XM_005248680.3 XM_005248681.2 XM_011535598.2 XM_011535599.2 XM_011535600.2 XM_017010495.1 XM_017010496.1 XM_017010498.1 XM_017010501.1 XM_017010502.1 XM_017010503.1 XM_017010505.1 XM_017010508.1 XM_017010510.1 XM_017010511.1 XM_017010512.1 XM_017010513.1 XM_017010514.1

SNP Detail

SNP ID Chromosome Location
rs902665624 Chr.6: 89562794-89562794 on Build GRCh38
rs796469815 Chr.6: 89562822-89562822 on Build GRCh38
rs1011590717 Chr.6: 89562784-89562784 on Build GRCh38
rs984426188 Chr.6: 89562842-89562842 on Build GRCh38
rs373121056 Chr.6: 89563050-89563050 on Build GRCh38
rs756521791 Chr.6: 89562956-89562956 on Build GRCh38
rs550759855 Chr.6: 89562831-89562831 on Build GRCh38
rs970001434 Chr.6: 89562769-89562769 on Build GRCh38
rs149760535 Chr.6: 89562702-89562702 on Build GRCh38
rs868577735 Chr.6: 89562698-89562698 on Build GRCh38
rs866379186 Chr.6: 89562748-89562748 on Build GRCh38
rs911472246 Chr.6: 89562888-89562888 on Build GRCh38
rs939893338 Chr.6: 89562669-89562669 on Build GRCh38
rs187044122 Chr.6: 89563032-89563032 on Build GRCh38
rs922317638 Chr.6: 89563025-89563025 on Build GRCh38
rs1022440428 Chr.6: 89562793-89562793 on Build GRCh38
rs769586290 Chr.6: 89562866-89562866 on Build GRCh38
rs888505581 Chr.6: 89562665-89562665 on Build GRCh38
rs145719671 Chr.6: 89563087-89563087 on Build GRCh38
rs780821954 Chr.6: 89562773-89562773 on Build GRCh38
rs762809986 Chr.6: 89563018-89563018 on Build GRCh38
rs909829281 Chr.6: 89562870-89562870 on Build GRCh38
rs999730334 Chr.6: 89562798-89562798 on Build GRCh38
rs867947010 Chr.6: 89563088-89563088 on Build GRCh38
rs775383201 Chr.6: 89562971-89562971 on Build GRCh38
rs1035098611 Chr.6: 89562809-89562809 on Build GRCh38
rs561986420 Chr.6: 89562774-89562774 on Build GRCh38
rs942931047 Chr.6: 89562901-89562901 on Build GRCh38
rs78358725 Chr.6: 89562839-89562839 on Build GRCh38
rs767797061 Chr.6: 89562815-89562815 on Build GRCh38
rs192177668 Chr.6: 89562710-89562710 on Build GRCh38
rs1041694900 Chr.6: 89562999-89562999 on Build GRCh38
rs909725044 Chr.6: 89562654-89562654 on Build GRCh38
rs528380430 Chr.6: 89562707-89562707 on Build GRCh38
rs145716683 Chr.6: 89562892-89562892 on Build GRCh38
rs369702311 Chr.6: 89562931-89562931 on Build GRCh38
rs182371693 Chr.6: 89562830-89562830 on Build GRCh38
rs1036728596 Chr.6: 89562680-89562680 on Build GRCh38
rs888419855 Chr.6: 89562741-89562741 on Build GRCh38
rs994224490 Chr.6: 89562690-89562690 on Build GRCh38
rs191849862 Chr.6: 89563033-89563033 on Build GRCh38
rs1016168687 Chr.6: 89562749-89562749 on Build GRCh38
rs574793188 Chr.6: 89563057-89563057 on Build GRCh38
rs947041510 Chr.6: 89562847-89562847 on Build GRCh38
rs140393378 Chr.6: 89562653-89562653 on Build GRCh38
rs572521755 Chr.6: 89562807-89562807 on Build GRCh38
rs140584705 Chr.6: 89563031-89563031 on Build GRCh38
rs372886420 Chr.6: 89562795-89562795 on Build GRCh38
rs978423200 Chr.6: 89562852-89562852 on Build GRCh38
rs551067520 Chr.6: 89562860-89562860 on Build GRCh38
rs568849065 Chr.6: 89562835-89562835 on Build GRCh38

View Full Product Details