Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 114056637-114057144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS3ST5
Entrez Gene ID
222537
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 5
Gene Aliases
3-OST-5, 3OST5, HS3OST5, NBLA04021
UniGene
Hs.645477
Gene Chromosome Location
Chr.6: 114055586-114342388 on Build GRCh38
Transcripts
NM_153612.3 XM_006715379.2 XM_011535588.2 XM_017010470.1 XM_017010471.1 XM_017010472.1 XM_017010473.1 XM_017010474.1 NR_125845.1

Gene Symbol
LOC101927768
Entrez Gene ID
101927768
Gene Name
uncharacterized LOC101927768
Gene Aliases
-
UniGene
Hs.129280
Gene Chromosome Location
Chr.6: 113969701-114340729 on Build GRCh38
Transcripts
NM_153612.3 XM_006715379.2 XM_011535588.2 XM_017010470.1 XM_017010471.1 XM_017010472.1 XM_017010473.1 XM_017010474.1 NR_125845.1

SNP Detail

SNP ID Chromosome Location
rs573632185 Chr.6: 114056815-114056815 on Build GRCh38
rs988206490 Chr.6: 114056695-114056695 on Build GRCh38
rs113461156 Chr.6: 114056933-114056933 on Build GRCh38
rs976704691 Chr.6: 114057026-114057026 on Build GRCh38
rs17075821 Chr.6: 114056982-114056982 on Build GRCh38
rs942549722 Chr.6: 114056758-114056758 on Build GRCh38
rs756053330 Chr.6: 114057060-114057060 on Build GRCh38
rs761975532 Chr.6: 114056815-114056816 on Build GRCh38
rs1028257861 Chr.6: 114057041-114057041 on Build GRCh38
rs1002426983 Chr.6: 114056916-114056916 on Build GRCh38
rs544459341 Chr.6: 114057069-114057070 on Build GRCh38
rs958119926 Chr.6: 114056685-114056685 on Build GRCh38
rs905462041 Chr.6: 114056889-114056889 on Build GRCh38
rs1012485304 Chr.6: 114056935-114056935 on Build GRCh38
rs574108026 Chr.6: 114056848-114056848 on Build GRCh38
rs906875379 Chr.6: 114056842-114056842 on Build GRCh38
rs949734100 Chr.6: 114056872-114056872 on Build GRCh38
rs556370075 Chr.6: 114056903-114056903 on Build GRCh38
rs550062854 Chr.6: 114057109-114057109 on Build GRCh38
rs565749652 Chr.6: 114056694-114056694 on Build GRCh38
rs1041459589 Chr.6: 114056834-114056834 on Build GRCh38
rs986734407 Chr.6: 114056981-114056982 on Build GRCh38
rs764477498 Chr.6: 114057065-114057065 on Build GRCh38
rs140456892 Chr.6: 114056818-114056818 on Build GRCh38
rs745938513 Chr.6: 114056857-114056857 on Build GRCh38
rs932512193 Chr.6: 114056798-114056798 on Build GRCh38
rs1034050333 Chr.6: 114056956-114056956 on Build GRCh38
rs895369826 Chr.6: 114056891-114056891 on Build GRCh38
rs1055495788 Chr.6: 114057098-114057098 on Build GRCh38
rs558305057 Chr.6: 114056989-114056989 on Build GRCh38
rs1054726283 Chr.6: 114056812-114056812 on Build GRCh38
rs1017561019 Chr.6: 114056945-114056945 on Build GRCh38
rs1002590747 Chr.6: 114056949-114056949 on Build GRCh38
rs938338211 Chr.6: 114057070-114057070 on Build GRCh38
rs1032518240 Chr.6: 114056948-114056948 on Build GRCh38
rs1021374813 Chr.6: 114056736-114056736 on Build GRCh38
rs575430357 Chr.6: 114056662-114056662 on Build GRCh38
rs571817864 Chr.6: 114057119-114057119 on Build GRCh38
rs557681371 Chr.6: 114056822-114056822 on Build GRCh38
rs80329057 Chr.6: 114057070-114057070 on Build GRCh38
rs796353937 Chr.6: 114056845-114056846 on Build GRCh38
rs939774366 Chr.6: 114056833-114056833 on Build GRCh38
rs983969276 Chr.6: 114057053-114057053 on Build GRCh38
rs74668424 Chr.6: 114056969-114056969 on Build GRCh38
rs75633161 Chr.6: 114057068-114057068 on Build GRCh38
rs921168692 Chr.6: 114056784-114056784 on Build GRCh38
rs372505575 Chr.6: 114057037-114057037 on Build GRCh38
rs1056748339 Chr.6: 114056880-114056880 on Build GRCh38
rs1046704633 Chr.6: 114056876-114056876 on Build GRCh38
rs532080527 Chr.6: 114057107-114057107 on Build GRCh38
rs994670174 Chr.6: 114056951-114056951 on Build GRCh38
rs928344011 Chr.6: 114057069-114057069 on Build GRCh38
rs555218633 Chr.6: 114056760-114056760 on Build GRCh38
rs1002499171 Chr.6: 114056846-114056846 on Build GRCh38
rs955393757 Chr.6: 114056974-114056974 on Build GRCh38
rs533805477 Chr.6: 114056745-114056745 on Build GRCh38
rs1032486929 Chr.6: 114056684-114056684 on Build GRCh38
rs561560716 Chr.6: 114057100-114057100 on Build GRCh38
rs542679924 Chr.6: 114057073-114057073 on Build GRCh38
rs965789799 Chr.6: 114056998-114056998 on Build GRCh38
rs527258323 Chr.6: 114057052-114057052 on Build GRCh38
rs977110861 Chr.6: 114056768-114056768 on Build GRCh38
rs577618478 Chr.6: 114056946-114056946 on Build GRCh38

View Full Product Details