Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125760595-125761108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEY2
Entrez Gene ID
23493
Gene Name
hes related family bHLH transcription factor with YRPW motif 2
Gene Aliases
CHF1, GRIDLOCK, GRL, HERP1, HESR2, HRT2, bHLHb32
UniGene
Hs.144287
Gene Chromosome Location
Chr.6: 125747592-125762243 on Build GRCh38
Transcripts
NM_012259.2 XM_017010627.1 XM_017010628.1 XM_017010629.1

SNP Detail

SNP ID Chromosome Location
rs779962556 Chr.6: 125760817-125760817 on Build GRCh38
rs576229362 Chr.6: 125760640-125760640 on Build GRCh38
rs778246120 Chr.6: 125760903-125760903 on Build GRCh38
rs1006275057 Chr.6: 125760742-125760742 on Build GRCh38
rs749163582 Chr.6: 125760859-125760859 on Build GRCh38
rs779701608 Chr.6: 125760806-125760806 on Build GRCh38
rs1040703051 Chr.6: 125760995-125760995 on Build GRCh38
rs972069518 Chr.6: 125760821-125760821 on Build GRCh38
rs887773350 Chr.6: 125761065-125761065 on Build GRCh38
rs544102568 Chr.6: 125760677-125760677 on Build GRCh38
rs528379150 Chr.6: 125761046-125761046 on Build GRCh38
rs996735900 Chr.6: 125760999-125760999 on Build GRCh38
rs928653833 Chr.6: 125760848-125760848 on Build GRCh38
rs1002103793 Chr.6: 125761055-125761055 on Build GRCh38
rs529712818 Chr.6: 125760705-125760705 on Build GRCh38
rs952525066 Chr.6: 125760738-125760738 on Build GRCh38
rs994000194 Chr.6: 125761084-125761084 on Build GRCh38
rs562208058 Chr.6: 125760699-125760699 on Build GRCh38
rs986788965 Chr.6: 125760984-125760984 on Build GRCh38
rs1013399483 Chr.6: 125760649-125760649 on Build GRCh38
rs984865465 Chr.6: 125760855-125760855 on Build GRCh38
rs936346238 Chr.6: 125760990-125760990 on Build GRCh38
rs191249768 Chr.6: 125760894-125760894 on Build GRCh38
rs541487154 Chr.6: 125760719-125760719 on Build GRCh38
rs888398659 Chr.6: 125761034-125761034 on Build GRCh38
rs760830552 Chr.6: 125761008-125761010 on Build GRCh38
rs1048802969 Chr.6: 125761063-125761063 on Build GRCh38
rs892269915 Chr.6: 125760628-125760628 on Build GRCh38
rs923512121 Chr.6: 125760926-125760926 on Build GRCh38
rs1028129191 Chr.6: 125760736-125760739 on Build GRCh38
rs998526551 Chr.6: 125760689-125760689 on Build GRCh38
rs913677231 Chr.6: 125761009-125761009 on Build GRCh38
rs766411759 Chr.6: 125760700-125760703 on Build GRCh38
rs745982551 Chr.6: 125761038-125761038 on Build GRCh38
rs753034359 Chr.6: 125760861-125760861 on Build GRCh38
rs747527320 Chr.6: 125760905-125760905 on Build GRCh38
rs924362343 Chr.6: 125760960-125760960 on Build GRCh38
rs1016349437 Chr.6: 125760790-125760790 on Build GRCh38
rs866399785 Chr.6: 125760941-125760941 on Build GRCh38
rs552298764 Chr.6: 125760968-125760968 on Build GRCh38
rs866161707 Chr.6: 125760863-125760863 on Build GRCh38
rs947115361 Chr.6: 125760646-125760646 on Build GRCh38
rs924946927 Chr.6: 125760989-125760989 on Build GRCh38
rs962137065 Chr.6: 125761069-125761069 on Build GRCh38
rs1015457138 Chr.6: 125761045-125761045 on Build GRCh38
rs75759893 Chr.6: 125761025-125761025 on Build GRCh38
rs770120208 Chr.6: 125761072-125761072 on Build GRCh38
rs547478367 Chr.6: 125760996-125760996 on Build GRCh38
rs932220622 Chr.6: 125760942-125760942 on Build GRCh38
rs186391404 Chr.6: 125760720-125760720 on Build GRCh38

View Full Product Details