Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154323149-154323650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP10
Entrez Gene ID
89872
Gene Name
aquaporin 10
Gene Aliases
AQPA_HUMAN
UniGene
Hs.259048
Gene Chromosome Location
Chr.1: 154321059-154325325 on Build GRCh38
Transcripts
NM_080429.2 XM_011510104.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4924906 c.534G>C p.L178L Chr.1: 154323633-154323633 on Build GRCh38
4022784 c.385T>C p.Y129H Chr.1: 154323255-154323255 on Build GRCh38
3474658 c.431C>T p.S144F Chr.1: 154323301-154323301 on Build GRCh38
896567 c.444C>T p.T148T Chr.1: 154323314-154323314 on Build GRCh38
6058403 c.373G>T p.A125S Chr.1: 154323243-154323243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781413989 Chr.1: 154323611-154323611 on Build GRCh38
rs767138950 Chr.1: 154323198-154323198 on Build GRCh38
rs1015924680 Chr.1: 154323439-154323439 on Build GRCh38
rs756601568 Chr.1: 154323454-154323454 on Build GRCh38
rs536046256 Chr.1: 154323562-154323562 on Build GRCh38
rs747142402 Chr.1: 154323284-154323284 on Build GRCh38
rs201774045 Chr.1: 154323376-154323376 on Build GRCh38
rs763566273 Chr.1: 154323382-154323382 on Build GRCh38
rs562889451 Chr.1: 154323336-154323336 on Build GRCh38
rs780082562 Chr.1: 154323255-154323255 on Build GRCh38
rs770661309 Chr.1: 154323389-154323389 on Build GRCh38
rs754254481 Chr.1: 154323225-154323225 on Build GRCh38
rs749263103 Chr.1: 154323317-154323317 on Build GRCh38
rs895955199 Chr.1: 154323323-154323323 on Build GRCh38
rs767200900 Chr.1: 154323369-154323369 on Build GRCh38
rs373931873 Chr.1: 154323547-154323547 on Build GRCh38
rs1026071451 Chr.1: 154323448-154323448 on Build GRCh38
rs1043108846 Chr.1: 154323272-154323272 on Build GRCh38
rs776338550 Chr.1: 154323569-154323569 on Build GRCh38
rs542543589 Chr.1: 154323304-154323304 on Build GRCh38
rs1014428988 Chr.1: 154323325-154323325 on Build GRCh38
rs201947699 Chr.1: 154323229-154323229 on Build GRCh38
rs759248478 Chr.1: 154323168-154323168 on Build GRCh38
rs765827342 Chr.1: 154323395-154323395 on Build GRCh38
rs770847978 Chr.1: 154323321-154323321 on Build GRCh38
rs993211564 Chr.1: 154323438-154323438 on Build GRCh38
rs760289199 Chr.1: 154323565-154323565 on Build GRCh38
rs750832917 Chr.1: 154323401-154323401 on Build GRCh38
rs954525331 Chr.1: 154323464-154323464 on Build GRCh38
rs768201669 Chr.1: 154323369-154323370 on Build GRCh38
rs779460726 Chr.1: 154323228-154323228 on Build GRCh38
rs934722709 Chr.1: 154323277-154323277 on Build GRCh38
rs925975397 Chr.1: 154323568-154323568 on Build GRCh38
rs550233622 Chr.1: 154323533-154323533 on Build GRCh38
rs577422894 Chr.1: 154323219-154323219 on Build GRCh38
rs963052947 Chr.1: 154323418-154323418 on Build GRCh38
rs755209873 Chr.1: 154323604-154323604 on Build GRCh38
rs778252397 Chr.1: 154323430-154323430 on Build GRCh38
rs187770486 Chr.1: 154323408-154323408 on Build GRCh38
rs529013967 Chr.1: 154323532-154323532 on Build GRCh38
rs1045043554 Chr.1: 154323346-154323346 on Build GRCh38
rs532092647 Chr.1: 154323520-154323520 on Build GRCh38
rs768350602 Chr.1: 154323567-154323567 on Build GRCh38
rs760321077 Chr.1: 154323227-154323227 on Build GRCh38
rs371278059 Chr.1: 154323381-154323381 on Build GRCh38
rs762478363 Chr.1: 154323394-154323394 on Build GRCh38
rs753371419 Chr.1: 154323386-154323386 on Build GRCh38
rs148220488 Chr.1: 154323326-154323326 on Build GRCh38
rs751948567 Chr.1: 154323602-154323602 on Build GRCh38
rs943356980 Chr.1: 154323184-154323184 on Build GRCh38
rs748209434 Chr.1: 154323542-154323542 on Build GRCh38
rs1001658320 Chr.1: 154323282-154323282 on Build GRCh38
rs954878410 Chr.1: 154323517-154323517 on Build GRCh38
rs752776491 Chr.1: 154323616-154323616 on Build GRCh38
rs564501584 Chr.1: 154323476-154323476 on Build GRCh38
rs780343755 Chr.1: 154323407-154323407 on Build GRCh38
rs758960762 Chr.1: 154323595-154323595 on Build GRCh38
rs548507197 Chr.1: 154323578-154323578 on Build GRCh38
rs906140914 Chr.1: 154323403-154323403 on Build GRCh38
rs752262909 Chr.1: 154323202-154323202 on Build GRCh38
rs373206571 Chr.1: 154323220-154323220 on Build GRCh38
rs934672125 Chr.1: 154323589-154323589 on Build GRCh38
rs910447637 Chr.1: 154323519-154323519 on Build GRCh38
rs568743248 Chr.1: 154323539-154323539 on Build GRCh38
rs750910346 Chr.1: 154323588-154323588 on Build GRCh38
rs894944033 Chr.1: 154323318-154323318 on Build GRCh38
rs776244248 Chr.1: 154323390-154323391 on Build GRCh38
rs867034814 Chr.1: 154323350-154323350 on Build GRCh38
rs904588777 Chr.1: 154323281-154323281 on Build GRCh38
rs753161641 Chr.1: 154323251-154323251 on Build GRCh38
rs774969157 Chr.1: 154323373-154323373 on Build GRCh38
rs760257654 Chr.1: 154323378-154323378 on Build GRCh38
rs758771450 Chr.1: 154323404-154323404 on Build GRCh38
rs963092781 Chr.1: 154323473-154323473 on Build GRCh38
rs559057464 Chr.1: 154323173-154323173 on Build GRCh38
rs776750157 Chr.1: 154323312-154323312 on Build GRCh38
rs201073653 Chr.1: 154323233-154323233 on Build GRCh38
rs978755119 Chr.1: 154323540-154323540 on Build GRCh38
rs1194610 Chr.1: 154323600-154323600 on Build GRCh38
rs200551947 Chr.1: 154323332-154323332 on Build GRCh38
rs757819904 Chr.1: 154323227-154323227 on Build GRCh38
rs770864468 Chr.1: 154323546-154323546 on Build GRCh38
rs1029164906 Chr.1: 154323482-154323482 on Build GRCh38
rs182344148 Chr.1: 154323237-154323237 on Build GRCh38
rs774186017 Chr.1: 154323322-154323322 on Build GRCh38
rs771717485 Chr.1: 154323561-154323561 on Build GRCh38
rs745697594 Chr.1: 154323555-154323555 on Build GRCh38
rs566653446 Chr.1: 154323579-154323579 on Build GRCh38

View Full Product Details