Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154576088-154576593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB2
Entrez Gene ID
1141
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Gene Aliases
EFNL3, nAChRB2
UniGene
Hs.2306
Gene Chromosome Location
Chr.1: 154567781-154580026 on Build GRCh38
Transcripts
NM_000748.2 XM_017000180.1

SNP Detail

SNP ID Chromosome Location
rs200726730 Chr.1: 154576222-154576222 on Build GRCh38
rs200050649 Chr.1: 154576304-154576304 on Build GRCh38
rs201303081 Chr.1: 154576377-154576377 on Build GRCh38
rs1021704233 Chr.1: 154576447-154576447 on Build GRCh38
rs1013822758 Chr.1: 154576207-154576207 on Build GRCh38
rs2072661 Chr.1: 154576404-154576404 on Build GRCh38
rs200295099 Chr.1: 154576147-154576147 on Build GRCh38
rs199579680 Chr.1: 154576417-154576417 on Build GRCh38
rs986665401 Chr.1: 154576345-154576345 on Build GRCh38
rs896265024 Chr.1: 154576141-154576141 on Build GRCh38
rs199996766 Chr.1: 154576458-154576458 on Build GRCh38
rs772927236 Chr.1: 154576512-154576512 on Build GRCh38
rs956169135 Chr.1: 154576219-154576219 on Build GRCh38
rs185796768 Chr.1: 154576270-154576270 on Build GRCh38
rs200156184 Chr.1: 154576482-154576482 on Build GRCh38
rs918504236 Chr.1: 154576145-154576145 on Build GRCh38
rs201710469 Chr.1: 154576349-154576349 on Build GRCh38
rs914584021 Chr.1: 154576243-154576244 on Build GRCh38
rs199545188 Chr.1: 154576151-154576151 on Build GRCh38
rs201141086 Chr.1: 154576150-154576150 on Build GRCh38
rs34019399 Chr.1: 154576199-154576199 on Build GRCh38
rs199934611 Chr.1: 154576496-154576496 on Build GRCh38
rs998072335 Chr.1: 154576396-154576396 on Build GRCh38
rs200917319 Chr.1: 154576152-154576152 on Build GRCh38
rs201351693 Chr.1: 154576414-154576414 on Build GRCh38
rs78691486 Chr.1: 154576344-154576344 on Build GRCh38
rs973942512 Chr.1: 154576294-154576294 on Build GRCh38
rs116171514 Chr.1: 154576420-154576420 on Build GRCh38
rs1023990632 Chr.1: 154576215-154576215 on Build GRCh38
rs185006462 Chr.1: 154576365-154576365 on Build GRCh38
rs201694281 Chr.1: 154576156-154576156 on Build GRCh38
rs530692157 Chr.1: 154576246-154576246 on Build GRCh38
rs200573025 Chr.1: 154576175-154576175 on Build GRCh38
rs1052997239 Chr.1: 154576187-154576187 on Build GRCh38
rs773621716 Chr.1: 154576527-154576527 on Build GRCh38
rs549167761 Chr.1: 154576302-154576302 on Build GRCh38
rs200934493 Chr.1: 154576487-154576487 on Build GRCh38
rs202034594 Chr.1: 154576351-154576351 on Build GRCh38
rs200252337 Chr.1: 154576131-154576131 on Build GRCh38
rs1034102333 Chr.1: 154576254-154576254 on Build GRCh38
rs2072660 Chr.1: 154576245-154576245 on Build GRCh38
rs200457733 Chr.1: 154576533-154576533 on Build GRCh38
rs199833512 Chr.1: 154576143-154576143 on Build GRCh38
rs200933616 Chr.1: 154576501-154576501 on Build GRCh38
rs45490696 Chr.1: 154576516-154576516 on Build GRCh38
rs896199984 Chr.1: 154576299-154576299 on Build GRCh38
rs202226324 Chr.1: 154576269-154576269 on Build GRCh38
rs532932637 Chr.1: 154576180-154576180 on Build GRCh38
rs201735146 Chr.1: 154576233-154576233 on Build GRCh38
rs201796200 Chr.1: 154576495-154576495 on Build GRCh38
rs1018263199 Chr.1: 154576386-154576386 on Build GRCh38
rs749613312 Chr.1: 154576264-154576264 on Build GRCh38
rs201632996 Chr.1: 154576471-154576471 on Build GRCh38
rs988859704 Chr.1: 154576421-154576421 on Build GRCh38
rs193194163 Chr.1: 154576348-154576348 on Build GRCh38
rs200930444 Chr.1: 154576268-154576268 on Build GRCh38
rs928998183 Chr.1: 154576521-154576521 on Build GRCh38
rs904058551 Chr.1: 154576170-154576170 on Build GRCh38
rs887442230 Chr.1: 154576360-154576361 on Build GRCh38
rs188551694 Chr.1: 154576519-154576519 on Build GRCh38
rs927150974 Chr.1: 154576116-154576116 on Build GRCh38
rs199797230 Chr.1: 154576391-154576391 on Build GRCh38
rs201622270 Chr.1: 154576200-154576200 on Build GRCh38
rs570036243 Chr.1: 154576212-154576212 on Build GRCh38
rs202044279 Chr.1: 154576422-154576422 on Build GRCh38
rs968683415 Chr.1: 154576460-154576460 on Build GRCh38
rs959336366 Chr.1: 154576500-154576500 on Build GRCh38
rs4292956 Chr.1: 154576470-154576470 on Build GRCh38
rs201515151 Chr.1: 154576538-154576538 on Build GRCh38
rs1039769444 Chr.1: 154576169-154576169 on Build GRCh38
rs548833594 Chr.1: 154576265-154576265 on Build GRCh38
rs201972090 Chr.1: 154576550-154576550 on Build GRCh38
rs188760947 Chr.1: 154576343-154576343 on Build GRCh38
rs35522129 Chr.1: 154576517-154576517 on Build GRCh38
rs900779485 Chr.1: 154576394-154576394 on Build GRCh38
rs115971480 Chr.1: 154576163-154576163 on Build GRCh38
rs111787379 Chr.1: 154576204-154576204 on Build GRCh38
rs201024155 Chr.1: 154576142-154576142 on Build GRCh38

View Full Product Details