Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155052554-155053060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM15
Entrez Gene ID
8751
Gene Name
ADAM metallopeptidase domain 15
Gene Aliases
MDC15
UniGene
Hs.312098
Gene Chromosome Location
Chr.1: 155051272-155062776 on Build GRCh38
Transcripts
NM_001261464.1 NM_001261465.1 NM_001261466.1 NM_003815.4 NM_207191.2 NM_207194.2 NM_207195.2 NM_207196.2 NM_207197.2 NR_048577.1 NR_048578.1 NR_048579.1 NR_040773.1

Gene Symbol
LOC100505666
Entrez Gene ID
100505666
Gene Name
uncharacterized LOC100505666
Gene Aliases
-
UniGene
Hs.657533
Gene Chromosome Location
Chr.1: 155045191-155063991 on Build GRCh38
Transcripts
NM_001261464.1 NM_001261465.1 NM_001261466.1 NM_003815.4 NM_207191.2 NM_207194.2 NM_207195.2 NM_207196.2 NM_207197.2 NR_048577.1 NR_048578.1 NR_048579.1 NR_040773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4825118 c.154G>T p.D52Y Chr.1: 155052745-155052745 on Build GRCh38
4388832 c.93G>A p.E31E Chr.1: 155052684-155052684 on Build GRCh38
3474843 c.117C>T p.A39A Chr.1: 155052708-155052708 on Build GRCh38
1334536 c.111G>T p.E37D Chr.1: 155052702-155052702 on Build GRCh38
896751 c.87T>C p.T29T Chr.1: 155052678-155052678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773970204 Chr.1: 155052705-155052705 on Build GRCh38
rs745421787 Chr.1: 155052763-155052763 on Build GRCh38
rs752464548 Chr.1: 155052715-155052715 on Build GRCh38
rs1042712398 Chr.1: 155052867-155052867 on Build GRCh38
rs764327047 Chr.1: 155052623-155052623 on Build GRCh38
rs1026418900 Chr.1: 155052919-155052919 on Build GRCh38
rs936398494 Chr.1: 155052732-155052732 on Build GRCh38
rs987099392 Chr.1: 155052666-155052666 on Build GRCh38
rs139152568 Chr.1: 155052774-155052774 on Build GRCh38
rs918403895 Chr.1: 155052641-155052641 on Build GRCh38
rs763417047 Chr.1: 155052682-155052682 on Build GRCh38
rs770855644 Chr.1: 155052819-155052820 on Build GRCh38
rs892597335 Chr.1: 155052793-155052793 on Build GRCh38
rs766815515 Chr.1: 155052821-155052821 on Build GRCh38
rs946320988 Chr.1: 155052837-155052837 on Build GRCh38
rs770170364 Chr.1: 155052815-155052815 on Build GRCh38
rs924926242 Chr.1: 155053024-155053024 on Build GRCh38
rs963793514 Chr.1: 155052578-155052578 on Build GRCh38
rs942309361 Chr.1: 155052706-155052706 on Build GRCh38
rs565306693 Chr.1: 155052894-155052894 on Build GRCh38
rs775634486 Chr.1: 155052798-155052801 on Build GRCh38
rs760849719 Chr.1: 155052614-155052614 on Build GRCh38
rs555196210 Chr.1: 155052630-155052630 on Build GRCh38
rs993960896 Chr.1: 155052886-155052886 on Build GRCh38
rs924978926 Chr.1: 155052725-155052725 on Build GRCh38
rs569517540 Chr.1: 155052580-155052580 on Build GRCh38
rs975349669 Chr.1: 155053017-155053017 on Build GRCh38
rs575500075 Chr.1: 155052823-155052823 on Build GRCh38
rs201657685 Chr.1: 155052826-155052826 on Build GRCh38
rs1014053505 Chr.1: 155052950-155052950 on Build GRCh38
rs1019491209 Chr.1: 155052938-155052938 on Build GRCh38
rs578033720 Chr.1: 155052760-155052760 on Build GRCh38
rs377698128 Chr.1: 155052820-155052820 on Build GRCh38
rs113354502 Chr.1: 155052640-155052640 on Build GRCh38
rs751061570 Chr.1: 155052696-155052696 on Build GRCh38
rs986640083 Chr.1: 155052936-155052936 on Build GRCh38
rs777587581 Chr.1: 155052829-155052829 on Build GRCh38
rs370371360 Chr.1: 155052762-155052762 on Build GRCh38
rs369819091 Chr.1: 155053008-155053008 on Build GRCh38
rs765879414 Chr.1: 155052690-155052690 on Build GRCh38
rs867360330 Chr.1: 155052634-155052634 on Build GRCh38
rs111364353 Chr.1: 155052881-155052881 on Build GRCh38
rs140561140 Chr.1: 155052740-155052740 on Build GRCh38
rs199786202 Chr.1: 155052825-155052825 on Build GRCh38
rs775348283 Chr.1: 155052808-155052808 on Build GRCh38
rs770648729 Chr.1: 155052649-155052650 on Build GRCh38
rs2306125 Chr.1: 155052885-155052885 on Build GRCh38
rs749243962 Chr.1: 155052594-155052594 on Build GRCh38
rs58409393 Chr.1: 155052831-155052831 on Build GRCh38
rs372706086 Chr.1: 155052745-155052745 on Build GRCh38
rs535563524 Chr.1: 155052931-155052931 on Build GRCh38
rs188718790 Chr.1: 155052802-155052802 on Build GRCh38
rs200272760 Chr.1: 155052809-155052809 on Build GRCh38
rs765653167 Chr.1: 155052655-155052655 on Build GRCh38
rs904221541 Chr.1: 155052869-155052869 on Build GRCh38
rs750925838 Chr.1: 155052673-155052673 on Build GRCh38
rs772120990 Chr.1: 155052796-155052796 on Build GRCh38
rs781004867 Chr.1: 155052711-155052711 on Build GRCh38
rs773196353 Chr.1: 155052588-155052588 on Build GRCh38
rs201046872 Chr.1: 155052710-155052710 on Build GRCh38
rs950688015 Chr.1: 155052629-155052629 on Build GRCh38
rs951250047 Chr.1: 155052933-155052933 on Build GRCh38
rs772171440 Chr.1: 155052601-155052601 on Build GRCh38
rs140265124 Chr.1: 155052744-155052744 on Build GRCh38
rs537363225 Chr.1: 155052611-155052611 on Build GRCh38
rs768803750 Chr.1: 155052767-155052767 on Build GRCh38
rs951309702 Chr.1: 155053025-155053025 on Build GRCh38
rs747109064 Chr.1: 155052754-155052754 on Build GRCh38

View Full Product Details