Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155282397-155282940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCN3
Entrez Gene ID
57657
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
Gene Aliases
-
UniGene
Hs.706960
Gene Chromosome Location
Chr.1: 155277427-155289848 on Build GRCh38
Transcripts
NM_020897.2 NR_073074.1 XM_011509816.2 XM_011509817.2 XM_011509818.2 XM_017001918.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6540930 c.289G>T p.D97Y Chr.1: 155282421-155282421 on Build GRCh38
1639525 c.500G>A p.R167H Chr.1: 155282632-155282632 on Build GRCh38
5603729 c.354C>T p.F118F Chr.1: 155282486-155282486 on Build GRCh38
5626543 c.556A>G p.I186V Chr.1: 155282688-155282688 on Build GRCh38
6540931 c.628G>T p.V210F Chr.1: 155282760-155282760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756003377 Chr.1: 155282579-155282579 on Build GRCh38
rs751215604 Chr.1: 155282550-155282550 on Build GRCh38
rs899542595 Chr.1: 155282884-155282884 on Build GRCh38
rs771402332 Chr.1: 155282487-155282489 on Build GRCh38
rs144875017 Chr.1: 155282894-155282894 on Build GRCh38
rs777825053 Chr.1: 155282591-155282591 on Build GRCh38
rs747981924 Chr.1: 155282576-155282576 on Build GRCh38
rs757971773 Chr.1: 155282528-155282528 on Build GRCh38
rs757211906 Chr.1: 155282716-155282716 on Build GRCh38
rs772184733 Chr.1: 155282612-155282612 on Build GRCh38
rs987239148 Chr.1: 155282691-155282691 on Build GRCh38
rs1017947509 Chr.1: 155282805-155282805 on Build GRCh38
rs1043285006 Chr.1: 155282810-155282810 on Build GRCh38
rs781074367 Chr.1: 155282572-155282572 on Build GRCh38
rs960904131 Chr.1: 155282448-155282448 on Build GRCh38
rs371420737 Chr.1: 155282522-155282522 on Build GRCh38
rs766097977 Chr.1: 155282539-155282539 on Build GRCh38
rs371461301 Chr.1: 155282673-155282673 on Build GRCh38
rs759916758 Chr.1: 155282634-155282635 on Build GRCh38
rs140564885 Chr.1: 155282597-155282597 on Build GRCh38
rs374622488 Chr.1: 155282632-155282632 on Build GRCh38
rs367672217 Chr.1: 155282650-155282650 on Build GRCh38
rs781158679 Chr.1: 155282857-155282857 on Build GRCh38
rs760281033 Chr.1: 155282798-155282798 on Build GRCh38
rs201860832 Chr.1: 155282661-155282661 on Build GRCh38
rs773470248 Chr.1: 155282760-155282760 on Build GRCh38
rs141019470 Chr.1: 155282445-155282445 on Build GRCh38
rs767345999 Chr.1: 155282663-155282663 on Build GRCh38
rs550961408 Chr.1: 155282631-155282631 on Build GRCh38
rs754113276 Chr.1: 155282440-155282440 on Build GRCh38
rs373947209 Chr.1: 155282715-155282715 on Build GRCh38
rs201714728 Chr.1: 155282860-155282860 on Build GRCh38
rs777544371 Chr.1: 155282696-155282696 on Build GRCh38
rs746027472 Chr.1: 155282471-155282471 on Build GRCh38
rs971710679 Chr.1: 155282460-155282460 on Build GRCh38
rs752422015 Chr.1: 155282675-155282675 on Build GRCh38
rs751613252 Chr.1: 155282826-155282826 on Build GRCh38
rs115794670 Chr.1: 155282495-155282495 on Build GRCh38
rs201016236 Chr.1: 155282765-155282765 on Build GRCh38
rs1022836063 Chr.1: 155282837-155282837 on Build GRCh38
rs370726012 Chr.1: 155282845-155282845 on Build GRCh38
rs1053759494 Chr.1: 155282833-155282833 on Build GRCh38
rs772171194 Chr.1: 155282480-155282480 on Build GRCh38
rs765937571 Chr.1: 155282629-155282629 on Build GRCh38
rs763457688 Chr.1: 155282764-155282764 on Build GRCh38
rs771553056 Chr.1: 155282873-155282873 on Build GRCh38
rs201835186 Chr.1: 155282621-155282621 on Build GRCh38
rs762293040 Chr.1: 155282620-155282620 on Build GRCh38
rs905919203 Chr.1: 155282902-155282902 on Build GRCh38
rs376362078 Chr.1: 155282746-155282746 on Build GRCh38
rs1035115459 Chr.1: 155282862-155282862 on Build GRCh38
rs777004755 Chr.1: 155282507-155282507 on Build GRCh38
rs763904221 Chr.1: 155282801-155282801 on Build GRCh38
rs199850038 Chr.1: 155282642-155282642 on Build GRCh38
rs763400924 Chr.1: 155282628-155282628 on Build GRCh38
rs764405079 Chr.1: 155282497-155282497 on Build GRCh38
rs780053299 Chr.1: 155282730-155282730 on Build GRCh38
rs745914746 Chr.1: 155282606-155282606 on Build GRCh38
rs749735280 Chr.1: 155282863-155282863 on Build GRCh38
rs771587960 Chr.1: 155282745-155282745 on Build GRCh38
rs755986615 Chr.1: 155282679-155282679 on Build GRCh38
rs200499867 Chr.1: 155282859-155282859 on Build GRCh38
rs532768491 Chr.1: 155282617-155282617 on Build GRCh38
rs368133579 Chr.1: 155282741-155282741 on Build GRCh38
rs757122307 Chr.1: 155282818-155282818 on Build GRCh38
rs774853370 Chr.1: 155282488-155282490 on Build GRCh38
rs752434822 Chr.1: 155282795-155282795 on Build GRCh38
rs775743272 Chr.1: 155282489-155282489 on Build GRCh38
rs778877301 Chr.1: 155282822-155282822 on Build GRCh38
rs748591685 Chr.1: 155282625-155282625 on Build GRCh38
rs754698524 Chr.1: 155282556-155282556 on Build GRCh38
rs747187791 Chr.1: 155282615-155282615 on Build GRCh38
rs773805653 Chr.1: 155282626-155282626 on Build GRCh38
rs775582089 Chr.1: 155282614-155282614 on Build GRCh38
rs563256073 Chr.1: 155282433-155282433 on Build GRCh38
rs765836236 Chr.1: 155282514-155282514 on Build GRCh38
rs199853030 Chr.1: 155282858-155282858 on Build GRCh38
rs960299711 Chr.1: 155282462-155282462 on Build GRCh38
rs36042538 Chr.1: 155282836-155282837 on Build GRCh38
rs143660999 Chr.1: 155282562-155282562 on Build GRCh38
rs201129751 Chr.1: 155282744-155282744 on Build GRCh38
rs753591594 Chr.1: 155282815-155282815 on Build GRCh38
rs759177173 Chr.1: 155282776-155282776 on Build GRCh38
rs769786225 Chr.1: 155282465-155282465 on Build GRCh38
rs904681445 Chr.1: 155282817-155282817 on Build GRCh38
rs767257749 Chr.1: 155282792-155282792 on Build GRCh38
rs751047319 Chr.1: 155282808-155282808 on Build GRCh38
rs1005590934 Chr.1: 155282909-155282909 on Build GRCh38
rs34277242 Chr.1: 155282616-155282616 on Build GRCh38
rs770156011 Chr.1: 155282759-155282759 on Build GRCh38

View Full Product Details