Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155283891-155284390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCN3
Entrez Gene ID
57657
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
Gene Aliases
-
UniGene
Hs.706960
Gene Chromosome Location
Chr.1: 155277427-155289848 on Build GRCh38
Transcripts
NM_020897.2 NR_073074.1 XM_011509816.2 XM_011509817.2 XM_011509818.2 XM_017001918.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4222207 c.749G>A p.R250H Chr.1: 155284014-155284014 on Build GRCh38
4022989 c.751A>G p.I251V Chr.1: 155284016-155284016 on Build GRCh38
3418117 c.797G>T p.G266V Chr.1: 155284062-155284062 on Build GRCh38
6678055 c.817delC p.M274fs*17 Chr.1: 155284082-155284082 on Build GRCh38
3862911 c.748C>T p.R250C Chr.1: 155284013-155284013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566102626 Chr.1: 155284305-155284305 on Build GRCh38
rs761642949 Chr.1: 155284013-155284013 on Build GRCh38
rs1038738288 Chr.1: 155283936-155283936 on Build GRCh38
rs879739866 Chr.1: 155284291-155284291 on Build GRCh38
rs918586369 Chr.1: 155283955-155283955 on Build GRCh38
rs1048380347 Chr.1: 155283931-155283931 on Build GRCh38
rs900751562 Chr.1: 155283963-155283963 on Build GRCh38
rs1008704568 Chr.1: 155283976-155283976 on Build GRCh38
rs983584573 Chr.1: 155284224-155284224 on Build GRCh38
rs768329776 Chr.1: 155284136-155284136 on Build GRCh38
rs764958951 Chr.1: 155284145-155284145 on Build GRCh38
rs753651072 Chr.1: 155284004-155284004 on Build GRCh38
rs929380588 Chr.1: 155283921-155283921 on Build GRCh38
rs568997335 Chr.1: 155284222-155284222 on Build GRCh38
rs986895109 Chr.1: 155284367-155284367 on Build GRCh38
rs767575342 Chr.1: 155284033-155284033 on Build GRCh38
rs756353266 Chr.1: 155284046-155284046 on Build GRCh38
rs917893513 Chr.1: 155284138-155284138 on Build GRCh38
rs753496282 Chr.1: 155283986-155283987 on Build GRCh38
rs891449144 Chr.1: 155284168-155284168 on Build GRCh38
rs900376305 Chr.1: 155284072-155284072 on Build GRCh38
rs368797420 Chr.1: 155284106-155284106 on Build GRCh38
rs761479686 Chr.1: 155284142-155284142 on Build GRCh38
rs912614183 Chr.1: 155284368-155284368 on Build GRCh38
rs377307674 Chr.1: 155284040-155284040 on Build GRCh38
rs971551941 Chr.1: 155284259-155284259 on Build GRCh38
rs757736075 Chr.1: 155284080-155284080 on Build GRCh38
rs777916843 Chr.1: 155284048-155284048 on Build GRCh38
rs942251305 Chr.1: 155284289-155284289 on Build GRCh38
rs752695225 Chr.1: 155284267-155284267 on Build GRCh38
rs1051444239 Chr.1: 155284359-155284359 on Build GRCh38
rs929969804 Chr.1: 155284350-155284350 on Build GRCh38
rs930079258 Chr.1: 155283956-155283956 on Build GRCh38
rs774889651 Chr.1: 155283960-155283960 on Build GRCh38
rs114326234 Chr.1: 155284232-155284232 on Build GRCh38
rs968608221 Chr.1: 155284043-155284043 on Build GRCh38
rs927521676 Chr.1: 155283916-155283916 on Build GRCh38
rs372652695 Chr.1: 155283974-155283974 on Build GRCh38
rs369597587 Chr.1: 155283962-155283962 on Build GRCh38
rs771574816 Chr.1: 155283953-155283953 on Build GRCh38
rs746570449 Chr.1: 155284132-155284132 on Build GRCh38
rs532168007 Chr.1: 155284114-155284114 on Build GRCh38
rs1009962601 Chr.1: 155284026-155284026 on Build GRCh38
rs374487896 Chr.1: 155284175-155284193 on Build GRCh38
rs374546210 Chr.1: 155284165-155284165 on Build GRCh38
rs929280087 Chr.1: 155284221-155284221 on Build GRCh38
rs149206824 Chr.1: 155284133-155284133 on Build GRCh38
rs376804542 Chr.1: 155284141-155284141 on Build GRCh38
rs980206097 Chr.1: 155284287-155284287 on Build GRCh38
rs980342212 Chr.1: 155284052-155284052 on Build GRCh38
rs369355977 Chr.1: 155284156-155284156 on Build GRCh38
rs532857443 Chr.1: 155284089-155284089 on Build GRCh38
rs1033126731 Chr.1: 155284143-155284143 on Build GRCh38
rs368615772 Chr.1: 155284170-155284170 on Build GRCh38
rs1029969477 Chr.1: 155283972-155283972 on Build GRCh38
rs749649500 Chr.1: 155284058-155284058 on Build GRCh38
rs1023987552 Chr.1: 155284252-155284252 on Build GRCh38
rs750317134 Chr.1: 155284025-155284025 on Build GRCh38
rs762809740 Chr.1: 155284158-155284158 on Build GRCh38
rs1039180040 Chr.1: 155284304-155284304 on Build GRCh38
rs761407006 Chr.1: 155284170-155284172 on Build GRCh38
rs116597799 Chr.1: 155284276-155284276 on Build GRCh38
rs116867045 Chr.1: 155284344-155284344 on Build GRCh38
rs759087390 Chr.1: 155283937-155283937 on Build GRCh38
rs775047600 Chr.1: 155283929-155283929 on Build GRCh38
rs779452350 Chr.1: 155284105-155284105 on Build GRCh38
rs373380091 Chr.1: 155284121-155284121 on Build GRCh38
rs768010394 Chr.1: 155283985-155283986 on Build GRCh38
rs150455097 Chr.1: 155284029-155284029 on Build GRCh38
rs974336772 Chr.1: 155284343-155284343 on Build GRCh38
rs900762889 Chr.1: 155284312-155284315 on Build GRCh38
rs765123608 Chr.1: 155284023-155284023 on Build GRCh38
rs997186349 Chr.1: 155283965-155283965 on Build GRCh38

View Full Product Details