Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155339191-155339601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASH1L
Entrez Gene ID
55870
Gene Name
ASH1 like histone lysine methyltransferase
Gene Aliases
ASH1, ASH1L1, KMT2H
UniGene
Hs.491060
Gene Chromosome Location
Chr.1: 155335261-155563160 on Build GRCh38
Transcripts
NM_018489.2 XM_006711450.3 XM_006711451.3 XM_006711452.3 XM_011509770.2 XM_017001784.1 XM_017001785.1 XM_017001788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2210625 c.8501G>T p.R2834L Chr.1: 155339328-155339328 on Build GRCh38
7089616 c.8487G>T p.K2829N Chr.1: 155339342-155339342 on Build GRCh38
6605703 c.8501G>A p.R2834Q Chr.1: 155339328-155339328 on Build GRCh38
6605695 c.8469C>T p.Y2823Y Chr.1: 155339360-155339360 on Build GRCh38
896902 c.8500C>T p.R2834* Chr.1: 155339329-155339329 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753485684 Chr.1: 155339306-155339306 on Build GRCh38
rs900963908 Chr.1: 155339531-155339531 on Build GRCh38
rs372716651 Chr.1: 155339482-155339486 on Build GRCh38
rs754309681 Chr.1: 155339311-155339312 on Build GRCh38
rs940305841 Chr.1: 155339460-155339460 on Build GRCh38
rs769388354 Chr.1: 155339230-155339230 on Build GRCh38
rs915263543 Chr.1: 155339294-155339294 on Build GRCh38
rs541546996 Chr.1: 155339227-155339227 on Build GRCh38
rs371493386 Chr.1: 155339359-155339359 on Build GRCh38
rs1032185253 Chr.1: 155339558-155339558 on Build GRCh38
rs760045005 Chr.1: 155339277-155339277 on Build GRCh38
rs756976218 Chr.1: 155339309-155339309 on Build GRCh38
rs772600430 Chr.1: 155339385-155339385 on Build GRCh38
rs779899743 Chr.1: 155339318-155339318 on Build GRCh38
rs757587677 Chr.1: 155339316-155339317 on Build GRCh38
rs372607459 Chr.1: 155339289-155339289 on Build GRCh38
rs139922898 Chr.1: 155339281-155339281 on Build GRCh38
rs776386519 Chr.1: 155339509-155339509 on Build GRCh38
rs1008505954 Chr.1: 155339514-155339514 on Build GRCh38
rs889519620 Chr.1: 155339503-155339503 on Build GRCh38
rs758337819 Chr.1: 155339315-155339315 on Build GRCh38
rs930959539 Chr.1: 155339495-155339495 on Build GRCh38
rs765545973 Chr.1: 155339380-155339380 on Build GRCh38
rs183259160 Chr.1: 155339550-155339550 on Build GRCh38
rs112938330 Chr.1: 155339366-155339366 on Build GRCh38
rs888126652 Chr.1: 155339473-155339473 on Build GRCh38
rs756229863 Chr.1: 155339434-155339434 on Build GRCh38
rs954327379 Chr.1: 155339255-155339255 on Build GRCh38
rs775926452 Chr.1: 155339399-155339399 on Build GRCh38
rs775042527 Chr.1: 155339286-155339286 on Build GRCh38
rs751186013 Chr.1: 155339384-155339384 on Build GRCh38
rs945348235 Chr.1: 155339311-155339311 on Build GRCh38
rs954330502 Chr.1: 155339570-155339570 on Build GRCh38
rs748257840 Chr.1: 155339328-155339328 on Build GRCh38
rs749444214 Chr.1: 155339342-155339342 on Build GRCh38
rs1003910303 Chr.1: 155339508-155339508 on Build GRCh38
rs574154438 Chr.1: 155339212-155339212 on Build GRCh38
rs367682239 Chr.1: 155339351-155339351 on Build GRCh38
rs756247812 Chr.1: 155339329-155339329 on Build GRCh38
rs376071723 Chr.1: 155339402-155339402 on Build GRCh38
rs966639277 Chr.1: 155339257-155339258 on Build GRCh38
rs188881460 Chr.1: 155339251-155339251 on Build GRCh38
rs765084257 Chr.1: 155339314-155339314 on Build GRCh38
rs370789867 Chr.1: 155339288-155339288 on Build GRCh38
rs948669439 Chr.1: 155339427-155339427 on Build GRCh38
rs553735283 Chr.1: 155339371-155339371 on Build GRCh38
rs149742311 Chr.1: 155339533-155339533 on Build GRCh38
rs763833102 Chr.1: 155339292-155339292 on Build GRCh38
rs776228501 Chr.1: 155339360-155339360 on Build GRCh38
rs567702147 Chr.1: 155339518-155339518 on Build GRCh38
rs1057000069 Chr.1: 155339387-155339387 on Build GRCh38
rs981622189 Chr.1: 155339431-155339431 on Build GRCh38
rs193273160 Chr.1: 155339337-155339337 on Build GRCh38
rs763632417 Chr.1: 155339409-155339409 on Build GRCh38
rs754719148 Chr.1: 155339405-155339405 on Build GRCh38

View Full Product Details