Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155344013-155344511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASH1L
Entrez Gene ID
55870
Gene Name
ASH1 like histone lysine methyltransferase
Gene Aliases
ASH1, ASH1L1, KMT2H
UniGene
Hs.491060
Gene Chromosome Location
Chr.1: 155335261-155563160 on Build GRCh38
Transcripts
NM_018489.2 XM_006711450.3 XM_006711451.3 XM_006711452.3 XM_011509770.2 XM_017001784.1 XM_017001785.1 XM_017001788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6605716 c.7940A>G p.Y2647C Chr.1: 155344224-155344224 on Build GRCh38
6459388 c.7901T>A p.M2634K Chr.1: 155344263-155344263 on Build GRCh38
5377844 c.7905C>T p.I2635I Chr.1: 155344259-155344259 on Build GRCh38
7251852 c.7891-10C>T p.? Chr.1: 155344283-155344283 on Build GRCh38
4626835 c.7914C>T p.P2638P Chr.1: 155344250-155344250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772130740 Chr.1: 155344321-155344321 on Build GRCh38
rs1037757184 Chr.1: 155344264-155344264 on Build GRCh38
rs533889862 Chr.1: 155344045-155344045 on Build GRCh38
rs780858038 Chr.1: 155344133-155344133 on Build GRCh38
rs901649982 Chr.1: 155344405-155344405 on Build GRCh38
rs1051146508 Chr.1: 155344360-155344360 on Build GRCh38
rs1047090743 Chr.1: 155344353-155344353 on Build GRCh38
rs931324978 Chr.1: 155344350-155344350 on Build GRCh38
rs893038511 Chr.1: 155344462-155344462 on Build GRCh38
rs754185073 Chr.1: 155344205-155344205 on Build GRCh38
rs772416513 Chr.1: 155344179-155344179 on Build GRCh38
rs1004758138 Chr.1: 155344086-155344086 on Build GRCh38
rs769820371 Chr.1: 155344303-155344303 on Build GRCh38
rs369466292 Chr.1: 155344189-155344189 on Build GRCh38
rs780995608 Chr.1: 155344296-155344296 on Build GRCh38
rs754440380 Chr.1: 155344436-155344436 on Build GRCh38
rs773522157 Chr.1: 155344193-155344193 on Build GRCh38
rs181628419 Chr.1: 155344048-155344048 on Build GRCh38
rs766734750 Chr.1: 155344278-155344278 on Build GRCh38
rs538103112 Chr.1: 155344366-155344366 on Build GRCh38
rs749932376 Chr.1: 155344239-155344239 on Build GRCh38
rs748063150 Chr.1: 155344143-155344143 on Build GRCh38
rs751159490 Chr.1: 155344271-155344271 on Build GRCh38
rs890727055 Chr.1: 155344385-155344385 on Build GRCh38
rs754622167 Chr.1: 155344281-155344281 on Build GRCh38
rs1032761596 Chr.1: 155344449-155344449 on Build GRCh38
rs765040911 Chr.1: 155344488-155344488 on Build GRCh38
rs556422925 Chr.1: 155344367-155344367 on Build GRCh38
rs762286819 Chr.1: 155344396-155344396 on Build GRCh38
rs745926333 Chr.1: 155344317-155344317 on Build GRCh38
rs982120543 Chr.1: 155344083-155344083 on Build GRCh38
rs757964056 Chr.1: 155344245-155344245 on Build GRCh38
rs767840436 Chr.1: 155344088-155344088 on Build GRCh38
rs748001710 Chr.1: 155344302-155344302 on Build GRCh38
rs999938264 Chr.1: 155344411-155344411 on Build GRCh38
rs774546860 Chr.1: 155344155-155344155 on Build GRCh38
rs116211337 Chr.1: 155344112-155344112 on Build GRCh38
rs762285533 Chr.1: 155344206-155344206 on Build GRCh38
rs996010357 Chr.1: 155344132-155344132 on Build GRCh38
rs868594014 Chr.1: 155344442-155344442 on Build GRCh38
rs949392108 Chr.1: 155344074-155344074 on Build GRCh38
rs1011238480 Chr.1: 155344464-155344464 on Build GRCh38
rs1028896125 Chr.1: 155344446-155344446 on Build GRCh38
rs1019912152 Chr.1: 155344392-155344392 on Build GRCh38
rs769871383 Chr.1: 155344144-155344144 on Build GRCh38
rs928754131 Chr.1: 155344311-155344311 on Build GRCh38
rs759727262 Chr.1: 155344165-155344165 on Build GRCh38
rs762218071 Chr.1: 155344057-155344057 on Build GRCh38
rs917158000 Chr.1: 155344042-155344042 on Build GRCh38
rs765774025 Chr.1: 155344230-155344230 on Build GRCh38
rs937592591 Chr.1: 155344316-155344316 on Build GRCh38
rs368843735 Chr.1: 155344256-155344256 on Build GRCh38
rs1019185506 Chr.1: 155344110-155344110 on Build GRCh38
rs201394353 Chr.1: 155344188-155344188 on Build GRCh38
rs920161122 Chr.1: 155344322-155344322 on Build GRCh38
rs944085817 Chr.1: 155344386-155344386 on Build GRCh38
rs777921369 Chr.1: 155344314-155344314 on Build GRCh38

View Full Product Details