Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125972167-125972573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HINT3
Entrez Gene ID
135114
Gene Name
histidine triad nucleotide binding protein 3
Gene Aliases
HINT4
UniGene
Hs.72325
Gene Chromosome Location
Chr.6: 125956715-125980244 on Build GRCh38
Transcripts
NM_138571.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3858051 c.332T>C p.V111A Chr.6: 125972271-125972271 on Build GRCh38
1072949 c.359G>T p.R120I Chr.6: 125972298-125972298 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774046468 Chr.6: 125972304-125972304 on Build GRCh38
rs748237915 Chr.6: 125972261-125972261 on Build GRCh38
rs749563161 Chr.6: 125972283-125972283 on Build GRCh38
rs984233891 Chr.6: 125972509-125972509 on Build GRCh38
rs145582221 Chr.6: 125972507-125972507 on Build GRCh38
rs756166289 Chr.6: 125972363-125972363 on Build GRCh38
rs748697632 Chr.6: 125972362-125972362 on Build GRCh38
rs769916214 Chr.6: 125972276-125972276 on Build GRCh38
rs558027241 Chr.6: 125972400-125972400 on Build GRCh38
rs147966956 Chr.6: 125972493-125972493 on Build GRCh38
rs546178562 Chr.6: 125972372-125972372 on Build GRCh38
rs763734703 Chr.6: 125972322-125972322 on Build GRCh38
rs796910042 Chr.6: 125972549-125972549 on Build GRCh38
rs372560513 Chr.6: 125972282-125972282 on Build GRCh38
rs145594277 Chr.6: 125972288-125972288 on Build GRCh38
rs751870175 Chr.6: 125972335-125972335 on Build GRCh38
rs199906226 Chr.6: 125972319-125972319 on Build GRCh38
rs370489985 Chr.6: 125972333-125972333 on Build GRCh38
rs986701038 Chr.6: 125972385-125972385 on Build GRCh38
rs747544630 Chr.6: 125972345-125972346 on Build GRCh38
rs1024261877 Chr.6: 125972414-125972414 on Build GRCh38
rs781178315 Chr.6: 125972231-125972231 on Build GRCh38
rs895609362 Chr.6: 125972275-125972275 on Build GRCh38
rs539526998 Chr.6: 125972396-125972396 on Build GRCh38
rs139464016 Chr.6: 125972196-125972196 on Build GRCh38
rs576302987 Chr.6: 125972433-125972433 on Build GRCh38
rs868069359 Chr.6: 125972257-125972257 on Build GRCh38
rs144919432 Chr.6: 125972311-125972311 on Build GRCh38
rs769070558 Chr.6: 125972221-125972221 on Build GRCh38
rs954934088 Chr.6: 125972379-125972379 on Build GRCh38
rs905275353 Chr.6: 125972294-125972294 on Build GRCh38
rs377239838 Chr.6: 125972328-125972328 on Build GRCh38
rs373220582 Chr.6: 125972365-125972365 on Build GRCh38
rs781652784 Chr.6: 125972360-125972360 on Build GRCh38
rs774138084 Chr.6: 125972373-125972373 on Build GRCh38
rs531411056 Chr.6: 125972369-125972369 on Build GRCh38
rs755452295 Chr.6: 125972353-125972353 on Build GRCh38
rs979974454 Chr.6: 125972483-125972483 on Build GRCh38
rs147740313 Chr.6: 125972315-125972315 on Build GRCh38
rs756880654 Chr.6: 125972331-125972331 on Build GRCh38
rs553463639 Chr.6: 125972320-125972320 on Build GRCh38

View Full Product Details