Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125974666-125975177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HINT3
Entrez Gene ID
135114
Gene Name
histidine triad nucleotide binding protein 3
Gene Aliases
HINT4
UniGene
Hs.72325
Gene Chromosome Location
Chr.6: 125956715-125980244 on Build GRCh38
Transcripts
NM_138571.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4495791 c.460C>T p.Q154* Chr.6: 125974917-125974917 on Build GRCh38
3620163 c.510T>C p.F170F Chr.6: 125974967-125974967 on Build GRCh38
4690328 c.426C>A p.S142S Chr.6: 125974883-125974883 on Build GRCh38
6864119 c.487_488delTA p.Y163fs*1 Chr.6: 125974944-125974945 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768324743 Chr.6: 125974907-125974908 on Build GRCh38
rs768715314 Chr.6: 125974945-125974945 on Build GRCh38
rs750561303 Chr.6: 125975024-125975024 on Build GRCh38
rs914721199 Chr.6: 125975149-125975149 on Build GRCh38
rs967122719 Chr.6: 125975151-125975152 on Build GRCh38
rs746161433 Chr.6: 125974899-125974899 on Build GRCh38
rs757254631 Chr.6: 125975003-125975003 on Build GRCh38
rs767922040 Chr.6: 125974863-125974863 on Build GRCh38
rs557368100 Chr.6: 125974786-125974786 on Build GRCh38
rs184832630 Chr.6: 125975065-125975065 on Build GRCh38
rs762691620 Chr.6: 125974961-125974961 on Build GRCh38
rs968782678 Chr.6: 125974923-125974923 on Build GRCh38
rs1037993845 Chr.6: 125975064-125975064 on Build GRCh38
rs544917261 Chr.6: 125975058-125975058 on Build GRCh38
rs577650419 Chr.6: 125974954-125974954 on Build GRCh38
rs370635400 Chr.6: 125974852-125974852 on Build GRCh38
rs142669902 Chr.6: 125974847-125974847 on Build GRCh38
rs1047387743 Chr.6: 125975129-125975129 on Build GRCh38
rs779000488 Chr.6: 125974918-125974918 on Build GRCh38
rs761212828 Chr.6: 125974951-125974951 on Build GRCh38
rs760773184 Chr.6: 125974993-125974993 on Build GRCh38
rs761113453 Chr.6: 125974994-125974994 on Build GRCh38
rs75747983 Chr.6: 125975105-125975105 on Build GRCh38
rs755291518 Chr.6: 125974858-125974858 on Build GRCh38
rs145709677 Chr.6: 125974850-125974850 on Build GRCh38
rs757619798 Chr.6: 125974802-125974802 on Build GRCh38
rs756648725 Chr.6: 125974882-125974882 on Build GRCh38
rs948884210 Chr.6: 125975044-125975044 on Build GRCh38
rs569157088 Chr.6: 125974827-125974827 on Build GRCh38
rs754436073 Chr.6: 125975001-125975001 on Build GRCh38
rs771967946 Chr.6: 125974936-125974936 on Build GRCh38
rs780017028 Chr.6: 125974939-125974939 on Build GRCh38
rs943963042 Chr.6: 125974878-125974878 on Build GRCh38
rs1017295338 Chr.6: 125975153-125975153 on Build GRCh38
rs749341560 Chr.6: 125974893-125974893 on Build GRCh38
rs373946404 Chr.6: 125975133-125975133 on Build GRCh38
rs528150632 Chr.6: 125975145-125975145 on Build GRCh38
rs990297407 Chr.6: 125974711-125974711 on Build GRCh38
rs753156772 Chr.6: 125974868-125974868 on Build GRCh38
rs771971264 Chr.6: 125974818-125974818 on Build GRCh38
rs559175785 Chr.6: 125974940-125974940 on Build GRCh38
rs767832192 Chr.6: 125974992-125974992 on Build GRCh38
rs1026763507 Chr.6: 125974980-125974980 on Build GRCh38
rs202125498 Chr.6: 125974972-125974972 on Build GRCh38
rs865974355 Chr.6: 125974840-125974840 on Build GRCh38
rs776144718 Chr.6: 125974947-125974947 on Build GRCh38
rs149760356 Chr.6: 125974849-125974849 on Build GRCh38
rs530430801 Chr.6: 125975103-125975103 on Build GRCh38
rs536604153 Chr.6: 125974834-125974834 on Build GRCh38
rs763648744 Chr.6: 125974826-125974826 on Build GRCh38
rs745581270 Chr.6: 125974729-125974730 on Build GRCh38
rs775536519 Chr.6: 125974822-125974822 on Build GRCh38
rs890209085 Chr.6: 125974705-125974705 on Build GRCh38
rs1017409069 Chr.6: 125974736-125974736 on Build GRCh38
rs981846701 Chr.6: 125975114-125975114 on Build GRCh38
rs938769061 Chr.6: 125974982-125974982 on Build GRCh38
rs756881997 Chr.6: 125974813-125974850 on Build GRCh38
rs777764486 Chr.6: 125974887-125974887 on Build GRCh38
rs772720084 Chr.6: 125974957-125974957 on Build GRCh38
rs779097059 Chr.6: 125975004-125975004 on Build GRCh38
rs200985274 Chr.6: 125974839-125974839 on Build GRCh38
rs376235645 Chr.6: 125974836-125974836 on Build GRCh38
rs764572487 Chr.6: 125975000-125975000 on Build GRCh38
rs750146321 Chr.6: 125974838-125974838 on Build GRCh38
rs1008561405 Chr.6: 125974727-125974727 on Build GRCh38
rs757417568 Chr.6: 125974911-125974911 on Build GRCh38
rs745639091 Chr.6: 125974815-125974815 on Build GRCh38
rs148947145 Chr.6: 125974934-125974934 on Build GRCh38

View Full Product Details