Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 137143730-137144233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL22RA2
Entrez Gene ID
116379
Gene Name
interleukin 22 receptor subunit alpha 2
Gene Aliases
CRF2-10, CRF2-S1, CRF2X, IL-22BP, IL-22R-alpha-2, IL-22RA2, ZCYTOR16
UniGene
Hs.126891
Gene Chromosome Location
Chr.6: 137143820-137173648 on Build GRCh38
Transcripts
NM_052962.2 NM_181309.1 NM_181310.1

SNP Detail

SNP ID Chromosome Location
rs956077075 Chr.6: 137143931-137143931 on Build GRCh38
rs992406232 Chr.6: 137144092-137144092 on Build GRCh38
rs748470084 Chr.6: 137143904-137143904 on Build GRCh38
rs1031895210 Chr.6: 137144178-137144178 on Build GRCh38
rs968613241 Chr.6: 137143849-137143849 on Build GRCh38
rs956129521 Chr.6: 137144200-137144203 on Build GRCh38
rs202566 Chr.6: 137143937-137143937 on Build GRCh38
rs944836878 Chr.6: 137143898-137143898 on Build GRCh38
rs202565 Chr.6: 137143779-137143779 on Build GRCh38
rs781711389 Chr.6: 137143896-137143896 on Build GRCh38
rs774600004 Chr.6: 137144083-137144083 on Build GRCh38
rs918795879 Chr.6: 137143775-137143775 on Build GRCh38
rs1051441060 Chr.6: 137144086-137144086 on Build GRCh38
rs143852774 Chr.6: 137144146-137144146 on Build GRCh38
rs773889596 Chr.6: 137144085-137144085 on Build GRCh38
rs533773093 Chr.6: 137143977-137143977 on Build GRCh38
rs149489220 Chr.6: 137143861-137143861 on Build GRCh38
rs375675212 Chr.6: 137144161-137144161 on Build GRCh38
rs575819254 Chr.6: 137144173-137144173 on Build GRCh38
rs28362176 Chr.6: 137143947-137143947 on Build GRCh38
rs1031756607 Chr.6: 137143900-137143900 on Build GRCh38
rs530070165 Chr.6: 137143944-137143944 on Build GRCh38
rs1007627981 Chr.6: 137144115-137144115 on Build GRCh38
rs777221684 Chr.6: 137143992-137143992 on Build GRCh38
rs930243225 Chr.6: 137143778-137143778 on Build GRCh38
rs544329873 Chr.6: 137144201-137144201 on Build GRCh38
rs1000039285 Chr.6: 137144148-137144148 on Build GRCh38
rs974304690 Chr.6: 137143756-137143756 on Build GRCh38
rs866706506 Chr.6: 137143974-137143974 on Build GRCh38
rs1017923748 Chr.6: 137143839-137143839 on Build GRCh38
rs752060022 Chr.6: 137143806-137143806 on Build GRCh38
rs949806141 Chr.6: 137143859-137143859 on Build GRCh38
rs916918055 Chr.6: 137144109-137144109 on Build GRCh38
rs370388465 Chr.6: 137144040-137144040 on Build GRCh38
rs41288973 Chr.6: 137144143-137144143 on Build GRCh38
rs962853635 Chr.6: 137144047-137144047 on Build GRCh38
rs573851119 Chr.6: 137143864-137143864 on Build GRCh38
rs1018280520 Chr.6: 137144036-137144036 on Build GRCh38
rs189910063 Chr.6: 137144056-137144056 on Build GRCh38
rs576668500 Chr.6: 137144124-137144124 on Build GRCh38
rs890148368 Chr.6: 137144105-137144105 on Build GRCh38
rs1010145483 Chr.6: 137143951-137143951 on Build GRCh38
rs949669587 Chr.6: 137144113-137144113 on Build GRCh38
rs770403172 Chr.6: 137144058-137144058 on Build GRCh38
rs1007453843 Chr.6: 137143781-137143781 on Build GRCh38
rs9494668 Chr.6: 137143998-137143998 on Build GRCh38

View Full Product Details