Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 137158260-137158753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL22RA2
Entrez Gene ID
116379
Gene Name
interleukin 22 receptor subunit alpha 2
Gene Aliases
CRF2-10, CRF2-S1, CRF2X, IL-22BP, IL-22R-alpha-2, IL-22RA2, ZCYTOR16
UniGene
Hs.126891
Gene Chromosome Location
Chr.6: 137143820-137173648 on Build GRCh38
Transcripts
NM_052962.2 NM_181309.1 NM_181310.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5404450 c.91_92CC>TT p.P31F Chr.6: 137158452-137158453 on Build GRCh38
1440700 c.121T>G p.F41V Chr.6: 137158423-137158423 on Build GRCh38
5599696 c.150G>T p.G50G Chr.6: 137158394-137158394 on Build GRCh38
7372065 c.158T>C p.L53P Chr.6: 137158386-137158386 on Build GRCh38
6685237 c.94C>G p.Q32E Chr.6: 137158450-137158450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533102602 Chr.6: 137158606-137158606 on Build GRCh38
rs185208638 Chr.6: 137158543-137158543 on Build GRCh38
rs961596355 Chr.6: 137158691-137158691 on Build GRCh38
rs953800732 Chr.6: 137158661-137158661 on Build GRCh38
rs780627214 Chr.6: 137158425-137158425 on Build GRCh38
rs764997279 Chr.6: 137158316-137158316 on Build GRCh38
rs189150070 Chr.6: 137158575-137158575 on Build GRCh38
rs374860051 Chr.6: 137158469-137158469 on Build GRCh38
rs758393708 Chr.6: 137158576-137158576 on Build GRCh38
rs746469094 Chr.6: 137158337-137158337 on Build GRCh38
rs550031620 Chr.6: 137158662-137158662 on Build GRCh38
rs946904815 Chr.6: 137158517-137158517 on Build GRCh38
rs200411020 Chr.6: 137158509-137158509 on Build GRCh38
rs376997317 Chr.6: 137158507-137158507 on Build GRCh38
rs568586290 Chr.6: 137158702-137158702 on Build GRCh38
rs1031319174 Chr.6: 137158356-137158356 on Build GRCh38
rs533484155 Chr.6: 137158487-137158487 on Build GRCh38
rs946799791 Chr.6: 137158721-137158721 on Build GRCh38
rs761705086 Chr.6: 137158392-137158392 on Build GRCh38
rs757070092 Chr.6: 137158313-137158313 on Build GRCh38
rs767227640 Chr.6: 137158514-137158514 on Build GRCh38
rs193043200 Chr.6: 137158498-137158498 on Build GRCh38
rs760680350 Chr.6: 137158388-137158388 on Build GRCh38
rs765051005 Chr.6: 137158393-137158393 on Build GRCh38
rs137884605 Chr.6: 137158705-137158705 on Build GRCh38
rs1028112134 Chr.6: 137158649-137158649 on Build GRCh38
rs764102281 Chr.6: 137158493-137158493 on Build GRCh38
rs772427516 Chr.6: 137158444-137158444 on Build GRCh38
rs761985099 Chr.6: 137158490-137158490 on Build GRCh38
rs753723324 Chr.6: 137158307-137158307 on Build GRCh38
rs774589687 Chr.6: 137158282-137158282 on Build GRCh38
rs750146939 Chr.6: 137158396-137158396 on Build GRCh38
rs999750370 Chr.6: 137158559-137158559 on Build GRCh38
rs780249958 Chr.6: 137158468-137158468 on Build GRCh38
rs1011070723 Chr.6: 137158327-137158327 on Build GRCh38
rs117006380 Chr.6: 137158695-137158695 on Build GRCh38
rs754351808 Chr.6: 137158428-137158428 on Build GRCh38
rs775700387 Chr.6: 137158387-137158387 on Build GRCh38
rs201767019 Chr.6: 137158405-137158405 on Build GRCh38
rs746169416 Chr.6: 137158431-137158431 on Build GRCh38
rs754151454 Chr.6: 137158616-137158616 on Build GRCh38
rs772042407 Chr.6: 137158385-137158385 on Build GRCh38
rs35962884 Chr.6: 137158352-137158352 on Build GRCh38
rs140545983 Chr.6: 137158404-137158404 on Build GRCh38
rs761065579 Chr.6: 137158470-137158470 on Build GRCh38
rs758167056 Chr.6: 137158324-137158324 on Build GRCh38
rs1021055518 Chr.6: 137158583-137158583 on Build GRCh38
rs976466601 Chr.6: 137158398-137158398 on Build GRCh38
rs201560036 Chr.6: 137158429-137158429 on Build GRCh38
rs772229715 Chr.6: 137158386-137158386 on Build GRCh38
rs1052780377 Chr.6: 137158605-137158605 on Build GRCh38
rs776612446 Chr.6: 137158484-137158484 on Build GRCh38
rs776124172 Chr.6: 137158448-137158448 on Build GRCh38
rs375007805 Chr.6: 137158494-137158494 on Build GRCh38
rs145385106 Chr.6: 137158666-137158666 on Build GRCh38
rs967114790 Chr.6: 137158601-137158601 on Build GRCh38
rs548699649 Chr.6: 137158351-137158351 on Build GRCh38
rs376298751 Chr.6: 137158406-137158406 on Build GRCh38
rs537997575 Chr.6: 137158318-137158318 on Build GRCh38
rs574793113 Chr.6: 137158397-137158397 on Build GRCh38
rs140270270 Chr.6: 137158544-137158544 on Build GRCh38
rs199542741 Chr.6: 137158390-137158390 on Build GRCh38
rs895278467 Chr.6: 137158317-137158317 on Build GRCh38
rs563707552 Chr.6: 137158588-137158588 on Build GRCh38
rs759631943 Chr.6: 137158287-137158287 on Build GRCh38
rs959970249 Chr.6: 137158625-137158627 on Build GRCh38
rs866315107 Chr.6: 137158496-137158496 on Build GRCh38
rs1055176279 Chr.6: 137158513-137158513 on Build GRCh38
rs115602108 Chr.6: 137158504-137158504 on Build GRCh38
rs556514846 Chr.6: 137158500-137158500 on Build GRCh38
rs374224961 Chr.6: 137158377-137158377 on Build GRCh38
rs916675809 Chr.6: 137158712-137158712 on Build GRCh38
rs563385239 Chr.6: 137158663-137158663 on Build GRCh38
rs28362851 Chr.6: 137158492-137158492 on Build GRCh38
rs1048273738 Chr.6: 137158674-137158674 on Build GRCh38
rs371156597 Chr.6: 137158342-137158342 on Build GRCh38
rs558752550 Chr.6: 137158409-137158409 on Build GRCh38
rs369995086 Chr.6: 137158706-137158706 on Build GRCh38
rs1055227864 Chr.6: 137158294-137158294 on Build GRCh38
rs779884183 Chr.6: 137158334-137158334 on Build GRCh38
rs77622675 Chr.6: 137158467-137158467 on Build GRCh38
rs371564041 Chr.6: 137158531-137158531 on Build GRCh38
rs771317080 Chr.6: 137158373-137158373 on Build GRCh38

View Full Product Details