Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138209800-138210343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGEF3
Entrez Gene ID
57221
Gene Name
ARFGEF family member 3
Gene Aliases
A7322, BIG3, C6orf92, KIAA1244, PPP1R33, dJ171N11.1
UniGene
Hs.194408
Gene Chromosome Location
Chr.6: 138161690-138344663 on Build GRCh38
Transcripts
NM_020340.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5533297 c.132G>A p.V44V Chr.6: 138210035-138210035 on Build GRCh38
3621046 c.259G>A p.D87N Chr.6: 138209949-138209949 on Build GRCh38
3858576 c.296A>G p.N99S Chr.6: 138209986-138209986 on Build GRCh38
3858578 c.324C>T p.N108N Chr.6: 138210014-138210014 on Build GRCh38
4586256 c.321C>T p.L107L Chr.6: 138210011-138210011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760092112 Chr.6: 138209875-138209875 on Build GRCh38
rs376156596 Chr.6: 138209860-138209860 on Build GRCh38
rs747743897 Chr.6: 138210082-138210082 on Build GRCh38
rs928084535 Chr.6: 138210127-138210127 on Build GRCh38
rs569789439 Chr.6: 138210137-138210137 on Build GRCh38
rs573802241 Chr.6: 138209884-138209884 on Build GRCh38
rs879799598 Chr.6: 138210289-138210289 on Build GRCh38
rs183881992 Chr.6: 138209990-138209990 on Build GRCh38
rs141094520 Chr.6: 138209955-138209955 on Build GRCh38
rs371306054 Chr.6: 138210069-138210069 on Build GRCh38
rs566070452 Chr.6: 138210176-138210176 on Build GRCh38
rs772861520 Chr.6: 138210003-138210003 on Build GRCh38
rs906502530 Chr.6: 138209891-138209891 on Build GRCh38
rs956781105 Chr.6: 138210309-138210309 on Build GRCh38
rs192996208 Chr.6: 138209870-138209870 on Build GRCh38
rs979491124 Chr.6: 138210296-138210296 on Build GRCh38
rs144337526 Chr.6: 138210038-138210038 on Build GRCh38
rs968558412 Chr.6: 138209999-138209999 on Build GRCh38
rs769362995 Chr.6: 138210002-138210002 on Build GRCh38
rs765759246 Chr.6: 138210235-138210235 on Build GRCh38
rs191153062 Chr.6: 138210284-138210284 on Build GRCh38
rs894379509 Chr.6: 138210162-138210162 on Build GRCh38
rs764341526 Chr.6: 138209917-138209917 on Build GRCh38
rs765246285 Chr.6: 138210050-138210050 on Build GRCh38
rs199788147 Chr.6: 138210053-138210053 on Build GRCh38
rs117405205 Chr.6: 138210219-138210219 on Build GRCh38
rs139587659 Chr.6: 138210174-138210174 on Build GRCh38
rs201650935 Chr.6: 138209872-138209872 on Build GRCh38
rs765264174 Chr.6: 138210139-138210139 on Build GRCh38
rs768398718 Chr.6: 138209984-138209984 on Build GRCh38
rs1003505046 Chr.6: 138209901-138209901 on Build GRCh38
rs549839332 Chr.6: 138210128-138210128 on Build GRCh38
rs1037415414 Chr.6: 138210168-138210168 on Build GRCh38
rs1018707071 Chr.6: 138210287-138210287 on Build GRCh38
rs370635068 Chr.6: 138209970-138209970 on Build GRCh38
rs779745367 Chr.6: 138209876-138209877 on Build GRCh38
rs775689172 Chr.6: 138210008-138210008 on Build GRCh38
rs904324302 Chr.6: 138210266-138210266 on Build GRCh38
rs758267584 Chr.6: 138210065-138210065 on Build GRCh38
rs910027541 Chr.6: 138210060-138210060 on Build GRCh38
rs147619292 Chr.6: 138210161-138210161 on Build GRCh38
rs762031354 Chr.6: 138210048-138210048 on Build GRCh38
rs897574266 Chr.6: 138210190-138210190 on Build GRCh38
rs145271719 Chr.6: 138210011-138210011 on Build GRCh38
rs377139969 Chr.6: 138209960-138209960 on Build GRCh38
rs752358350 Chr.6: 138210080-138210080 on Build GRCh38
rs993363612 Chr.6: 138210212-138210212 on Build GRCh38
rs1045147600 Chr.6: 138210122-138210122 on Build GRCh38
rs745868206 Chr.6: 138209861-138209862 on Build GRCh38
rs767714804 Chr.6: 138209879-138209879 on Build GRCh38
rs776248190 Chr.6: 138209863-138209863 on Build GRCh38
rs769160281 Chr.6: 138209866-138209872 on Build GRCh38
rs558073952 Chr.6: 138210236-138210236 on Build GRCh38
rs750428295 Chr.6: 138210181-138210181 on Build GRCh38
rs758372554 Chr.6: 138209963-138209963 on Build GRCh38
rs149485557 Chr.6: 138210033-138210033 on Build GRCh38
rs776217445 Chr.6: 138210027-138210027 on Build GRCh38
rs138636575 Chr.6: 138209921-138209921 on Build GRCh38
rs746857899 Chr.6: 138209981-138209981 on Build GRCh38
rs115941727 Chr.6: 138210014-138210014 on Build GRCh38
rs200375307 Chr.6: 138209865-138209865 on Build GRCh38
rs757279376 Chr.6: 138209933-138209933 on Build GRCh38
rs745417623 Chr.6: 138210064-138210064 on Build GRCh38
rs1012734319 Chr.6: 138209982-138209982 on Build GRCh38
rs940192568 Chr.6: 138210068-138210068 on Build GRCh38
rs779000006 Chr.6: 138209951-138209951 on Build GRCh38
rs188823726 Chr.6: 138210103-138210103 on Build GRCh38
rs780939417 Chr.6: 138209989-138209989 on Build GRCh38
rs116456650 Chr.6: 138209843-138209843 on Build GRCh38
rs368629110 Chr.6: 138210077-138210077 on Build GRCh38
rs774959426 Chr.6: 138209873-138209873 on Build GRCh38
rs772700641 Chr.6: 138210007-138210007 on Build GRCh38
rs760805381 Chr.6: 138209894-138209894 on Build GRCh38
rs754521960 Chr.6: 138210274-138210274 on Build GRCh38
rs1057039362 Chr.6: 138209918-138209918 on Build GRCh38
rs751472322 Chr.6: 138210073-138210073 on Build GRCh38
rs372581008 Chr.6: 138210264-138210264 on Build GRCh38
rs534914100 Chr.6: 138209855-138209855 on Build GRCh38

View Full Product Details