Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143427865-143428410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAT2
Entrez Gene ID
134637
Gene Name
adenosine deaminase, tRNA specific 2
Gene Aliases
DEADC1, TAD2, dJ20N2, dJ20N2.1
UniGene
Hs.709561
Gene Chromosome Location
Chr.6: 143422832-143450750 on Build GRCh38
Transcripts
NM_001286259.1 NM_182503.2 XM_011535439.2 XM_011535440.1 XM_011535444.2 XM_017010260.1 XM_017010261.1 XM_017010262.1 XM_017010263.1

SNP Detail

SNP ID Chromosome Location
rs183286560 Chr.6: 143428337-143428337 on Build GRCh38
rs962458704 Chr.6: 143428086-143428086 on Build GRCh38
rs1065355 Chr.6: 143428158-143428158 on Build GRCh38
rs936802293 Chr.6: 143427935-143427935 on Build GRCh38
rs900742509 Chr.6: 143427970-143427970 on Build GRCh38
rs966699484 Chr.6: 143428192-143428192 on Build GRCh38
rs556567231 Chr.6: 143428047-143428047 on Build GRCh38
rs181502929 Chr.6: 143427990-143427990 on Build GRCh38
rs565516170 Chr.6: 143428268-143428268 on Build GRCh38
rs545697444 Chr.6: 143428222-143428222 on Build GRCh38
rs1012886977 Chr.6: 143428327-143428327 on Build GRCh38
rs895396371 Chr.6: 143427953-143427953 on Build GRCh38
rs1042891932 Chr.6: 143428141-143428141 on Build GRCh38
rs971186262 Chr.6: 143428001-143428001 on Build GRCh38
rs908734067 Chr.6: 143428124-143428124 on Build GRCh38
rs113136631 Chr.6: 143427945-143427945 on Build GRCh38
rs768999493 Chr.6: 143428155-143428155 on Build GRCh38
rs1033549005 Chr.6: 143428097-143428097 on Build GRCh38
rs1039216490 Chr.6: 143427947-143427947 on Build GRCh38
rs373733449 Chr.6: 143428082-143428082 on Build GRCh38
rs973730181 Chr.6: 143428081-143428081 on Build GRCh38
rs187581567 Chr.6: 143428383-143428383 on Build GRCh38
rs759282153 Chr.6: 143428070-143428072 on Build GRCh38
rs943873875 Chr.6: 143428113-143428113 on Build GRCh38
rs959531580 Chr.6: 143428148-143428148 on Build GRCh38
rs536656464 Chr.6: 143427918-143427918 on Build GRCh38
rs532652759 Chr.6: 143427911-143427911 on Build GRCh38
rs917947619 Chr.6: 143427913-143427913 on Build GRCh38
rs954059938 Chr.6: 143428084-143428084 on Build GRCh38
rs1000805909 Chr.6: 143427930-143427930 on Build GRCh38
rs925377852 Chr.6: 143427929-143427929 on Build GRCh38
rs978065492 Chr.6: 143428210-143428210 on Build GRCh38
rs936790865 Chr.6: 143428272-143428272 on Build GRCh38
rs190080800 Chr.6: 143428289-143428289 on Build GRCh38
rs749406652 Chr.6: 143428026-143428026 on Build GRCh38
rs912356409 Chr.6: 143428112-143428112 on Build GRCh38
rs376223175 Chr.6: 143428023-143428023 on Build GRCh38
rs917837478 Chr.6: 143428170-143428170 on Build GRCh38
rs985330301 Chr.6: 143428084-143428084 on Build GRCh38
rs1049992762 Chr.6: 143427899-143427900 on Build GRCh38
rs570258152 Chr.6: 143428162-143428162 on Build GRCh38
rs995337942 Chr.6: 143428096-143428096 on Build GRCh38
rs1012423029 Chr.6: 143427966-143427966 on Build GRCh38
rs895403079 Chr.6: 143428302-143428302 on Build GRCh38
rs1051185225 Chr.6: 143428004-143428004 on Build GRCh38
rs1004300181 Chr.6: 143428024-143428024 on Build GRCh38
rs997808152 Chr.6: 143427997-143427997 on Build GRCh38
rs959359105 Chr.6: 143427903-143427903 on Build GRCh38
rs186145080 Chr.6: 143428237-143428237 on Build GRCh38
rs6933612 Chr.6: 143427925-143427925 on Build GRCh38

View Full Product Details