Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 145626280-145626778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPM2A
Entrez Gene ID
7957
Gene Name
epilepsy, progressive myoclonus type 2A, Lafora disease (laforin)
Gene Aliases
EPM2, MELF
UniGene
Hs.486696
Gene Chromosome Location
Chr.6: 145500744-145736018 on Build GRCh38
Transcripts
NM_001018041.1 NM_005670.3 XM_006715564.3 XM_011536113.2 XM_011536116.1 XM_017011300.1 XM_017011301.1 XM_017011302.1

SNP Detail

SNP ID Chromosome Location
rs895165125 Chr.6: 145626532-145626532 on Build GRCh38
rs933057620 Chr.6: 145626708-145626708 on Build GRCh38
rs568826881 Chr.6: 145626548-145626548 on Build GRCh38
rs1037578576 Chr.6: 145626505-145626505 on Build GRCh38
rs570596325 Chr.6: 145626635-145626635 on Build GRCh38
rs1034148959 Chr.6: 145626530-145626530 on Build GRCh38
rs886386587 Chr.6: 145626478-145626478 on Build GRCh38
rs775757205 Chr.6: 145626345-145626345 on Build GRCh38
rs1047513417 Chr.6: 145626620-145626620 on Build GRCh38
rs1032515259 Chr.6: 145626608-145626608 on Build GRCh38
rs945379250 Chr.6: 145626518-145626518 on Build GRCh38
rs903758987 Chr.6: 145626576-145626576 on Build GRCh38
rs61095394 Chr.6: 145626456-145626456 on Build GRCh38
rs532554007 Chr.6: 145626651-145626651 on Build GRCh38
rs1016373044 Chr.6: 145626652-145626652 on Build GRCh38
rs60283049 Chr.6: 145626488-145626488 on Build GRCh38
rs184781317 Chr.6: 145626710-145626710 on Build GRCh38
rs41285847 Chr.6: 145626412-145626412 on Build GRCh38
rs982991896 Chr.6: 145626320-145626320 on Build GRCh38
rs921450966 Chr.6: 145626429-145626429 on Build GRCh38
rs546842567 Chr.6: 145626748-145626748 on Build GRCh38
rs1013306053 Chr.6: 145626615-145626615 on Build GRCh38
rs370728607 Chr.6: 145626490-145626490 on Build GRCh38
rs898953077 Chr.6: 145626519-145626519 on Build GRCh38
rs1054941722 Chr.6: 145626607-145626607 on Build GRCh38
rs1024970272 Chr.6: 145626543-145626543 on Build GRCh38
rs936474695 Chr.6: 145626393-145626393 on Build GRCh38
rs925437652 Chr.6: 145626374-145626374 on Build GRCh38
rs79253382 Chr.6: 145626743-145626743 on Build GRCh38
rs1028458063 Chr.6: 145626694-145626694 on Build GRCh38
rs966936388 Chr.6: 145626365-145626365 on Build GRCh38
rs1005882091 Chr.6: 145626638-145626638 on Build GRCh38
rs560480535 Chr.6: 145626650-145626650 on Build GRCh38
rs564401326 Chr.6: 145626309-145626309 on Build GRCh38
rs201975659 Chr.6: 145626579-145626580 on Build GRCh38
rs140540200 Chr.6: 145626308-145626308 on Build GRCh38
rs1013612983 Chr.6: 145626533-145626533 on Build GRCh38
rs117758752 Chr.6: 145626668-145626668 on Build GRCh38
rs978749252 Chr.6: 145626598-145626598 on Build GRCh38
rs551071462 Chr.6: 145626674-145626674 on Build GRCh38
rs1025205827 Chr.6: 145626359-145626359 on Build GRCh38
rs147291746 Chr.6: 145626578-145626579 on Build GRCh38
rs1051650373 Chr.6: 145626469-145626469 on Build GRCh38
rs974860542 Chr.6: 145626408-145626408 on Build GRCh38
rs191801098 Chr.6: 145626540-145626540 on Build GRCh38
rs966987390 Chr.6: 145626592-145626592 on Build GRCh38
rs753525886 Chr.6: 145626744-145626744 on Build GRCh38
rs995610493 Chr.6: 145626529-145626529 on Build GRCh38
rs540474350 Chr.6: 145626415-145626415 on Build GRCh38
rs763107324 Chr.6: 145626385-145626385 on Build GRCh38
rs766207516 Chr.6: 145626720-145626720 on Build GRCh38
rs978315720 Chr.6: 145626369-145626369 on Build GRCh38

View Full Product Details