Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151468825-151469322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMT1
Entrez Gene ID
79624
Gene Name
acidic residue methyltransferase 1
Gene Aliases
C6orf211
UniGene
Hs.15929
Gene Chromosome Location
Chr.6: 151452258-151470101 on Build GRCh38
Transcripts
NM_001286562.1 NM_024573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3019390 c.1057T>C p.Y353H Chr.6: 151468841-151468841 on Build GRCh38
3859153 c.1282A>G p.T428A Chr.6: 151469066-151469066 on Build GRCh38
5881264 c.1187C>G p.P396R Chr.6: 151468971-151468971 on Build GRCh38
3859154 c.1295A>G p.Y432C Chr.6: 151469079-151469079 on Build GRCh38
4917799 c.1159C>G p.Q387E Chr.6: 151468943-151468943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766688917 Chr.6: 151468973-151468973 on Build GRCh38
rs367760436 Chr.6: 151469100-151469100 on Build GRCh38
rs1052346025 Chr.6: 151469164-151469164 on Build GRCh38
rs536097951 Chr.6: 151469282-151469282 on Build GRCh38
rs756737330 Chr.6: 151469095-151469095 on Build GRCh38
rs1041330454 Chr.6: 151469222-151469222 on Build GRCh38
rs974670209 Chr.6: 151469249-151469249 on Build GRCh38
rs745472699 Chr.6: 151469007-151469007 on Build GRCh38
rs558614819 Chr.6: 151468878-151468878 on Build GRCh38
rs756757031 Chr.6: 151468991-151468991 on Build GRCh38
rs767698565 Chr.6: 151468968-151468968 on Build GRCh38
rs748184329 Chr.6: 151468879-151468879 on Build GRCh38
rs754347404 Chr.6: 151468974-151468974 on Build GRCh38
rs753223657 Chr.6: 151469084-151469084 on Build GRCh38
rs758870663 Chr.6: 151469295-151469295 on Build GRCh38
rs1024374299 Chr.6: 151468923-151468923 on Build GRCh38
rs374775847 Chr.6: 151469026-151469026 on Build GRCh38
rs748584179 Chr.6: 151469199-151469199 on Build GRCh38
rs1034881399 Chr.6: 151469245-151469245 on Build GRCh38
rs563031382 Chr.6: 151469180-151469180 on Build GRCh38
rs1045945469 Chr.6: 151469168-151469168 on Build GRCh38
rs527808498 Chr.6: 151469235-151469235 on Build GRCh38
rs770544009 Chr.6: 151468874-151468874 on Build GRCh38
rs754257764 Chr.6: 151469064-151469064 on Build GRCh38
rs151335520 Chr.6: 151468868-151468868 on Build GRCh38
rs972757664 Chr.6: 151469134-151469134 on Build GRCh38
rs374663926 Chr.6: 151468879-151468879 on Build GRCh38
rs888974908 Chr.6: 151469290-151469290 on Build GRCh38
rs1010319520 Chr.6: 151469211-151469211 on Build GRCh38
rs755774649 Chr.6: 151469102-151469102 on Build GRCh38
rs749163784 Chr.6: 151469158-151469158 on Build GRCh38
rs768772545 Chr.6: 151469020-151469020 on Build GRCh38
rs149658464 Chr.6: 151468990-151468990 on Build GRCh38
rs142746071 Chr.6: 151468936-151468936 on Build GRCh38
rs140681772 Chr.6: 151468961-151468961 on Build GRCh38
rs780753822 Chr.6: 151468997-151468997 on Build GRCh38
rs551736160 Chr.6: 151469201-151469201 on Build GRCh38
rs753042203 Chr.6: 151469237-151469237 on Build GRCh38
rs184190412 Chr.6: 151469166-151469166 on Build GRCh38
rs779458832 Chr.6: 151468984-151468984 on Build GRCh38
rs749255188 Chr.6: 151469019-151469019 on Build GRCh38
rs1054643038 Chr.6: 151469194-151469194 on Build GRCh38
rs755419809 Chr.6: 151468978-151468978 on Build GRCh38
rs892882635 Chr.6: 151469200-151469200 on Build GRCh38
rs760817421 Chr.6: 151469047-151469047 on Build GRCh38
rs201674567 Chr.6: 151468970-151468970 on Build GRCh38
rs140057628 Chr.6: 151468967-151468967 on Build GRCh38
rs571697633 Chr.6: 151469208-151469208 on Build GRCh38
rs200502959 Chr.6: 151469018-151469018 on Build GRCh38
rs774240808 Chr.6: 151468964-151468964 on Build GRCh38
rs929625641 Chr.6: 151468999-151468999 on Build GRCh38
rs774341693 Chr.6: 151469024-151469024 on Build GRCh38
rs892818372 Chr.6: 151468956-151468956 on Build GRCh38
rs372873070 Chr.6: 151468982-151468982 on Build GRCh38
rs765699732 Chr.6: 151469071-151469071 on Build GRCh38
rs988836237 Chr.6: 151468921-151468921 on Build GRCh38
rs754772438 Chr.6: 151469159-151469159 on Build GRCh38
rs759887047 Chr.6: 151469066-151469066 on Build GRCh38
rs962897028 Chr.6: 151469247-151469247 on Build GRCh38
rs775593762 Chr.6: 151468948-151468948 on Build GRCh38
rs762946533 Chr.6: 151468949-151468949 on Build GRCh38
rs773335371 Chr.6: 151469041-151469041 on Build GRCh38
rs971486366 Chr.6: 151468926-151468926 on Build GRCh38
rs918822880 Chr.6: 151469280-151469280 on Build GRCh38
rs1010213754 Chr.6: 151469191-151469191 on Build GRCh38
rs567337322 Chr.6: 151469275-151469275 on Build GRCh38
rs779843355 Chr.6: 151469152-151469152 on Build GRCh38
rs148839278 Chr.6: 151469246-151469246 on Build GRCh38
rs765653105 Chr.6: 151469028-151469028 on Build GRCh38
rs892397745 Chr.6: 151469165-151469165 on Build GRCh38
rs779160643 Chr.6: 151469060-151469060 on Build GRCh38
rs531946381 Chr.6: 151469193-151469193 on Build GRCh38
rs766744055 Chr.6: 151469054-151469054 on Build GRCh38
rs151094814 Chr.6: 151469096-151469096 on Build GRCh38
rs145468544 Chr.6: 151469209-151469209 on Build GRCh38

View Full Product Details