Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151469589-151470128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMT1
Entrez Gene ID
79624
Gene Name
acidic residue methyltransferase 1
Gene Aliases
C6orf211
UniGene
Hs.15929
Gene Chromosome Location
Chr.6: 151452258-151470101 on Build GRCh38
Transcripts
NM_001286562.1 NM_024573.2

SNP Detail

SNP ID Chromosome Location
rs898544875 Chr.6: 151470007-151470007 on Build GRCh38
rs987009119 Chr.6: 151469625-151469625 on Build GRCh38
rs949234733 Chr.6: 151469885-151469885 on Build GRCh38
rs1056213330 Chr.6: 151469955-151469955 on Build GRCh38
rs1016065309 Chr.6: 151469736-151469736 on Build GRCh38
rs576842619 Chr.6: 151470006-151470006 on Build GRCh38
rs1046334090 Chr.6: 151469899-151469899 on Build GRCh38
rs936427922 Chr.6: 151469830-151469830 on Build GRCh38
rs963666869 Chr.6: 151469683-151469683 on Build GRCh38
rs138110574 Chr.6: 151469647-151469647 on Build GRCh38
rs370812141 Chr.6: 151469703-151469703 on Build GRCh38
rs757372791 Chr.6: 151469888-151469888 on Build GRCh38
rs778375773 Chr.6: 151469741-151469741 on Build GRCh38
rs192394710 Chr.6: 151469702-151469702 on Build GRCh38
rs914720341 Chr.6: 151470009-151470009 on Build GRCh38
rs543298260 Chr.6: 151469948-151469948 on Build GRCh38
rs776464777 Chr.6: 151469943-151469943 on Build GRCh38
rs568709220 Chr.6: 151470103-151470103 on Build GRCh38
rs994231749 Chr.6: 151469778-151469778 on Build GRCh38
rs778468057 Chr.6: 151469617-151469617 on Build GRCh38
rs913735772 Chr.6: 151469864-151469864 on Build GRCh38
rs889036941 Chr.6: 151469682-151469682 on Build GRCh38
rs987955137 Chr.6: 151469862-151469862 on Build GRCh38
rs990734795 Chr.6: 151470004-151470004 on Build GRCh38
rs563173150 Chr.6: 151469950-151469950 on Build GRCh38
rs1025718385 Chr.6: 151469790-151469790 on Build GRCh38
rs554517750 Chr.6: 151469701-151469701 on Build GRCh38
rs925030480 Chr.6: 151469786-151469786 on Build GRCh38
rs545560829 Chr.6: 151470016-151470016 on Build GRCh38
rs971419039 Chr.6: 151469799-151469799 on Build GRCh38
rs1034602674 Chr.6: 151469915-151469915 on Build GRCh38
rs905134616 Chr.6: 151469942-151469942 on Build GRCh38
rs977169341 Chr.6: 151470045-151470048 on Build GRCh38
rs751964104 Chr.6: 151469860-151469860 on Build GRCh38
rs565359611 Chr.6: 151470029-151470029 on Build GRCh38

View Full Product Details