Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 166366878-166367374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPC1
Entrez Gene ID
51660
Gene Name
mitochondrial pyruvate carrier 1
Gene Aliases
BRP44L, CGI-129, MPYCD, dJ68L15.3
UniGene
Hs.172755
Gene Chromosome Location
Chr.6: 166364919-166383013 on Build GRCh38
Transcripts
NM_001270879.1 NM_016098.3 NR_073093.1 XM_011535894.1 XM_011535895.1

SNP Detail

SNP ID Chromosome Location
rs149799643 Chr.6: 166367126-166367126 on Build GRCh38
rs996355806 Chr.6: 166367121-166367121 on Build GRCh38
rs756865188 Chr.6: 166367013-166367013 on Build GRCh38
rs1014907475 Chr.6: 166367198-166367198 on Build GRCh38
rs774020255 Chr.6: 166366918-166366918 on Build GRCh38
rs1036560251 Chr.6: 166367308-166367308 on Build GRCh38
rs891564985 Chr.6: 166367036-166367036 on Build GRCh38
rs140110900 Chr.6: 166367002-166367002 on Build GRCh38
rs1004909539 Chr.6: 166367195-166367195 on Build GRCh38
rs1032473001 Chr.6: 166366998-166366998 on Build GRCh38
rs201621198 Chr.6: 166366991-166366991 on Build GRCh38
rs954928338 Chr.6: 166367169-166367169 on Build GRCh38
rs963467500 Chr.6: 166367215-166367215 on Build GRCh38
rs546635527 Chr.6: 166367042-166367042 on Build GRCh38
rs767143374 Chr.6: 166366918-166366919 on Build GRCh38
rs979948888 Chr.6: 166367244-166367244 on Build GRCh38
rs772199908 Chr.6: 166366981-166366981 on Build GRCh38
rs919724038 Chr.6: 166366933-166366933 on Build GRCh38
rs749353762 Chr.6: 166366940-166366940 on Build GRCh38
rs938381094 Chr.6: 166367008-166367008 on Build GRCh38
rs755347066 Chr.6: 166366956-166366956 on Build GRCh38
rs1031034294 Chr.6: 166367165-166367165 on Build GRCh38
rs938579824 Chr.6: 166367277-166367277 on Build GRCh38
rs970330852 Chr.6: 166367236-166367236 on Build GRCh38
rs534142863 Chr.6: 166366982-166366982 on Build GRCh38
rs991588772 Chr.6: 166367265-166367265 on Build GRCh38
rs746221035 Chr.6: 166366906-166366906 on Build GRCh38
rs774877313 Chr.6: 166367041-166367041 on Build GRCh38
rs774608175 Chr.6: 166366954-166366954 on Build GRCh38
rs997753811 Chr.6: 166366950-166366950 on Build GRCh38
rs556720411 Chr.6: 166367301-166367301 on Build GRCh38
rs933578919 Chr.6: 166367324-166367324 on Build GRCh38
rs756637240 Chr.6: 166366913-166366913 on Build GRCh38
rs776132314 Chr.6: 166367062-166367062 on Build GRCh38
rs754200686 Chr.6: 166366936-166366936 on Build GRCh38
rs751081220 Chr.6: 166367010-166367010 on Build GRCh38
rs973449135 Chr.6: 166367223-166367223 on Build GRCh38
rs919406451 Chr.6: 166367321-166367321 on Build GRCh38
rs763544588 Chr.6: 166367007-166367007 on Build GRCh38
rs1043239822 Chr.6: 166367045-166367045 on Build GRCh38
rs1051452161 Chr.6: 166366969-166366969 on Build GRCh38
rs750041596 Chr.6: 166366919-166366920 on Build GRCh38
rs970648823 Chr.6: 166367076-166367076 on Build GRCh38
rs909326095 Chr.6: 166367274-166367274 on Build GRCh38
rs567129578 Chr.6: 166366900-166366900 on Build GRCh38
rs528565212 Chr.6: 166366990-166366990 on Build GRCh38
rs780761474 Chr.6: 166366926-166366926 on Build GRCh38
rs749640399 Chr.6: 166367180-166367180 on Build GRCh38
rs900629676 Chr.6: 166367107-166367107 on Build GRCh38
rs748356133 Chr.6: 166366978-166366978 on Build GRCh38
rs181692044 Chr.6: 166367332-166367332 on Build GRCh38
rs958919942 Chr.6: 166367255-166367255 on Build GRCh38
rs6913353 Chr.6: 166366993-166366993 on Build GRCh38
rs779380917 Chr.6: 166366965-166366969 on Build GRCh38
rs1020738105 Chr.6: 166367022-166367022 on Build GRCh38
rs768893003 Chr.6: 166366943-166366943 on Build GRCh38
rs1052703662 Chr.6: 166367019-166367019 on Build GRCh38
rs760402541 Chr.6: 166366924-166366924 on Build GRCh38
rs762419969 Chr.6: 166367006-166367006 on Build GRCh38
rs1043239417 Chr.6: 166367342-166367342 on Build GRCh38
rs766616061 Chr.6: 166366929-166366933 on Build GRCh38
rs912865915 Chr.6: 166367296-166367296 on Build GRCh38
rs1052732195 Chr.6: 166367284-166367284 on Build GRCh38
rs16898896 Chr.6: 166367233-166367233 on Build GRCh38
rs928568651 Chr.6: 166367268-166367268 on Build GRCh38
rs574888578 Chr.6: 166367312-166367312 on Build GRCh38

View Full Product Details