Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 166382658-166383158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPC1
Entrez Gene ID
51660
Gene Name
mitochondrial pyruvate carrier 1
Gene Aliases
BRP44L, CGI-129, MPYCD, dJ68L15.3
UniGene
Hs.172755
Gene Chromosome Location
Chr.6: 166364919-166383013 on Build GRCh38
Transcripts
NM_001270879.1 NM_016098.3 NR_073093.1

SNP Detail

SNP ID Chromosome Location
rs998699944 Chr.6: 166382989-166382989 on Build GRCh38
rs1017456576 Chr.6: 166383011-166383011 on Build GRCh38
rs965669882 Chr.6: 166382924-166382924 on Build GRCh38
rs936782332 Chr.6: 166383114-166383114 on Build GRCh38
rs938034650 Chr.6: 166382810-166382810 on Build GRCh38
rs777827116 Chr.6: 166382850-166382850 on Build GRCh38
rs567798756 Chr.6: 166382865-166382865 on Build GRCh38
rs781449430 Chr.6: 166382760-166382760 on Build GRCh38
rs931139792 Chr.6: 166383016-166383017 on Build GRCh38
rs562563891 Chr.6: 166383109-166383109 on Build GRCh38
rs767692880 Chr.6: 166382836-166382836 on Build GRCh38
rs550593206 Chr.6: 166382716-166382716 on Build GRCh38
rs944497057 Chr.6: 166382731-166382731 on Build GRCh38
rs376218888 Chr.6: 166382908-166382908 on Build GRCh38
rs760787392 Chr.6: 166383035-166383035 on Build GRCh38
rs556570579 Chr.6: 166382920-166382920 on Build GRCh38
rs866223452 Chr.6: 166382968-166382968 on Build GRCh38
rs182458694 Chr.6: 166383076-166383076 on Build GRCh38
rs1049335304 Chr.6: 166382745-166382745 on Build GRCh38
rs9459659 Chr.6: 166382754-166382754 on Build GRCh38
rs370510209 Chr.6: 166382885-166382885 on Build GRCh38
rs756227158 Chr.6: 166382906-166382906 on Build GRCh38
rs892960094 Chr.6: 166383000-166383000 on Build GRCh38
rs995343627 Chr.6: 166382877-166382877 on Build GRCh38
rs763301295 Chr.6: 166382809-166382809 on Build GRCh38
rs886476318 Chr.6: 166382895-166382895 on Build GRCh38
rs929504943 Chr.6: 166382947-166382947 on Build GRCh38
rs562340889 Chr.6: 166382734-166382734 on Build GRCh38
rs760329969 Chr.6: 166382784-166382784 on Build GRCh38
rs751623493 Chr.6: 166382822-166382822 on Build GRCh38
rs1007917322 Chr.6: 166383006-166383006 on Build GRCh38
rs767531439 Chr.6: 166382898-166382898 on Build GRCh38
rs764484310 Chr.6: 166382886-166382886 on Build GRCh38
rs140927897 Chr.6: 166382993-166382993 on Build GRCh38
rs765478642 Chr.6: 166382796-166382796 on Build GRCh38
rs1042515305 Chr.6: 166382985-166382985 on Build GRCh38
rs577965165 Chr.6: 166382927-166382927 on Build GRCh38
rs776472438 Chr.6: 166382793-166382793 on Build GRCh38
rs750600635 Chr.6: 166382838-166382838 on Build GRCh38
rs984554789 Chr.6: 166383053-166383053 on Build GRCh38
rs979575161 Chr.6: 166383121-166383121 on Build GRCh38
rs538989340 Chr.6: 166382942-166382942 on Build GRCh38
rs965701078 Chr.6: 166383027-166383027 on Build GRCh38
rs770608276 Chr.6: 166382868-166382868 on Build GRCh38
rs950804669 Chr.6: 166382681-166382681 on Build GRCh38
rs1029537717 Chr.6: 166382930-166382930 on Build GRCh38
rs757373242 Chr.6: 166382830-166382830 on Build GRCh38
rs775713787 Chr.6: 166382878-166382878 on Build GRCh38
rs777850458 Chr.6: 166382771-166382772 on Build GRCh38
rs985330657 Chr.6: 166382948-166382948 on Build GRCh38
rs373243456 Chr.6: 166382765-166382765 on Build GRCh38
rs768378219 Chr.6: 166382772-166382772 on Build GRCh38
rs910302834 Chr.6: 166382722-166382722 on Build GRCh38
rs887013196 Chr.6: 166383056-166383056 on Build GRCh38
rs950480216 Chr.6: 166383047-166383047 on Build GRCh38
rs759949701 Chr.6: 166382794-166382794 on Build GRCh38
rs375956827 Chr.6: 166382801-166382801 on Build GRCh38
rs1017258000 Chr.6: 166382919-166382919 on Build GRCh38
rs1546854 Chr.6: 166382945-166382945 on Build GRCh38
rs777823374 Chr.6: 166382914-166382914 on Build GRCh38
rs957072301 Chr.6: 166382709-166382709 on Build GRCh38
rs146480117 Chr.6: 166382988-166382988 on Build GRCh38
rs73265159 Chr.6: 166382738-166382738 on Build GRCh38
rs756377719 Chr.6: 166382842-166382842 on Build GRCh38
rs1053859895 Chr.6: 166383115-166383115 on Build GRCh38
rs11557065 Chr.6: 166382837-166382837 on Build GRCh38
rs79498685 Chr.6: 166382847-166382847 on Build GRCh38
rs879330704 Chr.6: 166382956-166382956 on Build GRCh38
rs938302905 Chr.6: 166382958-166382958 on Build GRCh38
rs112458095 Chr.6: 166383059-166383059 on Build GRCh38
rs769491045 Chr.6: 166382870-166382870 on Build GRCh38
rs772152939 Chr.6: 166383030-166383030 on Build GRCh38
rs746906037 Chr.6: 166382915-166382915 on Build GRCh38
rs372957606 Chr.6: 166382894-166382894 on Build GRCh38
rs925845930 Chr.6: 166382676-166382676 on Build GRCh38
rs747983938 Chr.6: 166382848-166382848 on Build GRCh38
rs572382575 Chr.6: 166382971-166382971 on Build GRCh38
rs35617576 Chr.6: 166382985-166382985 on Build GRCh38
rs764728090 Chr.6: 166382819-166382819 on Build GRCh38
rs919454872 Chr.6: 166382737-166382737 on Build GRCh38
rs551398633 Chr.6: 166382755-166382755 on Build GRCh38
rs757282363 Chr.6: 166382918-166382918 on Build GRCh38
rs745470711 Chr.6: 166382869-166382869 on Build GRCh38
rs1026029334 Chr.6: 166383034-166383034 on Build GRCh38
rs745449909 Chr.6: 166382926-166382926 on Build GRCh38
rs1042370832 Chr.6: 166383081-166383081 on Build GRCh38
rs918151451 Chr.6: 166383066-166383066 on Build GRCh38
rs766596398 Chr.6: 166383061-166383061 on Build GRCh38
rs762336418 Chr.6: 166382896-166382896 on Build GRCh38
rs202222055 Chr.6: 166382789-166382789 on Build GRCh38
rs766694413 Chr.6: 166382907-166382907 on Build GRCh38
rs566742239 Chr.6: 166382771-166382771 on Build GRCh38
rs898435477 Chr.6: 166382876-166382876 on Build GRCh38
rs377468342 Chr.6: 166382679-166382679 on Build GRCh38
rs1032934940 Chr.6: 166382899-166382899 on Build GRCh38
rs923960904 Chr.6: 166383102-166383102 on Build GRCh38
rs749057502 Chr.6: 166382762-166382762 on Build GRCh38
rs949348113 Chr.6: 166382981-166382981 on Build GRCh38
rs747983025 Chr.6: 166382774-166382774 on Build GRCh38
rs758266163 Chr.6: 166382911-166382911 on Build GRCh38
rs934978925 Chr.6: 166382780-166382780 on Build GRCh38
rs924189517 Chr.6: 166382952-166382952 on Build GRCh38
rs909469279 Chr.6: 166382953-166382953 on Build GRCh38
rs573449898 Chr.6: 166383063-166383063 on Build GRCh38
rs909406026 Chr.6: 166383060-166383060 on Build GRCh38
rs975826380 Chr.6: 166382929-166382929 on Build GRCh38

View Full Product Details