Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 549553-550046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKAR1B
Entrez Gene ID
5575
Gene Name
protein kinase cAMP-dependent type I regulatory subunit beta
Gene Aliases
PRKAR1
UniGene
Hs.520851
Gene Chromosome Location
Chr.7: 549185-727676 on Build GRCh38
Transcripts
NM_001164758.1 NM_001164759.1 NM_001164760.1 NM_001164761.1 NM_001164762.1 NM_002735.2 XM_011515446.2 XM_017012433.1 XM_017012434.1

SNP Detail

SNP ID Chromosome Location
rs984673072 Chr.7: 549916-549916 on Build GRCh38
rs1023495279 Chr.7: 549851-549851 on Build GRCh38
rs910373955 Chr.7: 549730-549730 on Build GRCh38
rs947623883 Chr.7: 549779-549779 on Build GRCh38
rs982298714 Chr.7: 549725-549725 on Build GRCh38
rs543515772 Chr.7: 549720-549720 on Build GRCh38
rs576184054 Chr.7: 549672-549672 on Build GRCh38
rs566902090 Chr.7: 549889-549889 on Build GRCh38
rs888991800 Chr.7: 549622-549622 on Build GRCh38
rs371476930 Chr.7: 549939-549943 on Build GRCh38
rs34512801 Chr.7: 549584-549584 on Build GRCh38
rs541408134 Chr.7: 549726-549726 on Build GRCh38
rs150643000 Chr.7: 549926-549926 on Build GRCh38
rs1004882343 Chr.7: 549687-549687 on Build GRCh38
rs771487869 Chr.7: 549828-549828 on Build GRCh38
rs10620 Chr.7: 549885-549885 on Build GRCh38
rs1042328600 Chr.7: 549776-549776 on Build GRCh38
rs975013593 Chr.7: 549894-549894 on Build GRCh38
rs1052319739 Chr.7: 549814-549814 on Build GRCh38
rs970506468 Chr.7: 549721-549721 on Build GRCh38
rs2763 Chr.7: 550023-550023 on Build GRCh38
rs1036048623 Chr.7: 549579-549579 on Build GRCh38
rs1051945452 Chr.7: 549922-549922 on Build GRCh38
rs1007636000 Chr.7: 549643-549643 on Build GRCh38
rs1010681718 Chr.7: 550020-550020 on Build GRCh38
rs183771995 Chr.7: 549905-549905 on Build GRCh38
rs1003837887 Chr.7: 549975-549975 on Build GRCh38
rs989598893 Chr.7: 549888-549888 on Build GRCh38
rs561383120 Chr.7: 549749-549749 on Build GRCh38
rs375253316 Chr.7: 549960-549960 on Build GRCh38
rs539290575 Chr.7: 549963-549963 on Build GRCh38
rs374629404 Chr.7: 549744-549744 on Build GRCh38
rs11545043 Chr.7: 549793-549793 on Build GRCh38
rs984426929 Chr.7: 549936-549943 on Build GRCh38
rs886517616 Chr.7: 549827-549827 on Build GRCh38
rs914859878 Chr.7: 549777-549777 on Build GRCh38
rs66461018 Chr.7: 549936-549940 on Build GRCh38
rs900646614 Chr.7: 549671-549671 on Build GRCh38
rs547027832 Chr.7: 549874-549874 on Build GRCh38
rs566007836 Chr.7: 549859-549859 on Build GRCh38
rs899147823 Chr.7: 549847-549847 on Build GRCh38
rs540381017 Chr.7: 549578-549578 on Build GRCh38
rs543874990 Chr.7: 549842-549842 on Build GRCh38
rs1025562042 Chr.7: 549913-549913 on Build GRCh38
rs1030402204 Chr.7: 549680-549680 on Build GRCh38
rs572474119 Chr.7: 549718-549718 on Build GRCh38
rs1000799377 Chr.7: 549887-549887 on Build GRCh38
rs1012185379 Chr.7: 549948-549948 on Build GRCh38
rs1026092727 Chr.7: 549933-549933 on Build GRCh38
rs865839668 Chr.7: 549803-549803 on Build GRCh38
rs1011789007 Chr.7: 549613-549613 on Build GRCh38
rs758722266 Chr.7: 549602-549602 on Build GRCh38
rs940804661 Chr.7: 549927-549927 on Build GRCh38
rs963272086 Chr.7: 549714-549714 on Build GRCh38
rs1040575790 Chr.7: 549871-549871 on Build GRCh38
rs922174861 Chr.7: 549897-549897 on Build GRCh38
rs1017287060 Chr.7: 549937-549937 on Build GRCh38
rs765637116 Chr.7: 549719-549720 on Build GRCh38
rs893386664 Chr.7: 549606-549606 on Build GRCh38
rs532727909 Chr.7: 549850-549850 on Build GRCh38
rs923088010 Chr.7: 549967-549967 on Build GRCh38
rs956724008 Chr.7: 549994-549994 on Build GRCh38
rs1014144169 Chr.7: 549823-549823 on Build GRCh38
rs200788744 Chr.7: 549583-549583 on Build GRCh38
rs565502679 Chr.7: 549834-549835 on Build GRCh38
rs935000897 Chr.7: 549589-549589 on Build GRCh38
rs906453349 Chr.7: 549974-549974 on Build GRCh38
rs971878258 Chr.7: 549836-549836 on Build GRCh38
rs527308417 Chr.7: 549872-549872 on Build GRCh38
rs989154357 Chr.7: 549765-549765 on Build GRCh38
rs918919055 Chr.7: 549839-549840 on Build GRCh38
rs981965872 Chr.7: 549860-549860 on Build GRCh38
rs13238949 Chr.7: 549849-549849 on Build GRCh38
rs949141879 Chr.7: 549581-549581 on Build GRCh38
rs774961277 Chr.7: 549892-549892 on Build GRCh38
rs979654512 Chr.7: 549712-549712 on Build GRCh38
rs569762261 Chr.7: 549934-549934 on Build GRCh38
rs1053805608 Chr.7: 549600-549600 on Build GRCh38
rs751521256 Chr.7: 549904-549904 on Build GRCh38
rs141814354 Chr.7: 549852-549852 on Build GRCh38
rs925343444 Chr.7: 550027-550027 on Build GRCh38
rs903900765 Chr.7: 549788-549788 on Build GRCh38
rs538777593 Chr.7: 549971-549971 on Build GRCh38
rs1039229663 Chr.7: 549648-549648 on Build GRCh38
rs956493383 Chr.7: 549881-549881 on Build GRCh38
rs1048887230 Chr.7: 549605-549605 on Build GRCh38
rs879537595 Chr.7: 550006-550006 on Build GRCh38
rs1029365981 Chr.7: 549896-549896 on Build GRCh38
rs1038162936 Chr.7: 549932-549932 on Build GRCh38
rs1030839112 Chr.7: 549981-549981 on Build GRCh38
rs886461263 Chr.7: 549685-549685 on Build GRCh38
rs899201597 Chr.7: 549944-549944 on Build GRCh38
rs1016628533 Chr.7: 549693-549693 on Build GRCh38
rs1039271880 Chr.7: 549789-549789 on Build GRCh38
rs545017074 Chr.7: 549688-549688 on Build GRCh38
rs1042481651 Chr.7: 549883-549883 on Build GRCh38
rs77752769 Chr.7: 549717-549717 on Build GRCh38
rs959668793 Chr.7: 549820-549820 on Build GRCh38

View Full Product Details