Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155946756-155947250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF2
Entrez Gene ID
9181
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases
GEF, GEF-H1, GEFH1, LFP40, P40
UniGene
Hs.743352
Gene Chromosome Location
Chr.1: 155946839-155991263 on Build GRCh38
Transcripts
NM_001162383.1 NM_001162384.1 NM_004723.3 XM_005245587.2 XM_005245588.2 XM_005245589.4 XM_005245590.3 XM_005245591.3 XM_005245592.2 XM_005245593.4 XM_005245594.2 XM_006711622.3 XM_006711623.3 XM_006711624.3 XM_006711625.3 XM_006711626.2 XM_011510136.2 XM_011510137.2 XM_011510139.2 XM_011510141.2 XM_011510143.2 XM_017002794.1 XM_017002795.1 XM_017002796.1 XM_017002797.1 XM_017002798.1 XM_017002799.1

SNP Detail

SNP ID Chromosome Location
rs1009394259 Chr.1: 155947022-155947022 on Build GRCh38
rs1047863824 Chr.1: 155947006-155947006 on Build GRCh38
rs15296 Chr.1: 155947013-155947013 on Build GRCh38
rs550477681 Chr.1: 155946911-155946911 on Build GRCh38
rs890104775 Chr.1: 155947007-155947007 on Build GRCh38
rs765371022 Chr.1: 155947106-155947106 on Build GRCh38
rs1009179087 Chr.1: 155947092-155947092 on Build GRCh38
rs576612065 Chr.1: 155947182-155947182 on Build GRCh38
rs1801836 Chr.1: 155946882-155946882 on Build GRCh38
rs571366123 Chr.1: 155946786-155946786 on Build GRCh38
rs971352358 Chr.1: 155947223-155947223 on Build GRCh38
rs535030112 Chr.1: 155947129-155947129 on Build GRCh38
rs149136860 Chr.1: 155946797-155946797 on Build GRCh38
rs1056206233 Chr.1: 155946883-155946884 on Build GRCh38
rs753015263 Chr.1: 155947162-155947162 on Build GRCh38
rs955681384 Chr.1: 155946989-155946989 on Build GRCh38
rs920343010 Chr.1: 155946926-155946926 on Build GRCh38
rs937701139 Chr.1: 155946877-155946877 on Build GRCh38
rs967918793 Chr.1: 155947104-155947104 on Build GRCh38
rs755992652 Chr.1: 155946925-155946925 on Build GRCh38
rs900703945 Chr.1: 155947110-155947110 on Build GRCh38
rs917668184 Chr.1: 155947187-155947187 on Build GRCh38
rs551031434 Chr.1: 155946782-155946782 on Build GRCh38
rs537283268 Chr.1: 155946898-155946898 on Build GRCh38
rs12037399 Chr.1: 155947109-155947109 on Build GRCh38
rs1037496426 Chr.1: 155946913-155946913 on Build GRCh38
rs929037066 Chr.1: 155946961-155946961 on Build GRCh38
rs1047268095 Chr.1: 155946910-155946912 on Build GRCh38
rs989218251 Chr.1: 155946779-155946779 on Build GRCh38
rs555573983 Chr.1: 155946968-155946968 on Build GRCh38
rs970632101 Chr.1: 155946806-155946806 on Build GRCh38
rs1020770889 Chr.1: 155947069-155947069 on Build GRCh38
rs567510194 Chr.1: 155947103-155947103 on Build GRCh38
rs950205265 Chr.1: 155946902-155946902 on Build GRCh38
rs1000361967 Chr.1: 155947135-155947135 on Build GRCh38
rs56928822 Chr.1: 155947028-155947028 on Build GRCh38
rs997262133 Chr.1: 155946944-155946944 on Build GRCh38
rs956325094 Chr.1: 155947180-155947180 on Build GRCh38
rs749394837 Chr.1: 155946849-155946849 on Build GRCh38
rs981971276 Chr.1: 155946821-155946821 on Build GRCh38
rs899802242 Chr.1: 155946943-155946945 on Build GRCh38
rs1024745780 Chr.1: 155947215-155947215 on Build GRCh38
rs1033140122 Chr.1: 155947176-155947176 on Build GRCh38
rs200909000 Chr.1: 155946884-155946884 on Build GRCh38
rs577979719 Chr.1: 155947227-155947227 on Build GRCh38

View Full Product Details