Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156598135-156598633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH4
Entrez Gene ID
54865
Gene Name
G-patch domain containing 4
Gene Aliases
GPATC4
UniGene
Hs.193832
Gene Chromosome Location
Chr.1: 156594308-156601496 on Build GRCh38
Transcripts
NM_015590.3 NM_182679.2 XM_005245287.3 XM_017000319.1

Gene Symbol
NAXE
Entrez Gene ID
128240
Gene Name
NAD(P)HX epimerase
Gene Aliases
AIBP, APOA1BP, YJEFN1
UniGene
Hs.528320
Gene Chromosome Location
Chr.1: 156591762-156599818 on Build GRCh38
Transcripts
NM_015590.3 NM_182679.2 XM_005245287.3 XM_017000319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1644637 c.238A>C p.T80P Chr.1: 156598235-156598235 on Build GRCh38
4687738 c.168G>A p.G56G Chr.1: 156598421-156598421 on Build GRCh38
4417354 c.114T>C p.T38T Chr.1: 156598460-156598460 on Build GRCh38
6673059 c.81G>A p.A27A Chr.1: 156598254-156598254 on Build GRCh38
5843982 c.207_208delCA p.F69fs*23 Chr.1: 156598265-156598266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042048671 Chr.1: 156598329-156598329 on Build GRCh38
rs750428847 Chr.1: 156598473-156598473 on Build GRCh38
rs533811598 Chr.1: 156598591-156598591 on Build GRCh38
rs773795317 Chr.1: 156598509-156598509 on Build GRCh38
rs772830681 Chr.1: 156598403-156598403 on Build GRCh38
rs574565283 Chr.1: 156598550-156598550 on Build GRCh38
rs765202405 Chr.1: 156598181-156598181 on Build GRCh38
rs764826651 Chr.1: 156598315-156598315 on Build GRCh38
rs757448527 Chr.1: 156598461-156598461 on Build GRCh38
rs574637674 Chr.1: 156598223-156598223 on Build GRCh38
rs762973750 Chr.1: 156598233-156598233 on Build GRCh38
rs912368215 Chr.1: 156598595-156598595 on Build GRCh38
rs757539065 Chr.1: 156598360-156598360 on Build GRCh38
rs761122138 Chr.1: 156598273-156598273 on Build GRCh38
rs967108956 Chr.1: 156598383-156598383 on Build GRCh38
rs374228106 Chr.1: 156598214-156598214 on Build GRCh38
rs753348857 Chr.1: 156598170-156598170 on Build GRCh38
rs147917741 Chr.1: 156598254-156598254 on Build GRCh38
rs375536694 Chr.1: 156598215-156598215 on Build GRCh38
rs41267391 Chr.1: 156598561-156598561 on Build GRCh38
rs200011635 Chr.1: 156598201-156598201 on Build GRCh38
rs371089441 Chr.1: 156598358-156598358 on Build GRCh38
rs763855015 Chr.1: 156598159-156598159 on Build GRCh38
rs767291966 Chr.1: 156598454-156598454 on Build GRCh38
rs778932604 Chr.1: 156598361-156598361 on Build GRCh38
rs772407854 Chr.1: 156598366-156598366 on Build GRCh38
rs777797755 Chr.1: 156598355-156598355 on Build GRCh38
rs751266258 Chr.1: 156598341-156598341 on Build GRCh38
rs761554507 Chr.1: 156598396-156598396 on Build GRCh38
rs759447268 Chr.1: 156598447-156598447 on Build GRCh38
rs747271731 Chr.1: 156598374-156598374 on Build GRCh38
rs147777488 Chr.1: 156598484-156598484 on Build GRCh38
rs532338500 Chr.1: 156598513-156598513 on Build GRCh38
rs750021334 Chr.1: 156598323-156598323 on Build GRCh38
rs966439878 Chr.1: 156598603-156598603 on Build GRCh38
rs759483322 Chr.1: 156598511-156598511 on Build GRCh38
rs777193869 Chr.1: 156598391-156598391 on Build GRCh38
rs747540233 Chr.1: 156598481-156598481 on Build GRCh38
rs750399575 Chr.1: 156598182-156598182 on Build GRCh38
rs756269980 Chr.1: 156598207-156598207 on Build GRCh38
rs759214393 Chr.1: 156598512-156598512 on Build GRCh38
rs369610510 Chr.1: 156598188-156598188 on Build GRCh38
rs371131822 Chr.1: 156598479-156598479 on Build GRCh38
rs550841840 Chr.1: 156598535-156598535 on Build GRCh38
rs775695376 Chr.1: 156598255-156598255 on Build GRCh38
rs779129143 Chr.1: 156598465-156598465 on Build GRCh38
rs751525333 Chr.1: 156598421-156598421 on Build GRCh38
rs11264505 Chr.1: 156598460-156598460 on Build GRCh38
rs529774613 Chr.1: 156598557-156598557 on Build GRCh38
rs746865789 Chr.1: 156598202-156598202 on Build GRCh38
rs367803727 Chr.1: 156598483-156598483 on Build GRCh38
rs558969678 Chr.1: 156598600-156598600 on Build GRCh38
rs780369480 Chr.1: 156598480-156598480 on Build GRCh38
rs201969131 Chr.1: 156598349-156598349 on Build GRCh38
rs751107469 Chr.1: 156598173-156598173 on Build GRCh38
rs766190366 Chr.1: 156598420-156598420 on Build GRCh38
rs575427299 Chr.1: 156598269-156598269 on Build GRCh38
rs912279225 Chr.1: 156598264-156598264 on Build GRCh38
rs146971918 Chr.1: 156598338-156598338 on Build GRCh38
rs763612382 Chr.1: 156598281-156598281 on Build GRCh38
rs778081442 Chr.1: 156598212-156598212 on Build GRCh38
rs996105200 Chr.1: 156598317-156598317 on Build GRCh38
rs566316598 Chr.1: 156598562-156598562 on Build GRCh38
rs1018871906 Chr.1: 156598263-156598263 on Build GRCh38
rs762468259 Chr.1: 156598159-156598161 on Build GRCh38
rs773033597 Chr.1: 156598497-156598497 on Build GRCh38
rs745997074 Chr.1: 156598362-156598362 on Build GRCh38
rs769614933 Chr.1: 156598400-156598400 on Build GRCh38
rs372927651 Chr.1: 156598337-156598337 on Build GRCh38
rs927976280 Chr.1: 156598442-156598442 on Build GRCh38
rs375112655 Chr.1: 156598157-156598157 on Build GRCh38
rs544436631 Chr.1: 156598175-156598175 on Build GRCh38
rs776429208 Chr.1: 156598301-156598301 on Build GRCh38
rs1028644792 Chr.1: 156598560-156598560 on Build GRCh38
rs748576154 Chr.1: 156598486-156598486 on Build GRCh38
rs369352887 Chr.1: 156598312-156598312 on Build GRCh38
rs189330330 Chr.1: 156598407-156598407 on Build GRCh38
rs376109339 Chr.1: 156598375-156598375 on Build GRCh38
rs755870846 Chr.1: 156598368-156598368 on Build GRCh38
rs765821417 Chr.1: 156598305-156598306 on Build GRCh38
rs762308439 Chr.1: 156598508-156598508 on Build GRCh38
rs769524657 Chr.1: 156598493-156598493 on Build GRCh38
rs1005043948 Chr.1: 156598573-156598573 on Build GRCh38
rs569077934 Chr.1: 156598536-156598536 on Build GRCh38

View Full Product Details