Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157575610-157576118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL4
Entrez Gene ID
83417
Gene Name
Fc receptor like 4
Gene Aliases
CD307d, FCRH4, IGFP2, IRTA1
UniGene
Hs.120260
Gene Chromosome Location
Chr.1: 157573749-157598080 on Build GRCh38
Transcripts
NM_031282.2 XM_011510034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
28472 c.1461+5G>A p.? Chr.1: 157575694-157575694 on Build GRCh38
4394502 c.1456G>A p.D486N Chr.1: 157575704-157575704 on Build GRCh38
1985182 c.1439C>A p.S480Y Chr.1: 157575721-157575721 on Build GRCh38
5714850 c.1461G>A p.K487K Chr.1: 157575699-157575699 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs917869622 Chr.1: 157575943-157575943 on Build GRCh38
rs749251080 Chr.1: 157575763-157575763 on Build GRCh38
rs979552746 Chr.1: 157575865-157575865 on Build GRCh38
rs1045363247 Chr.1: 157575945-157575945 on Build GRCh38
rs890718196 Chr.1: 157575849-157575849 on Build GRCh38
rs74118680 Chr.1: 157576031-157576031 on Build GRCh38
rs751374514 Chr.1: 157575771-157575771 on Build GRCh38
rs567657314 Chr.1: 157575788-157575788 on Build GRCh38
rs887548684 Chr.1: 157575659-157575659 on Build GRCh38
rs753258971 Chr.1: 157575744-157575744 on Build GRCh38
rs774422123 Chr.1: 157576075-157576075 on Build GRCh38
rs140634688 Chr.1: 157575693-157575693 on Build GRCh38
rs776326515 Chr.1: 157575755-157575755 on Build GRCh38
rs549538199 Chr.1: 157575960-157575960 on Build GRCh38
rs763696250 Chr.1: 157575753-157575753 on Build GRCh38
rs368411125 Chr.1: 157575637-157575637 on Build GRCh38
rs534661708 Chr.1: 157575802-157575802 on Build GRCh38
rs554918653 Chr.1: 157575944-157575944 on Build GRCh38
rs772443837 Chr.1: 157575707-157575707 on Build GRCh38
rs961697757 Chr.1: 157575756-157575756 on Build GRCh38
rs375180001 Chr.1: 157575776-157575776 on Build GRCh38
rs34703498 Chr.1: 157575873-157575874 on Build GRCh38
rs112034180 Chr.1: 157575747-157575747 on Build GRCh38
rs749363901 Chr.1: 157575689-157575689 on Build GRCh38
rs769965358 Chr.1: 157575671-157575671 on Build GRCh38
rs779042776 Chr.1: 157575641-157575641 on Build GRCh38
rs372191425 Chr.1: 157575740-157575740 on Build GRCh38
rs1036076276 Chr.1: 157576065-157576065 on Build GRCh38
rs774857897 Chr.1: 157575699-157575699 on Build GRCh38
rs192081827 Chr.1: 157575845-157575845 on Build GRCh38
rs543346604 Chr.1: 157575941-157575941 on Build GRCh38
rs766552379 Chr.1: 157575856-157575856 on Build GRCh38
rs375783353 Chr.1: 157575910-157575910 on Build GRCh38
rs779927177 Chr.1: 157575664-157575664 on Build GRCh38
rs924809235 Chr.1: 157575806-157575806 on Build GRCh38
rs374476793 Chr.1: 157575731-157575731 on Build GRCh38
rs564737207 Chr.1: 157576051-157576051 on Build GRCh38
rs911374043 Chr.1: 157575822-157575822 on Build GRCh38
rs35328355 Chr.1: 157575998-157575999 on Build GRCh38
rs1026860925 Chr.1: 157575654-157575654 on Build GRCh38
rs78489028 Chr.1: 157575880-157575880 on Build GRCh38
rs923986865 Chr.1: 157575870-157575870 on Build GRCh38
rs367654161 Chr.1: 157575737-157575737 on Build GRCh38
rs762228769 Chr.1: 157575705-157575705 on Build GRCh38
rs754060446 Chr.1: 157575989-157575989 on Build GRCh38
rs748415070 Chr.1: 157575668-157575668 on Build GRCh38
rs528015393 Chr.1: 157575857-157575857 on Build GRCh38
rs758351246 Chr.1: 157575769-157575769 on Build GRCh38
rs764799871 Chr.1: 157575761-157575761 on Build GRCh38
rs759973261 Chr.1: 157575701-157575701 on Build GRCh38
rs758299870 Chr.1: 157575655-157575655 on Build GRCh38
rs145665825 Chr.1: 157575821-157575821 on Build GRCh38
rs997170966 Chr.1: 157576074-157576074 on Build GRCh38
rs767525096 Chr.1: 157575710-157575710 on Build GRCh38
rs750338031 Chr.1: 157575643-157575643 on Build GRCh38
rs778048886 Chr.1: 157575679-157575679 on Build GRCh38
rs975683809 Chr.1: 157575760-157575760 on Build GRCh38
rs887632612 Chr.1: 157576040-157576040 on Build GRCh38
rs572818668 Chr.1: 157576036-157576036 on Build GRCh38
rs60560678 Chr.1: 157575889-157575889 on Build GRCh38
rs756761107 Chr.1: 157575638-157575638 on Build GRCh38
rs994956775 Chr.1: 157575907-157575907 on Build GRCh38
rs1004473171 Chr.1: 157575956-157575956 on Build GRCh38
rs564524633 Chr.1: 157576072-157576072 on Build GRCh38
rs947175924 Chr.1: 157575783-157575783 on Build GRCh38
rs761253435 Chr.1: 157575759-157575759 on Build GRCh38

View Full Product Details