Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27162317-27162862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA10-HOXA9
Entrez Gene ID
100534589
Gene Name
HOXA10-HOXA9 readthrough
Gene Aliases
-
UniGene
Hs.659350
Gene Chromosome Location
Chr.7: 27162438-27180261 on Build GRCh38
Transcripts
NR_037940.1 NM_152739.3

Gene Symbol
HOXA9
Entrez Gene ID
3205
Gene Name
homeobox A9
Gene Aliases
ABD-B, HOX1, HOX1.7, HOX1G
UniGene
Hs.659350
Gene Chromosome Location
Chr.7: 27162438-27165530 on Build GRCh38
Transcripts
NR_037940.1 NM_152739.3

SNP Detail

SNP ID Chromosome Location
rs971402857 Chr.7: 27162611-27162611 on Build GRCh38
rs892678285 Chr.7: 27162698-27162698 on Build GRCh38
rs1025143709 Chr.7: 27162715-27162715 on Build GRCh38
rs780882126 Chr.7: 27162472-27162472 on Build GRCh38
rs1013530021 Chr.7: 27162597-27162597 on Build GRCh38
rs1016378995 Chr.7: 27162749-27162749 on Build GRCh38
rs1049205710 Chr.7: 27162352-27162352 on Build GRCh38
rs537921686 Chr.7: 27162773-27162773 on Build GRCh38
rs530226198 Chr.7: 27162395-27162395 on Build GRCh38
rs908842613 Chr.7: 27162355-27162355 on Build GRCh38
rs941821183 Chr.7: 27162816-27162816 on Build GRCh38
rs370814748 Chr.7: 27162722-27162722 on Build GRCh38
rs1013810770 Chr.7: 27162829-27162829 on Build GRCh38
rs563289988 Chr.7: 27162494-27162496 on Build GRCh38
rs972445398 Chr.7: 27162801-27162801 on Build GRCh38
rs1050009334 Chr.7: 27162775-27162775 on Build GRCh38
rs913389696 Chr.7: 27162375-27162375 on Build GRCh38
rs963735434 Chr.7: 27162494-27162494 on Build GRCh38
rs1050145581 Chr.7: 27162503-27162503 on Build GRCh38
rs181737685 Chr.7: 27162666-27162666 on Build GRCh38
rs10259620 Chr.7: 27162670-27162670 on Build GRCh38
rs1024876124 Chr.7: 27162605-27162605 on Build GRCh38
rs552137702 Chr.7: 27162427-27162427 on Build GRCh38
rs565976296 Chr.7: 27162668-27162668 on Build GRCh38
rs938275097 Chr.7: 27162361-27162361 on Build GRCh38
rs530685033 Chr.7: 27162630-27162630 on Build GRCh38
rs752231477 Chr.7: 27162587-27162587 on Build GRCh38
rs145905865 Chr.7: 27162734-27162734 on Build GRCh38
rs570502030 Chr.7: 27162791-27162791 on Build GRCh38
rs35335761 Chr.7: 27162490-27162492 on Build GRCh38
rs568128306 Chr.7: 27162708-27162708 on Build GRCh38
rs1036506280 Chr.7: 27162817-27162817 on Build GRCh38
rs963511788 Chr.7: 27162782-27162782 on Build GRCh38
rs777287088 Chr.7: 27162669-27162669 on Build GRCh38
rs548387720 Chr.7: 27162704-27162704 on Build GRCh38
rs561294242 Chr.7: 27162360-27162360 on Build GRCh38
rs758756193 Chr.7: 27162779-27162779 on Build GRCh38
rs1025157041 Chr.7: 27162350-27162350 on Build GRCh38
rs552096220 Chr.7: 27162650-27162650 on Build GRCh38
rs887493602 Chr.7: 27162526-27162526 on Build GRCh38
rs766684947 Chr.7: 27162756-27162756 on Build GRCh38
rs1013841592 Chr.7: 27162711-27162711 on Build GRCh38
rs764532234 Chr.7: 27162809-27162809 on Build GRCh38
rs901540701 Chr.7: 27162479-27162481 on Build GRCh38
rs1054591139 Chr.7: 27162355-27162356 on Build GRCh38
rs543493227 Chr.7: 27162420-27162420 on Build GRCh38
rs912435066 Chr.7: 27162603-27162603 on Build GRCh38
rs927366199 Chr.7: 27162644-27162644 on Build GRCh38
rs929922292 Chr.7: 27162585-27162585 on Build GRCh38
rs946354356 Chr.7: 27162390-27162390 on Build GRCh38
rs532370978 Chr.7: 27162642-27162642 on Build GRCh38
rs1016263986 Chr.7: 27162453-27162453 on Build GRCh38
rs1024906920 Chr.7: 27162836-27162836 on Build GRCh38
rs755828947 Chr.7: 27162641-27162641 on Build GRCh38
rs539449195 Chr.7: 27162824-27162824 on Build GRCh38
rs759592020 Chr.7: 27162544-27162544 on Build GRCh38
rs190122610 Chr.7: 27162549-27162549 on Build GRCh38
rs149001266 Chr.7: 27162721-27162721 on Build GRCh38

View Full Product Details