Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 33152561-33153087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS9
Entrez Gene ID
27241
Gene Name
Bardet-Biedl syndrome 9
Gene Aliases
B1, C18, D1, PTHB1
UniGene
Hs.372360
Gene Chromosome Location
Chr.7: 33129244-33637238 on Build GRCh38
Transcripts
NM_001033604.1 NM_001033605.1 NM_014451.3 NM_198428.2 XM_005249700.3 XM_005249701.2 XM_011515264.2 XM_011515265.2 XM_011515266.2 XM_011515267.2 XM_011515268.1 XM_011515269.2 XM_011515270.2 XM_017011990.1 XM_017011991.1 XM_017011992.1 XM_017011993.1 XM_017011994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5919231 c.230C>T p.P77L Chr.7: 33152818-33152818 on Build GRCh38
3720954 c.215T>A p.V72E Chr.7: 33152803-33152803 on Build GRCh38
3950444 c.187G>A p.A63T Chr.7: 33152775-33152775 on Build GRCh38
7259788 c.162C>T p.P54P Chr.7: 33152750-33152750 on Build GRCh38
6610182 c.217G>A p.D73N Chr.7: 33152805-33152805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745966272 Chr.7: 33152891-33152891 on Build GRCh38
rs917477982 Chr.7: 33153054-33153054 on Build GRCh38
rs931618905 Chr.7: 33152594-33152594 on Build GRCh38
rs62449324 Chr.7: 33152690-33152690 on Build GRCh38
rs374383847 Chr.7: 33152864-33152864 on Build GRCh38
rs898791327 Chr.7: 33152919-33152919 on Build GRCh38
rs1008748533 Chr.7: 33152754-33152754 on Build GRCh38
rs758009096 Chr.7: 33152732-33152732 on Build GRCh38
rs765240871 Chr.7: 33152802-33152802 on Build GRCh38
rs771129780 Chr.7: 33152794-33152794 on Build GRCh38
rs766917697 Chr.7: 33152839-33152839 on Build GRCh38
rs890394799 Chr.7: 33152960-33152960 on Build GRCh38
rs776811498 Chr.7: 33152796-33152796 on Build GRCh38
rs778349431 Chr.7: 33152876-33152876 on Build GRCh38
rs752099175 Chr.7: 33152677-33152678 on Build GRCh38
rs543714605 Chr.7: 33152687-33152687 on Build GRCh38
rs377626008 Chr.7: 33152837-33152837 on Build GRCh38
rs368821728 Chr.7: 33152884-33152884 on Build GRCh38
rs769256027 Chr.7: 33152778-33152778 on Build GRCh38
rs751305909 Chr.7: 33152609-33152609 on Build GRCh38
rs996103797 Chr.7: 33152595-33152595 on Build GRCh38
rs751209650 Chr.7: 33152836-33152836 on Build GRCh38
rs1008841963 Chr.7: 33152969-33152969 on Build GRCh38
rs1015915957 Chr.7: 33152647-33152647 on Build GRCh38
rs762752188 Chr.7: 33152814-33152814 on Build GRCh38
rs35872534 Chr.7: 33152665-33152666 on Build GRCh38
rs139401222 Chr.7: 33152589-33152589 on Build GRCh38
rs931651739 Chr.7: 33152902-33152902 on Build GRCh38
rs763936170 Chr.7: 33152819-33152819 on Build GRCh38
rs756750359 Chr.7: 33152743-33152743 on Build GRCh38
rs370916293 Chr.7: 33152855-33152855 on Build GRCh38
rs370508659 Chr.7: 33152823-33152823 on Build GRCh38
rs879931572 Chr.7: 33152989-33152989 on Build GRCh38
rs777409972 Chr.7: 33152733-33152733 on Build GRCh38
rs991662754 Chr.7: 33152860-33152860 on Build GRCh38
rs538470220 Chr.7: 33152676-33152677 on Build GRCh38
rs761920074 Chr.7: 33152662-33152662 on Build GRCh38
rs17169881 Chr.7: 33152890-33152890 on Build GRCh38
rs200845910 Chr.7: 33152786-33152786 on Build GRCh38
rs779477193 Chr.7: 33152862-33152862 on Build GRCh38
rs995848131 Chr.7: 33152941-33152941 on Build GRCh38
rs773577974 Chr.7: 33152784-33152784 on Build GRCh38
rs775081992 Chr.7: 33152811-33152811 on Build GRCh38
rs749727731 Chr.7: 33152764-33152764 on Build GRCh38
rs111473409 Chr.7: 33152885-33152885 on Build GRCh38
rs764906719 Chr.7: 33152716-33152716 on Build GRCh38
rs10241188 Chr.7: 33152979-33152979 on Build GRCh38
rs753878819 Chr.7: 33152677-33152677 on Build GRCh38
rs148812031 Chr.7: 33152762-33152762 on Build GRCh38
rs925968981 Chr.7: 33152810-33152810 on Build GRCh38
rs376183075 Chr.7: 33152799-33152799 on Build GRCh38
rs749974697 Chr.7: 33152851-33152851 on Build GRCh38
rs374140910 Chr.7: 33152892-33152892 on Build GRCh38
rs1037893058 Chr.7: 33152911-33152911 on Build GRCh38
rs549303303 Chr.7: 33152906-33152906 on Build GRCh38
rs759795035 Chr.7: 33152612-33152613 on Build GRCh38
rs766360617 Chr.7: 33152688-33152688 on Build GRCh38
rs754919644 Chr.7: 33152633-33152633 on Build GRCh38
rs1028914924 Chr.7: 33152724-33152724 on Build GRCh38
rs912179458 Chr.7: 33152963-33152963 on Build GRCh38
rs755981354 Chr.7: 33153011-33153011 on Build GRCh38
rs371505366 Chr.7: 33152615-33152615 on Build GRCh38
rs373063776 Chr.7: 33152783-33152783 on Build GRCh38
rs1009074155 Chr.7: 33152663-33152663 on Build GRCh38
rs113260332 Chr.7: 33152881-33152881 on Build GRCh38
rs567428001 Chr.7: 33152942-33152942 on Build GRCh38
rs991464557 Chr.7: 33153053-33153053 on Build GRCh38
rs34249217 Chr.7: 33152988-33152988 on Build GRCh38
rs145241295 Chr.7: 33152726-33152726 on Build GRCh38
rs764116871 Chr.7: 33152989-33152989 on Build GRCh38
rs773383871 Chr.7: 33152678-33152678 on Build GRCh38
rs534533649 Chr.7: 33152950-33152950 on Build GRCh38
rs375757123 Chr.7: 33152793-33152793 on Build GRCh38
rs137962929 Chr.7: 33152852-33152852 on Build GRCh38
rs1020102844 Chr.7: 33152970-33152970 on Build GRCh38
rs751016663 Chr.7: 33152735-33152735 on Build GRCh38
rs775486299 Chr.7: 33152896-33152896 on Build GRCh38
rs568886417 Chr.7: 33152640-33152640 on Build GRCh38
rs186038329 Chr.7: 33152625-33152625 on Build GRCh38
rs958666918 Chr.7: 33153035-33153035 on Build GRCh38
rs753701101 Chr.7: 33152701-33152701 on Build GRCh38
rs753930404 Chr.7: 33152715-33152715 on Build GRCh38

View Full Product Details