Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36549289-36549824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOAH
Entrez Gene ID
313
Gene Name
acyloxyacyl hydrolase
Gene Aliases
-
UniGene
Hs.488007
Gene Chromosome Location
Chr.7: 36512042-36724549 on Build GRCh38
Transcripts
NM_001177506.1 NM_001177507.1 NM_001637.3 XM_011515333.2 XM_011515334.2 XM_011515335.2 XM_011515336.2 XM_011515338.2 XM_011515339.2 XM_011515340.2 XM_011515341.2 XM_011515342.2 XM_017012102.1 XM_017012103.1 XM_017012104.1 XM_017012105.1 XM_017012106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3228931 c.1022-1G>T p.? Chr.7: 36549476-36549476 on Build GRCh38
5912567 c.1022-1G>A p.? Chr.7: 36549476-36549476 on Build GRCh38
1635295 c.1025C>T p.A342V Chr.7: 36549472-36549472 on Build GRCh38
3950480 c.1058G>A p.S353N Chr.7: 36549439-36549439 on Build GRCh38
5997351 c.1036A>G p.N346D Chr.7: 36549461-36549461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943456485 Chr.7: 36549750-36549750 on Build GRCh38
rs779690818 Chr.7: 36549482-36549487 on Build GRCh38
rs186707363 Chr.7: 36549391-36549391 on Build GRCh38
rs553566047 Chr.7: 36549371-36549371 on Build GRCh38
rs564896421 Chr.7: 36549456-36549456 on Build GRCh38
rs977152378 Chr.7: 36549684-36549684 on Build GRCh38
rs183645533 Chr.7: 36549577-36549577 on Build GRCh38
rs763318015 Chr.7: 36549493-36549493 on Build GRCh38
rs187897689 Chr.7: 36549581-36549581 on Build GRCh38
rs919272019 Chr.7: 36549661-36549661 on Build GRCh38
rs760939192 Chr.7: 36549497-36549499 on Build GRCh38
rs201018752 Chr.7: 36549419-36549419 on Build GRCh38
rs56036432 Chr.7: 36549580-36549580 on Build GRCh38
rs929404239 Chr.7: 36549453-36549453 on Build GRCh38
rs959177708 Chr.7: 36549416-36549416 on Build GRCh38
rs770312641 Chr.7: 36549463-36549463 on Build GRCh38
rs759991483 Chr.7: 36549459-36549459 on Build GRCh38
rs911943680 Chr.7: 36549743-36549743 on Build GRCh38
rs771656372 Chr.7: 36549446-36549446 on Build GRCh38
rs3735397 Chr.7: 36549349-36549349 on Build GRCh38
rs575253954 Chr.7: 36549518-36549518 on Build GRCh38
rs915503895 Chr.7: 36549771-36549771 on Build GRCh38
rs571227303 Chr.7: 36549747-36549747 on Build GRCh38
rs759474536 Chr.7: 36549724-36549724 on Build GRCh38
rs528550051 Chr.7: 36549422-36549426 on Build GRCh38
rs768136513 Chr.7: 36549406-36549406 on Build GRCh38
rs990031593 Chr.7: 36549756-36549756 on Build GRCh38
rs956872358 Chr.7: 36549754-36549754 on Build GRCh38
rs915450438 Chr.7: 36549404-36549404 on Build GRCh38
rs368151055 Chr.7: 36549440-36549440 on Build GRCh38
rs965825379 Chr.7: 36549641-36549641 on Build GRCh38
rs972632146 Chr.7: 36549644-36549644 on Build GRCh38
rs764354370 Chr.7: 36549394-36549394 on Build GRCh38
rs775761869 Chr.7: 36549464-36549464 on Build GRCh38
rs764352317 Chr.7: 36549767-36549767 on Build GRCh38
rs764206288 Chr.7: 36549505-36549505 on Build GRCh38
rs572351259 Chr.7: 36549341-36549341 on Build GRCh38
rs902160875 Chr.7: 36549549-36549549 on Build GRCh38
rs377063687 Chr.7: 36549630-36549630 on Build GRCh38
rs943492558 Chr.7: 36549475-36549475 on Build GRCh38
rs192489643 Chr.7: 36549575-36549575 on Build GRCh38
rs981815986 Chr.7: 36549321-36549321 on Build GRCh38
rs759425783 Chr.7: 36549672-36549672 on Build GRCh38
rs751827146 Chr.7: 36549420-36549422 on Build GRCh38
rs973851561 Chr.7: 36549438-36549438 on Build GRCh38
rs563415448 Chr.7: 36549567-36549567 on Build GRCh38
rs906651659 Chr.7: 36549476-36549476 on Build GRCh38
rs545103372 Chr.7: 36549394-36549394 on Build GRCh38
rs371956492 Chr.7: 36549520-36549520 on Build GRCh38
rs189711639 Chr.7: 36549776-36549776 on Build GRCh38
rs994599244 Chr.7: 36549560-36549560 on Build GRCh38
rs897648983 Chr.7: 36549534-36549534 on Build GRCh38
rs78699547 Chr.7: 36549737-36549737 on Build GRCh38
rs754828838 Chr.7: 36549500-36549503 on Build GRCh38
rs778686603 Chr.7: 36549421-36549421 on Build GRCh38
rs747610695 Chr.7: 36549422-36549422 on Build GRCh38
rs1039647275 Chr.7: 36549753-36549753 on Build GRCh38

View Full Product Details