Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 40188123-40188662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUGCT
Entrez Gene ID
79783
Gene Name
succinyl-CoA:glutarate-CoA transferase
Gene Aliases
C7orf10, DERP13, GA3, ORF19
UniGene
Hs.586313
Gene Chromosome Location
Chr.7: 40134887-40860767 on Build GRCh38
Transcripts
NM_001193311.1 NM_001193312.1 NM_001193313.1 NM_024728.2 XM_006715775.3 XM_011515525.2 XM_011515526.2 XM_011515527.2 XM_011515528.2 XM_011515529.2 XM_011515530.2 XM_017012621.1 XM_017012622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6430128 c.242C>T p.T81I Chr.7: 40188510-40188510 on Build GRCh38
1089579 c.323G>A p.R108Q Chr.7: 40188570-40188570 on Build GRCh38
4976302 c.248C>A p.T83N Chr.7: 40188516-40188516 on Build GRCh38
6177896 c.307T>C p.Y103H Chr.7: 40188554-40188554 on Build GRCh38
5965642 c.291C>T p.L97L Chr.7: 40188559-40188559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544993739 Chr.7: 40188451-40188451 on Build GRCh38
rs138579332 Chr.7: 40188313-40188313 on Build GRCh38
rs761081046 Chr.7: 40188550-40188550 on Build GRCh38
rs1053546049 Chr.7: 40188362-40188362 on Build GRCh38
rs868533936 Chr.7: 40188500-40188500 on Build GRCh38
rs184082488 Chr.7: 40188172-40188172 on Build GRCh38
rs764513898 Chr.7: 40188461-40188462 on Build GRCh38
rs757782836 Chr.7: 40188555-40188559 on Build GRCh38
rs645016 Chr.7: 40188459-40188459 on Build GRCh38
rs766721841 Chr.7: 40188558-40188558 on Build GRCh38
rs936255299 Chr.7: 40188340-40188340 on Build GRCh38
rs758483577 Chr.7: 40188586-40188586 on Build GRCh38
rs887441756 Chr.7: 40188629-40188629 on Build GRCh38
rs926403891 Chr.7: 40188167-40188167 on Build GRCh38
rs1016385031 Chr.7: 40188449-40188449 on Build GRCh38
rs756435996 Chr.7: 40188466-40188466 on Build GRCh38
rs922665575 Chr.7: 40188440-40188441 on Build GRCh38
rs368991810 Chr.7: 40188454-40188456 on Build GRCh38
rs1010869375 Chr.7: 40188194-40188194 on Build GRCh38
rs769359903 Chr.7: 40188514-40188514 on Build GRCh38
rs759175316 Chr.7: 40188445-40188445 on Build GRCh38
rs771802565 Chr.7: 40188452-40188456 on Build GRCh38
rs1045518931 Chr.7: 40188542-40188542 on Build GRCh38
rs758231131 Chr.7: 40188499-40188499 on Build GRCh38
rs549381700 Chr.7: 40188261-40188261 on Build GRCh38
rs1024018244 Chr.7: 40188210-40188210 on Build GRCh38
rs902128316 Chr.7: 40188393-40188393 on Build GRCh38
rs752118948 Chr.7: 40188533-40188533 on Build GRCh38
rs746391689 Chr.7: 40188434-40188435 on Build GRCh38
rs945008885 Chr.7: 40188557-40188557 on Build GRCh38
rs891963048 Chr.7: 40188363-40188363 on Build GRCh38
rs781076109 Chr.7: 40188626-40188626 on Build GRCh38
rs190709227 Chr.7: 40188161-40188161 on Build GRCh38
rs752763152 Chr.7: 40188583-40188583 on Build GRCh38
rs933575025 Chr.7: 40188395-40188395 on Build GRCh38
rs564584016 Chr.7: 40188452-40188452 on Build GRCh38
rs985057534 Chr.7: 40188252-40188252 on Build GRCh38
rs989631466 Chr.7: 40188524-40188524 on Build GRCh38
rs772606842 Chr.7: 40188528-40188528 on Build GRCh38
rs557143955 Chr.7: 40188353-40188353 on Build GRCh38
rs774925854 Chr.7: 40188456-40188456 on Build GRCh38
rs542190042 Chr.7: 40188158-40188158 on Build GRCh38
rs972794689 Chr.7: 40188435-40188446 on Build GRCh38
rs762411634 Chr.7: 40188527-40188527 on Build GRCh38
rs566119739 Chr.7: 40188299-40188299 on Build GRCh38
rs182876521 Chr.7: 40188260-40188260 on Build GRCh38
rs1006133242 Chr.7: 40188425-40188425 on Build GRCh38
rs528655730 Chr.7: 40188624-40188624 on Build GRCh38
rs35948652 Chr.7: 40188435-40188436 on Build GRCh38
rs757610671 Chr.7: 40188214-40188214 on Build GRCh38
rs529624624 Chr.7: 40188365-40188365 on Build GRCh38
rs751538525 Chr.7: 40188594-40188594 on Build GRCh38
rs748143546 Chr.7: 40188492-40188492 on Build GRCh38
rs193174297 Chr.7: 40188205-40188205 on Build GRCh38
rs980544369 Chr.7: 40188344-40188344 on Build GRCh38
rs78060149 Chr.7: 40188479-40188479 on Build GRCh38
rs892132760 Chr.7: 40188177-40188177 on Build GRCh38
rs137852862 Chr.7: 40188569-40188569 on Build GRCh38
rs758857464 Chr.7: 40188460-40188463 on Build GRCh38
rs901177686 Chr.7: 40188152-40188152 on Build GRCh38
rs62456121 Chr.7: 40188463-40188463 on Build GRCh38
rs764649732 Chr.7: 40188446-40188446 on Build GRCh38
rs147264387 Chr.7: 40188380-40188380 on Build GRCh38
rs147109282 Chr.7: 40188456-40188456 on Build GRCh38
rs570023120 Chr.7: 40188251-40188251 on Build GRCh38
rs12667172 Chr.7: 40188435-40188435 on Build GRCh38
rs34697046 Chr.7: 40188467-40188468 on Build GRCh38
rs913673123 Chr.7: 40188533-40188533 on Build GRCh38
rs777825350 Chr.7: 40188511-40188511 on Build GRCh38
rs186242615 Chr.7: 40188159-40188159 on Build GRCh38
rs550085116 Chr.7: 40188564-40188564 on Build GRCh38
rs773160108 Chr.7: 40188451-40188452 on Build GRCh38
rs1050708824 Chr.7: 40188630-40188630 on Build GRCh38
rs644966 Chr.7: 40188429-40188429 on Build GRCh38
rs369992662 Chr.7: 40188589-40188589 on Build GRCh38
rs972783244 Chr.7: 40188283-40188283 on Build GRCh38
rs917873557 Chr.7: 40188325-40188325 on Build GRCh38
rs373608132 Chr.7: 40188455-40188456 on Build GRCh38
rs952076484 Chr.7: 40188406-40188406 on Build GRCh38
rs889295242 Chr.7: 40188447-40188447 on Build GRCh38
rs762169140 Chr.7: 40188460-40188460 on Build GRCh38
rs60252112 Chr.7: 40188433-40188434 on Build GRCh38
rs917050349 Chr.7: 40188532-40188533 on Build GRCh38
rs200690632 Chr.7: 40188513-40188513 on Build GRCh38
rs1024944924 Chr.7: 40188455-40188455 on Build GRCh38
rs1034241776 Chr.7: 40188238-40188238 on Build GRCh38
rs541476805 Chr.7: 40188326-40188326 on Build GRCh38
rs115327068 Chr.7: 40188636-40188636 on Build GRCh38
rs765884658 Chr.7: 40188453-40188456 on Build GRCh38
rs773981693 Chr.7: 40188352-40188352 on Build GRCh38
rs143928089 Chr.7: 40188333-40188333 on Build GRCh38
rs747590282 Chr.7: 40188515-40188515 on Build GRCh38
rs758367280 Chr.7: 40188432-40188433 on Build GRCh38
rs117708232 Chr.7: 40188162-40188162 on Build GRCh38
rs774813982 Chr.7: 40188517-40188517 on Build GRCh38
rs371476138 Chr.7: 40188377-40188377 on Build GRCh38
rs542350616 Chr.7: 40188370-40188370 on Build GRCh38
rs961870012 Chr.7: 40188435-40188442 on Build GRCh38
rs1006326139 Chr.7: 40188263-40188263 on Build GRCh38
rs980657289 Chr.7: 40188484-40188484 on Build GRCh38
rs776261298 Chr.7: 40188452-40188453 on Build GRCh38
rs745555927 Chr.7: 40188628-40188628 on Build GRCh38
rs766634150 Chr.7: 40188478-40188478 on Build GRCh38
rs778804466 Chr.7: 40188481-40188481 on Build GRCh38
rs370258889 Chr.7: 40188311-40188311 on Build GRCh38
rs1029070783 Chr.7: 40188236-40188236 on Build GRCh38
rs1006685545 Chr.7: 40188448-40188448 on Build GRCh38
rs536808745 Chr.7: 40188337-40188337 on Build GRCh38
rs746851191 Chr.7: 40188512-40188512 on Build GRCh38
rs376341860 Chr.7: 40188570-40188570 on Build GRCh38
rs558915993 Chr.7: 40188399-40188399 on Build GRCh38
rs773683780 Chr.7: 40188546-40188546 on Build GRCh38
rs936446906 Chr.7: 40188171-40188171 on Build GRCh38
rs188233722 Chr.7: 40188204-40188204 on Build GRCh38
rs1016641364 Chr.7: 40188267-40188267 on Build GRCh38
rs752755011 Chr.7: 40188163-40188163 on Build GRCh38
rs750745276 Chr.7: 40188464-40188464 on Build GRCh38

View Full Product Details