Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 40189549-40190073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUGCT
Entrez Gene ID
79783
Gene Name
succinyl-CoA:glutarate-CoA transferase
Gene Aliases
C7orf10, DERP13, GA3, ORF19
UniGene
Hs.586313
Gene Chromosome Location
Chr.7: 40134887-40860767 on Build GRCh38
Transcripts
NM_001193311.1 NM_001193312.1 NM_001193313.1 NM_024728.2 XM_006715775.3 XM_011515525.2 XM_011515526.2 XM_011515527.2 XM_011515528.2 XM_011515529.2 XM_011515530.2 XM_017012621.1 XM_017012622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7266533 c.345G>T p.G115G Chr.7: 40189576-40189576 on Build GRCh38
6246502 c.355G>T p.D119Y Chr.7: 40189565-40189565 on Build GRCh38
5101430 c.384+1G>T p.? Chr.7: 40189595-40189595 on Build GRCh38
6246503 c.334G>T p.D112Y Chr.7: 40189565-40189565 on Build GRCh38
4449828 c.323delT p.N109fs*8 Chr.7: 40189554-40189554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774772132 Chr.7: 40189738-40189738 on Build GRCh38
rs552765057 Chr.7: 40189743-40189743 on Build GRCh38
rs781757090 Chr.7: 40189750-40189750 on Build GRCh38
rs1013170565 Chr.7: 40189986-40189986 on Build GRCh38
rs963590463 Chr.7: 40190014-40190014 on Build GRCh38
rs898140751 Chr.7: 40189872-40189872 on Build GRCh38
rs762319962 Chr.7: 40189812-40189812 on Build GRCh38
rs993600797 Chr.7: 40190020-40190020 on Build GRCh38
rs757210588 Chr.7: 40189632-40189632 on Build GRCh38
rs750224714 Chr.7: 40189642-40189642 on Build GRCh38
rs558259075 Chr.7: 40189806-40189806 on Build GRCh38
rs1042310839 Chr.7: 40189667-40189667 on Build GRCh38
rs759916398 Chr.7: 40189589-40189589 on Build GRCh38
rs751557001 Chr.7: 40189625-40189625 on Build GRCh38
rs902461783 Chr.7: 40189699-40189699 on Build GRCh38
rs894463554 Chr.7: 40189932-40189932 on Build GRCh38
rs990920169 Chr.7: 40190031-40190031 on Build GRCh38
rs991322179 Chr.7: 40189628-40189628 on Build GRCh38
rs376552381 Chr.7: 40189590-40189590 on Build GRCh38
rs987128670 Chr.7: 40189618-40189618 on Build GRCh38
rs1026419988 Chr.7: 40190028-40190028 on Build GRCh38
rs928153059 Chr.7: 40189581-40189581 on Build GRCh38
rs535693245 Chr.7: 40189662-40189662 on Build GRCh38
rs982334267 Chr.7: 40189979-40189979 on Build GRCh38
rs765429584 Chr.7: 40189592-40189592 on Build GRCh38
rs889624574 Chr.7: 40189783-40189783 on Build GRCh38
rs1057188078 Chr.7: 40189921-40189921 on Build GRCh38
rs915240458 Chr.7: 40189648-40189648 on Build GRCh38
rs370847708 Chr.7: 40189612-40189612 on Build GRCh38
rs570267821 Chr.7: 40189781-40189781 on Build GRCh38
rs10237243 Chr.7: 40189915-40189915 on Build GRCh38
rs557837131 Chr.7: 40189779-40189779 on Build GRCh38
rs1006634328 Chr.7: 40189784-40189784 on Build GRCh38
rs993820584 Chr.7: 40189880-40189880 on Build GRCh38
rs915088470 Chr.7: 40189760-40189760 on Build GRCh38
rs752951817 Chr.7: 40189599-40189599 on Build GRCh38
rs1041843627 Chr.7: 40189827-40189827 on Build GRCh38
rs575240907 Chr.7: 40189833-40189833 on Build GRCh38
rs781162561 Chr.7: 40189635-40189635 on Build GRCh38
rs959843306 Chr.7: 40190029-40190029 on Build GRCh38
rs536003523 Chr.7: 40189723-40189723 on Build GRCh38
rs910214220 Chr.7: 40189696-40189696 on Build GRCh38
rs750816873 Chr.7: 40189977-40189977 on Build GRCh38
rs1035118521 Chr.7: 40190012-40190012 on Build GRCh38
rs550972462 Chr.7: 40189889-40189889 on Build GRCh38
rs539509845 Chr.7: 40189713-40189713 on Build GRCh38

View Full Product Details