Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 41689302-41689791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INHBA
Entrez Gene ID
3624
Gene Name
inhibin beta A subunit
Gene Aliases
EDF, FRP
UniGene
Hs.28792
Gene Chromosome Location
Chr.7: 41685101-41710532 on Build GRCh38
Transcripts
NM_002192.2 XM_017012174.1 XM_017012175.1 XM_017012176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4807994 c.1276T>A p.S426T Chr.7: 41689655-41689655 on Build GRCh38
6965301 c.1262A>T p.E421V Chr.7: 41689669-41689669 on Build GRCh38
311998 c.1141C>T p.H381Y Chr.7: 41689790-41689790 on Build GRCh38
3638669 c.1248G>A p.Q416Q Chr.7: 41689683-41689683 on Build GRCh38
379997 c.1227C>G p.N409K Chr.7: 41689704-41689704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759695534 Chr.7: 41689768-41689768 on Build GRCh38
rs927870191 Chr.7: 41689452-41689452 on Build GRCh38
rs1003967321 Chr.7: 41689514-41689514 on Build GRCh38
rs372914328 Chr.7: 41689619-41689619 on Build GRCh38
rs766398833 Chr.7: 41689607-41689607 on Build GRCh38
rs376978623 Chr.7: 41689400-41689400 on Build GRCh38
rs776889307 Chr.7: 41689617-41689617 on Build GRCh38
rs749414503 Chr.7: 41689431-41689431 on Build GRCh38
rs984086679 Chr.7: 41689551-41689551 on Build GRCh38
rs778157114 Chr.7: 41689731-41689731 on Build GRCh38
rs747229208 Chr.7: 41689432-41689432 on Build GRCh38
rs576489367 Chr.7: 41689331-41689331 on Build GRCh38
rs376487431 Chr.7: 41689656-41689656 on Build GRCh38
rs748595617 Chr.7: 41689692-41689692 on Build GRCh38
rs762855304 Chr.7: 41689632-41689632 on Build GRCh38
rs149050844 Chr.7: 41689755-41689755 on Build GRCh38
rs1047938669 Chr.7: 41689454-41689454 on Build GRCh38
rs779393124 Chr.7: 41689691-41689691 on Build GRCh38
rs187123304 Chr.7: 41689339-41689339 on Build GRCh38
rs996563855 Chr.7: 41689528-41689528 on Build GRCh38
rs529491006 Chr.7: 41689524-41689524 on Build GRCh38
rs752615625 Chr.7: 41689628-41689628 on Build GRCh38
rs1016658845 Chr.7: 41689522-41689522 on Build GRCh38
rs202220254 Chr.7: 41689523-41689524 on Build GRCh38
rs771243659 Chr.7: 41689747-41689747 on Build GRCh38
rs955344450 Chr.7: 41689530-41689531 on Build GRCh38
rs879242468 Chr.7: 41689524-41689524 on Build GRCh38
rs955841307 Chr.7: 41689653-41689653 on Build GRCh38
rs151304258 Chr.7: 41689513-41689514 on Build GRCh38
rs914585640 Chr.7: 41689374-41689374 on Build GRCh38
rs559661131 Chr.7: 41689566-41689566 on Build GRCh38
rs929995207 Chr.7: 41689376-41689376 on Build GRCh38
rs1054432355 Chr.7: 41689368-41689368 on Build GRCh38
rs372749601 Chr.7: 41689340-41689340 on Build GRCh38
rs113772875 Chr.7: 41689720-41689720 on Build GRCh38
rs576554932 Chr.7: 41689530-41689530 on Build GRCh38
rs1003525163 Chr.7: 41689453-41689453 on Build GRCh38
rs558930948 Chr.7: 41689366-41689366 on Build GRCh38
rs765335292 Chr.7: 41689625-41689625 on Build GRCh38
rs551233348 Chr.7: 41689560-41689560 on Build GRCh38
rs907476484 Chr.7: 41689450-41689450 on Build GRCh38
rs751206761 Chr.7: 41689649-41689649 on Build GRCh38
rs939324583 Chr.7: 41689511-41689511 on Build GRCh38
rs918548026 Chr.7: 41689361-41689361 on Build GRCh38
rs975742391 Chr.7: 41689322-41689322 on Build GRCh38
rs886583686 Chr.7: 41689513-41689513 on Build GRCh38
rs1055359616 Chr.7: 41689610-41689610 on Build GRCh38
rs974192572 Chr.7: 41689325-41689325 on Build GRCh38
rs540587378 Chr.7: 41689587-41689587 on Build GRCh38
rs763904229 Chr.7: 41689645-41689645 on Build GRCh38
rs191980835 Chr.7: 41689424-41689424 on Build GRCh38
rs894989688 Chr.7: 41689661-41689661 on Build GRCh38
rs962767909 Chr.7: 41689522-41689523 on Build GRCh38
rs77416038 Chr.7: 41689534-41689534 on Build GRCh38
rs755611862 Chr.7: 41689674-41689674 on Build GRCh38
rs78056785 Chr.7: 41689535-41689535 on Build GRCh38
rs543099568 Chr.7: 41689428-41689428 on Build GRCh38
rs755071449 Chr.7: 41689391-41689391 on Build GRCh38
rs747304324 Chr.7: 41689739-41689739 on Build GRCh38
rs749989847 Chr.7: 41689673-41689673 on Build GRCh38
rs772344962 Chr.7: 41689712-41689712 on Build GRCh38
rs983301858 Chr.7: 41689397-41689397 on Build GRCh38
rs986649027 Chr.7: 41689531-41689531 on Build GRCh38

View Full Product Details