Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45892876-45893384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFBP1
Entrez Gene ID
3484
Gene Name
insulin like growth factor binding protein 1
Gene Aliases
AFBP, IBP1, IGF-BP25, PP12, hIGFBP-1
UniGene
Hs.642938
Gene Chromosome Location
Chr.7: 45888360-45893668 on Build GRCh38
Transcripts
NM_000596.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5407576 c.717C>T p.I239I Chr.7: 45893028-45893028 on Build GRCh38
3762690 c.759A>G p.I253M Chr.7: 45893070-45893070 on Build GRCh38
5414333 c.719C>T p.P240L Chr.7: 45893030-45893030 on Build GRCh38
6892094 c.668G>A p.G223E Chr.7: 45892979-45892979 on Build GRCh38
6440596 c.706G>A p.G236R Chr.7: 45893017-45893017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760906614 Chr.7: 45893021-45893021 on Build GRCh38
rs144131273 Chr.7: 45893289-45893289 on Build GRCh38
rs764456292 Chr.7: 45893030-45893030 on Build GRCh38
rs531622833 Chr.7: 45893079-45893079 on Build GRCh38
rs4619 Chr.7: 45893070-45893070 on Build GRCh38
rs368771684 Chr.7: 45893158-45893158 on Build GRCh38
rs754497011 Chr.7: 45892927-45892927 on Build GRCh38
rs746568589 Chr.7: 45892924-45892924 on Build GRCh38
rs904838949 Chr.7: 45893259-45893259 on Build GRCh38
rs753251942 Chr.7: 45893172-45893172 on Build GRCh38
rs370766303 Chr.7: 45893002-45893002 on Build GRCh38
rs756050097 Chr.7: 45893141-45893141 on Build GRCh38
rs905081489 Chr.7: 45892998-45892998 on Build GRCh38
rs9658227 Chr.7: 45893167-45893168 on Build GRCh38
rs368307845 Chr.7: 45892965-45892965 on Build GRCh38
rs368331647 Chr.7: 45893118-45893118 on Build GRCh38
rs770923256 Chr.7: 45892986-45892986 on Build GRCh38
rs773089001 Chr.7: 45892969-45892969 on Build GRCh38
rs765553299 Chr.7: 45893122-45893122 on Build GRCh38
rs750924174 Chr.7: 45893129-45893129 on Build GRCh38
rs762162861 Chr.7: 45893038-45893038 on Build GRCh38
rs112808627 Chr.7: 45893256-45893257 on Build GRCh38
rs1036570215 Chr.7: 45893184-45893184 on Build GRCh38
rs780713458 Chr.7: 45893060-45893060 on Build GRCh38
rs1020324299 Chr.7: 45893063-45893063 on Build GRCh38
rs754485600 Chr.7: 45893051-45893051 on Build GRCh38
rs770829397 Chr.7: 45893083-45893083 on Build GRCh38
rs555524874 Chr.7: 45893323-45893323 on Build GRCh38
rs778465247 Chr.7: 45893348-45893348 on Build GRCh38
rs762115990 Chr.7: 45893031-45893031 on Build GRCh38
rs774132945 Chr.7: 45893019-45893020 on Build GRCh38
rs772071061 Chr.7: 45893103-45893103 on Build GRCh38
rs759586433 Chr.7: 45893000-45893000 on Build GRCh38
rs1034970689 Chr.7: 45893016-45893016 on Build GRCh38
rs750919434 Chr.7: 45893046-45893046 on Build GRCh38
rs940787456 Chr.7: 45893166-45893167 on Build GRCh38
rs1003241894 Chr.7: 45893282-45893282 on Build GRCh38
rs932238692 Chr.7: 45893182-45893182 on Build GRCh38
rs745782423 Chr.7: 45893099-45893099 on Build GRCh38
rs890730465 Chr.7: 45893222-45893222 on Build GRCh38
rs1009316935 Chr.7: 45893329-45893329 on Build GRCh38
rs899363604 Chr.7: 45893173-45893178 on Build GRCh38
rs72563789 Chr.7: 45893111-45893111 on Build GRCh38
rs1008939481 Chr.7: 45893242-45893245 on Build GRCh38
rs898882368 Chr.7: 45893191-45893191 on Build GRCh38
rs988110489 Chr.7: 45893133-45893133 on Build GRCh38
rs371743921 Chr.7: 45893334-45893334 on Build GRCh38
rs536386354 Chr.7: 45893302-45893302 on Build GRCh38
rs1022121022 Chr.7: 45893292-45893293 on Build GRCh38
rs1052652 Chr.7: 45893195-45893195 on Build GRCh38
rs13619 Chr.7: 45893252-45893252 on Build GRCh38
rs767509757 Chr.7: 45893115-45893116 on Build GRCh38
rs1010703059 Chr.7: 45893279-45893279 on Build GRCh38
rs373819923 Chr.7: 45893135-45893135 on Build GRCh38
rs892210173 Chr.7: 45893275-45893275 on Build GRCh38
rs766051003 Chr.7: 45893151-45893151 on Build GRCh38
rs774337911 Chr.7: 45892991-45892991 on Build GRCh38
rs375166026 Chr.7: 45893061-45893061 on Build GRCh38
rs761470489 Chr.7: 45893048-45893048 on Build GRCh38
rs750379996 Chr.7: 45893134-45893138 on Build GRCh38
rs778759405 Chr.7: 45893089-45893089 on Build GRCh38
rs141120529 Chr.7: 45893041-45893041 on Build GRCh38
rs775582580 Chr.7: 45893106-45893106 on Build GRCh38
rs747772753 Chr.7: 45893349-45893349 on Build GRCh38
rs1036100204 Chr.7: 45892941-45892941 on Build GRCh38
rs956735181 Chr.7: 45893033-45893033 on Build GRCh38
rs117054298 Chr.7: 45892964-45892964 on Build GRCh38
rs550128435 Chr.7: 45893097-45893097 on Build GRCh38
rs758205629 Chr.7: 45893351-45893351 on Build GRCh38
rs201339143 Chr.7: 45892979-45892979 on Build GRCh38
rs1022090374 Chr.7: 45893341-45893341 on Build GRCh38
rs892424163 Chr.7: 45893320-45893320 on Build GRCh38
rs201675959 Chr.7: 45893117-45893117 on Build GRCh38
rs760892744 Chr.7: 45893108-45893108 on Build GRCh38
rs766945964 Chr.7: 45893138-45893138 on Build GRCh38
rs4988516 Chr.7: 45893325-45893325 on Build GRCh38
rs9658228 Chr.7: 45893184-45893185 on Build GRCh38
rs4988515 Chr.7: 45893001-45893001 on Build GRCh38
rs930394005 Chr.7: 45893192-45893192 on Build GRCh38
rs775545221 Chr.7: 45892928-45892928 on Build GRCh38
rs145712942 Chr.7: 45893073-45893073 on Build GRCh38
rs1035343714 Chr.7: 45893294-45893294 on Build GRCh38
rs747805510 Chr.7: 45892945-45892945 on Build GRCh38

View Full Product Details