Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 47655215-47655720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf65
Entrez Gene ID
401335
Gene Name
chromosome 7 open reading frame 65
Gene Aliases
-
UniGene
Hs.520816
Gene Chromosome Location
Chr.7: 47655244-47661648 on Build GRCh38
Transcripts
NM_001123065.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1673178 c.49G>A p.G17S Chr.7: 47655327-47655327 on Build GRCh38
365875 c.9_10GG>AT p.M3>? Chr.7: 47655287-47655288 on Build GRCh38
6616659 c.37C>T p.R13W Chr.7: 47655315-47655315 on Build GRCh38
453138 c.47C>T p.P16L Chr.7: 47655325-47655325 on Build GRCh38
5546242 c.31G>A p.G11R Chr.7: 47655309-47655309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967594701 Chr.7: 47655301-47655301 on Build GRCh38
rs1048142218 Chr.7: 47655547-47655547 on Build GRCh38
rs372370028 Chr.7: 47655283-47655283 on Build GRCh38
rs539771185 Chr.7: 47655326-47655326 on Build GRCh38
rs928956143 Chr.7: 47655541-47655541 on Build GRCh38
rs556003684 Chr.7: 47655530-47655530 on Build GRCh38
rs998898207 Chr.7: 47655518-47655518 on Build GRCh38
rs753525683 Chr.7: 47655315-47655315 on Build GRCh38
rs74838059 Chr.7: 47655304-47655304 on Build GRCh38
rs376792870 Chr.7: 47655240-47655240 on Build GRCh38
rs553595625 Chr.7: 47655362-47655362 on Build GRCh38
rs778315138 Chr.7: 47655330-47655330 on Build GRCh38
rs901535815 Chr.7: 47655534-47655534 on Build GRCh38
rs201575562 Chr.7: 47655296-47655296 on Build GRCh38
rs928507812 Chr.7: 47655688-47655688 on Build GRCh38
rs765198568 Chr.7: 47655317-47655317 on Build GRCh38
rs781643262 Chr.7: 47655327-47655327 on Build GRCh38
rs758606274 Chr.7: 47655323-47655323 on Build GRCh38
rs903167140 Chr.7: 47655512-47655512 on Build GRCh38
rs763996078 Chr.7: 47655314-47655314 on Build GRCh38
rs770161662 Chr.7: 47655269-47655269 on Build GRCh38
rs984489795 Chr.7: 47655644-47655644 on Build GRCh38
rs1013693203 Chr.7: 47655605-47655605 on Build GRCh38
rs761456032 Chr.7: 47655369-47655369 on Build GRCh38
rs938505610 Chr.7: 47655482-47655483 on Build GRCh38
rs747806573 Chr.7: 47655620-47655620 on Build GRCh38
rs1055638487 Chr.7: 47655491-47655492 on Build GRCh38
rs557913032 Chr.7: 47655347-47655347 on Build GRCh38
rs747219038 Chr.7: 47655264-47655264 on Build GRCh38
rs748713671 Chr.7: 47655268-47655268 on Build GRCh38
rs986082425 Chr.7: 47655680-47655680 on Build GRCh38
rs921644623 Chr.7: 47655352-47655352 on Build GRCh38
rs1036219054 Chr.7: 47655445-47655445 on Build GRCh38
rs1011682610 Chr.7: 47655505-47655505 on Build GRCh38
rs749795939 Chr.7: 47655342-47655342 on Build GRCh38
rs35807095 Chr.7: 47655554-47655555 on Build GRCh38
rs1006044923 Chr.7: 47655287-47655287 on Build GRCh38
rs753850334 Chr.7: 47655239-47655239 on Build GRCh38
rs778837287 Chr.7: 47655387-47655388 on Build GRCh38
rs984396235 Chr.7: 47655373-47655373 on Build GRCh38
rs750544399 Chr.7: 47655321-47655321 on Build GRCh38
rs966674564 Chr.7: 47655363-47655363 on Build GRCh38
rs566762291 Chr.7: 47655511-47655511 on Build GRCh38
rs115666487 Chr.7: 47655498-47655498 on Build GRCh38
rs765108737 Chr.7: 47655370-47655370 on Build GRCh38
rs769129371 Chr.7: 47655267-47655267 on Build GRCh38
rs192455523 Chr.7: 47655661-47655661 on Build GRCh38
rs188912338 Chr.7: 47655460-47655460 on Build GRCh38
rs978772070 Chr.7: 47655405-47655405 on Build GRCh38
rs780365153 Chr.7: 47655260-47655260 on Build GRCh38
rs538956135 Chr.7: 47655549-47655549 on Build GRCh38
rs779377336 Chr.7: 47655272-47655272 on Build GRCh38
rs773349157 Chr.7: 47655372-47655372 on Build GRCh38
rs758698031 Chr.7: 47655259-47655259 on Build GRCh38
rs73697723 Chr.7: 47655246-47655246 on Build GRCh38
rs993388773 Chr.7: 47655266-47655266 on Build GRCh38
rs780275167 Chr.7: 47655325-47655325 on Build GRCh38
rs771234518 Chr.7: 47655656-47655656 on Build GRCh38
rs144638847 Chr.7: 47655592-47655592 on Build GRCh38
rs558674519 Chr.7: 47655587-47655587 on Build GRCh38
rs530284539 Chr.7: 47655666-47655666 on Build GRCh38
rs1006511233 Chr.7: 47655578-47655578 on Build GRCh38
rs894950824 Chr.7: 47655588-47655588 on Build GRCh38
rs571280087 Chr.7: 47655544-47655544 on Build GRCh38
rs768118071 Chr.7: 47655574-47655574 on Build GRCh38
rs1040073087 Chr.7: 47655531-47655531 on Build GRCh38
rs748395724 Chr.7: 47655328-47655328 on Build GRCh38
rs77018221 Chr.7: 47655627-47655627 on Build GRCh38
rs746692282 Chr.7: 47655351-47655351 on Build GRCh38
rs936872073 Chr.7: 47655459-47655459 on Build GRCh38
rs149267046 Chr.7: 47655361-47655362 on Build GRCh38
rs114198862 Chr.7: 47655606-47655606 on Build GRCh38
rs758992126 Chr.7: 47655295-47655295 on Build GRCh38
rs769412732 Chr.7: 47655409-47655409 on Build GRCh38
rs761837716 Chr.7: 47655316-47655316 on Build GRCh38
rs1033048206 Chr.7: 47655675-47655675 on Build GRCh38
rs56826338 Chr.7: 47655676-47655676 on Build GRCh38
rs545336607 Chr.7: 47655581-47655581 on Build GRCh38
rs145874758 Chr.7: 47655512-47655512 on Build GRCh38
rs138508901 Chr.7: 47655351-47655352 on Build GRCh38
rs768150720 Chr.7: 47655357-47655357 on Build GRCh38

View Full Product Details