Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 47803132-47803630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf69
Entrez Gene ID
80099
Gene Name
chromosome 7 open reading frame 69
Gene Aliases
-
UniGene
Hs.287647
Gene Chromosome Location
Chr.7: 47795291-47819847 on Build GRCh38
Transcripts
NM_001302627.1 NM_025031.2 NM_138295.3 XM_017011798.1

Gene Symbol
PKD1L1
Entrez Gene ID
168507
Gene Name
polycystin 1 like 1, transient receptor potential channel interacting
Gene Aliases
PRO19563
UniGene
Hs.195979
Gene Chromosome Location
Chr.7: 47774603-47958622 on Build GRCh38
Transcripts
NM_001302627.1 NM_025031.2 NM_138295.3 XM_017011798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3923691 c.7888C>T p.P2630S Chr.7: 47803284-47803284 on Build GRCh38
6747104 c.7900A>G p.N2634D Chr.7: 47803272-47803272 on Build GRCh38
190048 c.7843C>T p.R2615W Chr.7: 47803329-47803329 on Build GRCh38
1090133 c.7913C>A p.S2638Y Chr.7: 47803259-47803259 on Build GRCh38
1090132 c.7930C>T p.R2644C Chr.7: 47803242-47803242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962487903 Chr.7: 47803525-47803525 on Build GRCh38
rs772660280 Chr.7: 47803289-47803289 on Build GRCh38
rs139902184 Chr.7: 47803332-47803332 on Build GRCh38
rs765426545 Chr.7: 47803348-47803351 on Build GRCh38
rs746166152 Chr.7: 47803269-47803269 on Build GRCh38
rs867690873 Chr.7: 47803538-47803538 on Build GRCh38
rs772397321 Chr.7: 47803166-47803166 on Build GRCh38
rs964803539 Chr.7: 47803537-47803537 on Build GRCh38
rs368489146 Chr.7: 47803242-47803242 on Build GRCh38
rs764245777 Chr.7: 47803238-47803238 on Build GRCh38
rs966946757 Chr.7: 47803499-47803499 on Build GRCh38
rs780445852 Chr.7: 47803367-47803367 on Build GRCh38
rs755385013 Chr.7: 47803454-47803454 on Build GRCh38
rs754130484 Chr.7: 47803241-47803241 on Build GRCh38
rs1053197320 Chr.7: 47803393-47803393 on Build GRCh38
rs748971698 Chr.7: 47803293-47803293 on Build GRCh38
rs559248483 Chr.7: 47803169-47803169 on Build GRCh38
rs750656096 Chr.7: 47803364-47803364 on Build GRCh38
rs777278364 Chr.7: 47803378-47803378 on Build GRCh38
rs747521865 Chr.7: 47803280-47803280 on Build GRCh38
rs112131797 Chr.7: 47803474-47803474 on Build GRCh38
rs1032193725 Chr.7: 47803174-47803174 on Build GRCh38
rs368627673 Chr.7: 47803365-47803365 on Build GRCh38
rs760048648 Chr.7: 47803486-47803486 on Build GRCh38
rs148579321 Chr.7: 47803594-47803594 on Build GRCh38
rs759530622 Chr.7: 47803231-47803231 on Build GRCh38
rs752673990 Chr.7: 47803235-47803235 on Build GRCh38
rs955215166 Chr.7: 47803256-47803256 on Build GRCh38
rs536192882 Chr.7: 47803558-47803558 on Build GRCh38
rs1010724899 Chr.7: 47803192-47803192 on Build GRCh38
rs759314352 Chr.7: 47803312-47803312 on Build GRCh38
rs769345180 Chr.7: 47803281-47803281 on Build GRCh38
rs529846904 Chr.7: 47803328-47803328 on Build GRCh38
rs752029535 Chr.7: 47803371-47803371 on Build GRCh38
rs981664178 Chr.7: 47803567-47803567 on Build GRCh38
rs1033369060 Chr.7: 47803451-47803451 on Build GRCh38
rs747394094 Chr.7: 47803437-47803437 on Build GRCh38
rs780422407 Chr.7: 47803394-47803394 on Build GRCh38
rs561100449 Chr.7: 47803222-47803222 on Build GRCh38
rs906013439 Chr.7: 47803607-47803607 on Build GRCh38
rs758780182 Chr.7: 47803271-47803271 on Build GRCh38
rs755438540 Chr.7: 47803377-47803377 on Build GRCh38
rs776220522 Chr.7: 47803167-47803167 on Build GRCh38
rs892272150 Chr.7: 47803188-47803188 on Build GRCh38
rs1015323907 Chr.7: 47803504-47803504 on Build GRCh38
rs191498363 Chr.7: 47803542-47803542 on Build GRCh38
rs765250220 Chr.7: 47803489-47803489 on Build GRCh38
rs200087970 Chr.7: 47803356-47803356 on Build GRCh38
rs375628322 Chr.7: 47803360-47803360 on Build GRCh38
rs772893346 Chr.7: 47803178-47803178 on Build GRCh38
rs758738763 Chr.7: 47803372-47803372 on Build GRCh38
rs1043573295 Chr.7: 47803422-47803422 on Build GRCh38
rs576229940 Chr.7: 47803595-47803595 on Build GRCh38
rs149874243 Chr.7: 47803475-47803475 on Build GRCh38
rs759931520 Chr.7: 47803289-47803290 on Build GRCh38
rs928912649 Chr.7: 47803577-47803577 on Build GRCh38
rs139115507 Chr.7: 47803278-47803278 on Build GRCh38
rs762572525 Chr.7: 47803180-47803180 on Build GRCh38
rs766185571 Chr.7: 47803186-47803186 on Build GRCh38
rs754419298 Chr.7: 47803227-47803228 on Build GRCh38
rs371109435 Chr.7: 47803294-47803294 on Build GRCh38
rs1039042607 Chr.7: 47803267-47803267 on Build GRCh38
rs144907917 Chr.7: 47803557-47803557 on Build GRCh38
rs948915766 Chr.7: 47803334-47803334 on Build GRCh38
rs987228462 Chr.7: 47803522-47803522 on Build GRCh38
rs571378951 Chr.7: 47803426-47803426 on Build GRCh38
rs141685583 Chr.7: 47803296-47803296 on Build GRCh38
rs1010690471 Chr.7: 47803402-47803402 on Build GRCh38
rs899681168 Chr.7: 47803430-47803430 on Build GRCh38
rs779735171 Chr.7: 47803201-47803201 on Build GRCh38
rs769290303 Chr.7: 47803176-47803176 on Build GRCh38
rs779428038 Chr.7: 47803253-47803253 on Build GRCh38
rs756185807 Chr.7: 47803237-47803237 on Build GRCh38
rs369265310 Chr.7: 47803337-47803337 on Build GRCh38
rs767311484 Chr.7: 47803314-47803314 on Build GRCh38
rs201549067 Chr.7: 47803347-47803347 on Build GRCh38
rs150526436 Chr.7: 47803321-47803321 on Build GRCh38
rs200294826 Chr.7: 47803198-47803198 on Build GRCh38

View Full Product Details