Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 48036117-48036622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf57
Entrez Gene ID
136288
Gene Name
chromosome 7 open reading frame 57
Gene Aliases
-
UniGene
Hs.258357
Gene Chromosome Location
Chr.7: 48035511-48061297 on Build GRCh38
Transcripts
NM_001100159.2 NM_001267865.1 NM_001267866.1 XM_006715649.3 XM_006715650.2 XM_011515121.1 XM_011515122.2 XM_011515123.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5937283 c.26A>T p.Q9L Chr.7: 48036334-48036334 on Build GRCh38
3768408 c.31G>A p.A11T Chr.7: 48036339-48036339 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs991207071 Chr.7: 48036210-48036210 on Build GRCh38
rs910571672 Chr.7: 48036574-48036574 on Build GRCh38
rs767039010 Chr.7: 48036585-48036585 on Build GRCh38
rs770353356 Chr.7: 48036368-48036368 on Build GRCh38
rs547919970 Chr.7: 48036451-48036451 on Build GRCh38
rs757318512 Chr.7: 48036331-48036331 on Build GRCh38
rs71547874 Chr.7: 48036192-48036192 on Build GRCh38
rs777587593 Chr.7: 48036313-48036313 on Build GRCh38
rs1001306388 Chr.7: 48036414-48036414 on Build GRCh38
rs748814673 Chr.7: 48036433-48036433 on Build GRCh38
rs770011318 Chr.7: 48036173-48036173 on Build GRCh38
rs905619379 Chr.7: 48036155-48036155 on Build GRCh38
rs766968928 Chr.7: 48036380-48036380 on Build GRCh38
rs370390818 Chr.7: 48036260-48036260 on Build GRCh38
rs775649080 Chr.7: 48036338-48036347 on Build GRCh38
rs763376581 Chr.7: 48036207-48036207 on Build GRCh38
rs528179783 Chr.7: 48036399-48036399 on Build GRCh38
rs759043075 Chr.7: 48036293-48036293 on Build GRCh38
rs1008157533 Chr.7: 48036500-48036500 on Build GRCh38
rs943355996 Chr.7: 48036593-48036593 on Build GRCh38
rs950226303 Chr.7: 48036241-48036241 on Build GRCh38
rs763637098 Chr.7: 48036387-48036387 on Build GRCh38
rs958334784 Chr.7: 48036449-48036449 on Build GRCh38
rs760421400 Chr.7: 48036385-48036385 on Build GRCh38
rs955622684 Chr.7: 48036490-48036490 on Build GRCh38
rs370670796 Chr.7: 48036363-48036363 on Build GRCh38
rs745942739 Chr.7: 48036339-48036339 on Build GRCh38
rs772221181 Chr.7: 48036348-48036348 on Build GRCh38
rs924733240 Chr.7: 48036501-48036501 on Build GRCh38
rs745729969 Chr.7: 48036318-48036318 on Build GRCh38
rs905588121 Chr.7: 48036388-48036388 on Build GRCh38
rs866751703 Chr.7: 48036300-48036300 on Build GRCh38
rs1020459791 Chr.7: 48036463-48036463 on Build GRCh38
rs567714347 Chr.7: 48036453-48036453 on Build GRCh38
rs553479252 Chr.7: 48036163-48036163 on Build GRCh38
rs562054881 Chr.7: 48036186-48036186 on Build GRCh38
rs546811072 Chr.7: 48036309-48036309 on Build GRCh38
rs539637062 Chr.7: 48036158-48036158 on Build GRCh38
rs557266659 Chr.7: 48036149-48036149 on Build GRCh38
rs752340669 Chr.7: 48036307-48036307 on Build GRCh38
rs972006224 Chr.7: 48036465-48036465 on Build GRCh38
rs957828254 Chr.7: 48036191-48036191 on Build GRCh38
rs1054642958 Chr.7: 48036439-48036439 on Build GRCh38
rs373987914 Chr.7: 48036583-48036583 on Build GRCh38
rs537076192 Chr.7: 48036559-48036559 on Build GRCh38
rs759063588 Chr.7: 48036378-48036378 on Build GRCh38
rs1042389126 Chr.7: 48036247-48036247 on Build GRCh38
rs550569614 Chr.7: 48036566-48036566 on Build GRCh38
rs777104388 Chr.7: 48036362-48036362 on Build GRCh38
rs778985529 Chr.7: 48036335-48036335 on Build GRCh38
rs573380768 Chr.7: 48036165-48036165 on Build GRCh38
rs145111939 Chr.7: 48036284-48036284 on Build GRCh38
rs917438973 Chr.7: 48036216-48036216 on Build GRCh38
rs567504868 Chr.7: 48036156-48036156 on Build GRCh38
rs920425483 Chr.7: 48036298-48036298 on Build GRCh38
rs768727171 Chr.7: 48036361-48036361 on Build GRCh38
rs765278417 Chr.7: 48036407-48036407 on Build GRCh38
rs757041560 Chr.7: 48036403-48036403 on Build GRCh38
rs201858531 Chr.7: 48036269-48036269 on Build GRCh38
rs368113912 Chr.7: 48036353-48036353 on Build GRCh38
rs886392325 Chr.7: 48036354-48036354 on Build GRCh38
rs769014737 Chr.7: 48036255-48036255 on Build GRCh38
rs747213747 Chr.7: 48036359-48036359 on Build GRCh38
rs769864837 Chr.7: 48036369-48036369 on Build GRCh38
rs760627419 Chr.7: 48036464-48036464 on Build GRCh38
rs753552336 Chr.7: 48036326-48036326 on Build GRCh38
rs1010854506 Chr.7: 48036455-48036455 on Build GRCh38
rs779695952 Chr.7: 48036418-48036418 on Build GRCh38
rs753730472 Chr.7: 48036390-48036390 on Build GRCh38
rs760960686 Chr.7: 48036253-48036253 on Build GRCh38
rs375254209 Chr.7: 48036295-48036295 on Build GRCh38
rs977614069 Chr.7: 48036473-48036473 on Build GRCh38
rs1018242601 Chr.7: 48036564-48036564 on Build GRCh38
rs1047639225 Chr.7: 48036349-48036349 on Build GRCh38
rs542205577 Chr.7: 48036172-48036172 on Build GRCh38
rs775038911 Chr.7: 48036383-48036383 on Build GRCh38
rs544557830 Chr.7: 48036249-48036249 on Build GRCh38

View Full Product Details