Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55978175-55978585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBAS
Entrez Gene ID
2631
Gene Name
glioblastoma amplified sequence
Gene Aliases
NIPSNAP2
UniGene
Hs.591069
Gene Chromosome Location
Chr.7: 55964577-56000182 on Build GRCh38
Transcripts
NM_001202469.1 NM_001483.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6850925 c.193C>T p.L65L Chr.7: 55978226-55978226 on Build GRCh38
4686521 c.199delA p.K68fs*19 Chr.7: 55978232-55978232 on Build GRCh38
4587492 c.188A>G p.N63S Chr.7: 55978221-55978221 on Build GRCh38
6178020 c.193C>G p.L65V Chr.7: 55978226-55978226 on Build GRCh38
6669718 c.155G>A p.R52Q Chr.7: 55978188-55978188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772982475 Chr.7: 55978204-55978204 on Build GRCh38
rs763070402 Chr.7: 55978387-55978387 on Build GRCh38
rs1047613896 Chr.7: 55978541-55978541 on Build GRCh38
rs528807528 Chr.7: 55978515-55978515 on Build GRCh38
rs768613413 Chr.7: 55978357-55978357 on Build GRCh38
rs753047060 Chr.7: 55978412-55978412 on Build GRCh38
rs780603074 Chr.7: 55978294-55978294 on Build GRCh38
rs761608201 Chr.7: 55978377-55978377 on Build GRCh38
rs541204590 Chr.7: 55978252-55978252 on Build GRCh38
rs886297170 Chr.7: 55978543-55978543 on Build GRCh38
rs563237137 Chr.7: 55978420-55978420 on Build GRCh38
rs754297119 Chr.7: 55978221-55978221 on Build GRCh38
rs761984437 Chr.7: 55978280-55978280 on Build GRCh38
rs775117472 Chr.7: 55978358-55978358 on Build GRCh38
rs781439599 Chr.7: 55978407-55978407 on Build GRCh38
rs746283251 Chr.7: 55978439-55978442 on Build GRCh38
rs776360213 Chr.7: 55978366-55978366 on Build GRCh38
rs867333386 Chr.7: 55978517-55978517 on Build GRCh38
rs371051143 Chr.7: 55978287-55978287 on Build GRCh38
rs1042695524 Chr.7: 55978556-55978556 on Build GRCh38
rs752060117 Chr.7: 55978300-55978300 on Build GRCh38
rs757966247 Chr.7: 55978266-55978269 on Build GRCh38
rs1033976507 Chr.7: 55978318-55978318 on Build GRCh38
rs765493047 Chr.7: 55978494-55978494 on Build GRCh38
rs781717291 Chr.7: 55978309-55978309 on Build GRCh38
rs768965920 Chr.7: 55978201-55978201 on Build GRCh38
rs762274502 Chr.7: 55978409-55978409 on Build GRCh38
rs561225775 Chr.7: 55978442-55978442 on Build GRCh38
rs542272079 Chr.7: 55978441-55978441 on Build GRCh38
rs750677147 Chr.7: 55978386-55978386 on Build GRCh38
rs140053997 Chr.7: 55978328-55978328 on Build GRCh38
rs546784076 Chr.7: 55978537-55978537 on Build GRCh38
rs763926216 Chr.7: 55978262-55978262 on Build GRCh38
rs767426105 Chr.7: 55978233-55978233 on Build GRCh38
rs765466206 Chr.7: 55978284-55978284 on Build GRCh38
rs932301902 Chr.7: 55978536-55978536 on Build GRCh38
rs969791373 Chr.7: 55978236-55978236 on Build GRCh38
rs766415142 Chr.7: 55978399-55978399 on Build GRCh38
rs756854338 Chr.7: 55978305-55978306 on Build GRCh38
rs775360311 Chr.7: 55978237-55978237 on Build GRCh38
rs1039405625 Chr.7: 55978476-55978476 on Build GRCh38
rs778717369 Chr.7: 55978327-55978327 on Build GRCh38
rs748549406 Chr.7: 55978310-55978310 on Build GRCh38
rs776552549 Chr.7: 55978443-55978443 on Build GRCh38
rs145352702 Chr.7: 55978425-55978425 on Build GRCh38
rs752071145 Chr.7: 55978405-55978405 on Build GRCh38
rs763523916 Chr.7: 55978298-55978299 on Build GRCh38
rs553250783 Chr.7: 55978298-55978298 on Build GRCh38
rs923218605 Chr.7: 55978477-55978480 on Build GRCh38
rs765376797 Chr.7: 55978384-55978384 on Build GRCh38
rs773953831 Chr.7: 55978225-55978225 on Build GRCh38
rs775476225 Chr.7: 55978331-55978331 on Build GRCh38
rs1005951092 Chr.7: 55978272-55978272 on Build GRCh38
rs373444651 Chr.7: 55978339-55978339 on Build GRCh38
rs745557001 Chr.7: 55978421-55978421 on Build GRCh38
rs770651596 Chr.7: 55978313-55978313 on Build GRCh38
rs748995292 Chr.7: 55978217-55978217 on Build GRCh38
rs758652712 Chr.7: 55978548-55978548 on Build GRCh38
rs750444072 Chr.7: 55978286-55978286 on Build GRCh38
rs754030694 Chr.7: 55978275-55978275 on Build GRCh38
rs201209040 Chr.7: 55978223-55978223 on Build GRCh38
rs571469744 Chr.7: 55978553-55978553 on Build GRCh38
rs751224461 Chr.7: 55978299-55978299 on Build GRCh38
rs886200656 Chr.7: 55978248-55978248 on Build GRCh38
rs756616506 Chr.7: 55978413-55978413 on Build GRCh38
rs561230777 Chr.7: 55978304-55978304 on Build GRCh38
rs904193450 Chr.7: 55978560-55978560 on Build GRCh38
rs760571472 Chr.7: 55978356-55978356 on Build GRCh38
rs750747990 Chr.7: 55978498-55978498 on Build GRCh38

View Full Product Details