Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55983650-55984163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBAS
Entrez Gene ID
2631
Gene Name
glioblastoma amplified sequence
Gene Aliases
NIPSNAP2
UniGene
Hs.591069
Gene Chromosome Location
Chr.7: 55964577-56000182 on Build GRCh38
Transcripts
NM_001202469.1 NM_001483.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3267056 c.448T>G p.F150V Chr.7: 55983731-55983731 on Build GRCh38
6669719 c.455A>G p.E152G Chr.7: 55983738-55983738 on Build GRCh38
6178019 c.570G>T p.R190S Chr.7: 55983853-55983853 on Build GRCh38
3881889 c.547G>A p.G183R Chr.7: 55983830-55983830 on Build GRCh38
6471964 c.454G>A p.E152K Chr.7: 55983737-55983737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747079876 Chr.7: 55983711-55983711 on Build GRCh38
rs763497214 Chr.7: 55984122-55984125 on Build GRCh38
rs757170295 Chr.7: 55983821-55983821 on Build GRCh38
rs759430517 Chr.7: 55983744-55983745 on Build GRCh38
rs964608098 Chr.7: 55984066-55984066 on Build GRCh38
rs1005564081 Chr.7: 55983976-55983976 on Build GRCh38
rs75732486 Chr.7: 55984122-55984123 on Build GRCh38
rs776152741 Chr.7: 55983909-55983909 on Build GRCh38
rs765263887 Chr.7: 55983917-55983917 on Build GRCh38
rs749640855 Chr.7: 55983877-55983877 on Build GRCh38
rs141886054 Chr.7: 55983787-55983787 on Build GRCh38
rs76044047 Chr.7: 55983712-55983712 on Build GRCh38
rs1021002797 Chr.7: 55984038-55984038 on Build GRCh38
rs979756054 Chr.7: 55983724-55983724 on Build GRCh38
rs1017644543 Chr.7: 55983955-55983955 on Build GRCh38
rs1047175489 Chr.7: 55983940-55983940 on Build GRCh38
rs1019532528 Chr.7: 55983680-55983680 on Build GRCh38
rs1005848292 Chr.7: 55983924-55983924 on Build GRCh38
rs775406491 Chr.7: 55983914-55983915 on Build GRCh38
rs931026575 Chr.7: 55983938-55983938 on Build GRCh38
rs886652092 Chr.7: 55983957-55983957 on Build GRCh38
rs770226778 Chr.7: 55983867-55983867 on Build GRCh38
rs780430388 Chr.7: 55983708-55983708 on Build GRCh38
rs771334794 Chr.7: 55983883-55983883 on Build GRCh38
rs925150584 Chr.7: 55983766-55983766 on Build GRCh38
rs543641654 Chr.7: 55984057-55984057 on Build GRCh38
rs753319459 Chr.7: 55983811-55983811 on Build GRCh38
rs201857257 Chr.7: 55983756-55983756 on Build GRCh38
rs150298305 Chr.7: 55983979-55983980 on Build GRCh38
rs779433481 Chr.7: 55983848-55983848 on Build GRCh38
rs761237149 Chr.7: 55983912-55983912 on Build GRCh38
rs752408459 Chr.7: 55983772-55983772 on Build GRCh38
rs369625283 Chr.7: 55983829-55983829 on Build GRCh38
rs772380808 Chr.7: 55983682-55983702 on Build GRCh38
rs746360913 Chr.7: 55983884-55983884 on Build GRCh38
rs901619690 Chr.7: 55983847-55983847 on Build GRCh38
rs748197746 Chr.7: 55983865-55983865 on Build GRCh38
rs923915014 Chr.7: 55983730-55983730 on Build GRCh38
rs1019094323 Chr.7: 55983978-55983978 on Build GRCh38
rs1029122892 Chr.7: 55984115-55984115 on Build GRCh38
rs967115169 Chr.7: 55984055-55984055 on Build GRCh38
rs886114352 Chr.7: 55983906-55983907 on Build GRCh38
rs67861084 Chr.7: 55984017-55984018 on Build GRCh38
rs933135780 Chr.7: 55984120-55984123 on Build GRCh38
rs202092196 Chr.7: 55984001-55984001 on Build GRCh38
rs770231289 Chr.7: 55983743-55983743 on Build GRCh38
rs773281845 Chr.7: 55983686-55983686 on Build GRCh38
rs923114720 Chr.7: 55984119-55984119 on Build GRCh38
rs373243220 Chr.7: 55983845-55983845 on Build GRCh38
rs529926669 Chr.7: 55984047-55984047 on Build GRCh38
rs1047445895 Chr.7: 55983888-55983888 on Build GRCh38
rs902040155 Chr.7: 55983979-55983979 on Build GRCh38
rs925222671 Chr.7: 55984064-55984064 on Build GRCh38
rs979452744 Chr.7: 55984061-55984061 on Build GRCh38
rs201368231 Chr.7: 55983999-55983999 on Build GRCh38
rs183420133 Chr.7: 55983723-55983723 on Build GRCh38
rs780408156 Chr.7: 55983835-55983835 on Build GRCh38
rs763679995 Chr.7: 55983907-55983907 on Build GRCh38
rs576432730 Chr.7: 55983871-55983871 on Build GRCh38
rs573892637 Chr.7: 55984110-55984110 on Build GRCh38
rs760084984 Chr.7: 55983785-55983785 on Build GRCh38
rs773513938 Chr.7: 55983748-55983748 on Build GRCh38
rs534830958 Chr.7: 55983696-55983696 on Build GRCh38
rs68088835 Chr.7: 55984020-55984021 on Build GRCh38
rs538674610 Chr.7: 55983734-55983734 on Build GRCh38
rs764933166 Chr.7: 55983836-55983837 on Build GRCh38
rs992664372 Chr.7: 55984029-55984029 on Build GRCh38
rs760335350 Chr.7: 55983905-55983905 on Build GRCh38
rs960837130 Chr.7: 55984026-55984026 on Build GRCh38
rs758332860 Chr.7: 55983831-55983831 on Build GRCh38
rs201918543 Chr.7: 55984123-55984123 on Build GRCh38
rs187559705 Chr.7: 55984101-55984101 on Build GRCh38
rs60298274 Chr.7: 55983998-55983999 on Build GRCh38
rs750178567 Chr.7: 55983830-55983830 on Build GRCh38
rs991968632 Chr.7: 55983849-55983849 on Build GRCh38
rs992195319 Chr.7: 55983711-55983712 on Build GRCh38
rs966094369 Chr.7: 55983709-55983709 on Build GRCh38
rs747301651 Chr.7: 55983843-55983843 on Build GRCh38

View Full Product Details