Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 65960990-65961486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSB
Entrez Gene ID
2990
Gene Name
glucuronidase beta
Gene Aliases
BG, MPS7
UniGene
Hs.255230
Gene Chromosome Location
Chr.7: 65960684-65982314 on Build GRCh38
Transcripts
NM_000181.3 NM_001284290.1 NM_001293104.1 NM_001293105.1 NR_120531.1 XM_005250297.3 XM_011516113.1 XM_011516114.1 XM_017012091.1 XM_017012092.1 XM_017012093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6110761 c.1806G>C p.L602L Chr.7: 65961047-65961047 on Build GRCh38
6859061 c.1790-5delT p.? Chr.7: 65961068-65961068 on Build GRCh38
6856602 c.1790-3_1790-2insT p.? Chr.7: 65961065-65961066 on Build GRCh38
4774109 c.1820G>T p.G607V Chr.7: 65961033-65961033 on Build GRCh38
1091312 c.1793C>T p.P598L Chr.7: 65961060-65961060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755541359 Chr.7: 65961158-65961158 on Build GRCh38
rs956729849 Chr.7: 65961431-65961431 on Build GRCh38
rs1052501512 Chr.7: 65961169-65961169 on Build GRCh38
rs934427456 Chr.7: 65961350-65961350 on Build GRCh38
rs868417552 Chr.7: 65961223-65961223 on Build GRCh38
rs953646431 Chr.7: 65961088-65961088 on Build GRCh38
rs781740542 Chr.7: 65961204-65961204 on Build GRCh38
rs911853455 Chr.7: 65961154-65961154 on Build GRCh38
rs146527574 Chr.7: 65961406-65961408 on Build GRCh38
rs924392366 Chr.7: 65961341-65961341 on Build GRCh38
rs755492085 Chr.7: 65961096-65961096 on Build GRCh38
rs915308689 Chr.7: 65961181-65961181 on Build GRCh38
rs879010457 Chr.7: 65961067-65961067 on Build GRCh38
rs780346430 Chr.7: 65961115-65961115 on Build GRCh38
rs750022485 Chr.7: 65961068-65961068 on Build GRCh38
rs746043660 Chr.7: 65961103-65961103 on Build GRCh38
rs910238289 Chr.7: 65961297-65961302 on Build GRCh38
rs371981054 Chr.7: 65961067-65961067 on Build GRCh38
rs946593000 Chr.7: 65961203-65961203 on Build GRCh38
rs905118747 Chr.7: 65961250-65961250 on Build GRCh38
rs1030151298 Chr.7: 65961229-65961229 on Build GRCh38
rs987712533 Chr.7: 65961403-65961403 on Build GRCh38
rs1006666031 Chr.7: 65961456-65961456 on Build GRCh38
rs552724524 Chr.7: 65961056-65961056 on Build GRCh38
rs1023622676 Chr.7: 65961156-65961156 on Build GRCh38
rs962449223 Chr.7: 65961339-65961339 on Build GRCh38
rs752828079 Chr.7: 65961301-65961301 on Build GRCh38
rs539426890 Chr.7: 65961395-65961395 on Build GRCh38
rs753655611 Chr.7: 65961086-65961086 on Build GRCh38
rs765983374 Chr.7: 65961057-65961057 on Build GRCh38
rs1045250199 Chr.7: 65961419-65961419 on Build GRCh38
rs74802260 Chr.7: 65961426-65961426 on Build GRCh38
rs201487997 Chr.7: 65961331-65961331 on Build GRCh38
rs886743830 Chr.7: 65961313-65961313 on Build GRCh38
rs773791166 Chr.7: 65961065-65961066 on Build GRCh38
rs777173084 Chr.7: 65961031-65961031 on Build GRCh38
rs758984878 Chr.7: 65961060-65961060 on Build GRCh38
rs1006306632 Chr.7: 65961173-65961173 on Build GRCh38
rs764071830 Chr.7: 65961064-65961065 on Build GRCh38
rs1003425144 Chr.7: 65961251-65961251 on Build GRCh38
rs781380922 Chr.7: 65961213-65961215 on Build GRCh38
rs779006886 Chr.7: 65961099-65961099 on Build GRCh38
rs1016714509 Chr.7: 65961326-65961326 on Build GRCh38
rs111519653 Chr.7: 65961194-65961194 on Build GRCh38
rs767165241 Chr.7: 65961068-65961069 on Build GRCh38
rs972346388 Chr.7: 65961206-65961206 on Build GRCh38
rs935431734 Chr.7: 65961159-65961159 on Build GRCh38
rs1003867502 Chr.7: 65961320-65961320 on Build GRCh38
rs121918176 Chr.7: 65961022-65961022 on Build GRCh38
rs71564943 Chr.7: 65961263-65961263 on Build GRCh38
rs896647790 Chr.7: 65961405-65961405 on Build GRCh38
rs752200387 Chr.7: 65961068-65961068 on Build GRCh38
rs756495234 Chr.7: 65961067-65961068 on Build GRCh38
rs777986754 Chr.7: 65961385-65961385 on Build GRCh38
rs762241590 Chr.7: 65961051-65961051 on Build GRCh38
rs758534109 Chr.7: 65961111-65961111 on Build GRCh38
rs756087154 Chr.7: 65961074-65961074 on Build GRCh38
rs374209414 Chr.7: 65961092-65961092 on Build GRCh38
rs1016263198 Chr.7: 65961459-65961459 on Build GRCh38
rs763997663 Chr.7: 65961081-65961081 on Build GRCh38
rs754204979 Chr.7: 65961068-65961072 on Build GRCh38
rs751161248 Chr.7: 65961059-65961059 on Build GRCh38
rs756328110 Chr.7: 65961365-65961365 on Build GRCh38
rs767115386 Chr.7: 65961062-65961062 on Build GRCh38
rs1013774139 Chr.7: 65961153-65961153 on Build GRCh38

View Full Product Details