Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100680868-100681413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF1
Entrez Gene ID
64599
Gene Name
GRB10 interacting GYF protein 1
Gene Aliases
GYF1, PERQ1
UniGene
Hs.414396
Gene Chromosome Location
Chr.7: 100679507-100694282 on Build GRCh38
Transcripts
NM_022574.4 XM_005250532.1 XM_011516472.1 XM_011516477.1 XM_017012526.1 XM_017012527.1 XM_017012528.1 XM_017012529.1 XM_017012530.1 XM_017012531.1

SNP Detail

SNP ID Chromosome Location
rs1023154048 Chr.7: 100680937-100680937 on Build GRCh38
rs994872503 Chr.7: 100681382-100681382 on Build GRCh38
rs953323236 Chr.7: 100681041-100681041 on Build GRCh38
rs1030852818 Chr.7: 100681329-100681329 on Build GRCh38
rs747528440 Chr.7: 100680966-100680966 on Build GRCh38
rs537432115 Chr.7: 100681150-100681150 on Build GRCh38
rs1009820093 Chr.7: 100681024-100681024 on Build GRCh38
rs11461467 Chr.7: 100681374-100681374 on Build GRCh38
rs941331669 Chr.7: 100680969-100680969 on Build GRCh38
rs956513606 Chr.7: 100681367-100681367 on Build GRCh38
rs111978585 Chr.7: 100681006-100681006 on Build GRCh38
rs987196760 Chr.7: 100681302-100681303 on Build GRCh38
rs13246855 Chr.7: 100681376-100681376 on Build GRCh38
rs901501373 Chr.7: 100681223-100681223 on Build GRCh38
rs1028768656 Chr.7: 100681037-100681037 on Build GRCh38
rs1044864267 Chr.7: 100681049-100681049 on Build GRCh38
rs1005118096 Chr.7: 100681257-100681257 on Build GRCh38
rs780410637 Chr.7: 100680935-100680935 on Build GRCh38
rs112958935 Chr.7: 100680991-100680991 on Build GRCh38
rs989421245 Chr.7: 100681368-100681369 on Build GRCh38
rs1028623142 Chr.7: 100681297-100681298 on Build GRCh38
rs373098983 Chr.7: 100681200-100681200 on Build GRCh38
rs111314461 Chr.7: 100680902-100680902 on Build GRCh38
rs899130419 Chr.7: 100681375-100681375 on Build GRCh38
rs751544948 Chr.7: 100681374-100681375 on Build GRCh38
rs1003451542 Chr.7: 100681061-100681061 on Build GRCh38
rs186059947 Chr.7: 100680976-100680976 on Build GRCh38
rs1044525975 Chr.7: 100681223-100681223 on Build GRCh38
rs1005067210 Chr.7: 100681096-100681096 on Build GRCh38
rs371509308 Chr.7: 100681013-100681013 on Build GRCh38
rs190396800 Chr.7: 100681099-100681099 on Build GRCh38
rs1010436919 Chr.7: 100681210-100681210 on Build GRCh38
rs985097025 Chr.7: 100680945-100680945 on Build GRCh38
rs1016089705 Chr.7: 100681270-100681270 on Build GRCh38
rs887583993 Chr.7: 100680930-100680957 on Build GRCh38
rs905959237 Chr.7: 100681140-100681140 on Build GRCh38
rs548665481 Chr.7: 100681100-100681100 on Build GRCh38
rs908841835 Chr.7: 100680990-100680990 on Build GRCh38
rs1037035467 Chr.7: 100680975-100680975 on Build GRCh38
rs755648941 Chr.7: 100681097-100681097 on Build GRCh38
rs560710580 Chr.7: 100680891-100680891 on Build GRCh38
rs150822675 Chr.7: 100681033-100681033 on Build GRCh38
rs921754345 Chr.7: 100680918-100680918 on Build GRCh38
rs947510162 Chr.7: 100681039-100681039 on Build GRCh38
rs898870062 Chr.7: 100681292-100681292 on Build GRCh38
rs561412972 Chr.7: 100680959-100680959 on Build GRCh38
rs906007796 Chr.7: 100680998-100680998 on Build GRCh38
rs886702044 Chr.7: 100681067-100681067 on Build GRCh38
rs1045835672 Chr.7: 100681115-100681115 on Build GRCh38
rs893346363 Chr.7: 100681186-100681186 on Build GRCh38
rs1029155197 Chr.7: 100681149-100681149 on Build GRCh38
rs13233838 Chr.7: 100680992-100680992 on Build GRCh38
rs117231629 Chr.7: 100681056-100681056 on Build GRCh38
rs914610562 Chr.7: 100681194-100681194 on Build GRCh38
rs571179111 Chr.7: 100681300-100681300 on Build GRCh38
rs138816405 Chr.7: 100680962-100680963 on Build GRCh38
rs146231586 Chr.7: 100681327-100681327 on Build GRCh38
rs953175966 Chr.7: 100681259-100681259 on Build GRCh38
rs975801844 Chr.7: 100681035-100681035 on Build GRCh38
rs933128603 Chr.7: 100681009-100681009 on Build GRCh38
rs61220096 Chr.7: 100681296-100681297 on Build GRCh38
rs221791 Chr.7: 100680931-100680931 on Build GRCh38
rs987309550 Chr.7: 100681156-100681156 on Build GRCh38
rs909162763 Chr.7: 100680955-100680955 on Build GRCh38
rs909081263 Chr.7: 100681065-100681065 on Build GRCh38
rs572569048 Chr.7: 100680903-100680903 on Build GRCh38
rs972006546 Chr.7: 100681094-100681094 on Build GRCh38
rs149793679 Chr.7: 100681221-100681221 on Build GRCh38
rs940594298 Chr.7: 100681090-100681090 on Build GRCh38
rs997169547 Chr.7: 100681228-100681228 on Build GRCh38
rs915142933 Chr.7: 100681371-100681372 on Build GRCh38
rs1041991488 Chr.7: 100681215-100681215 on Build GRCh38
rs893406133 Chr.7: 100681016-100681016 on Build GRCh38
rs567063818 Chr.7: 100681147-100681147 on Build GRCh38
rs112138994 Chr.7: 100681338-100681338 on Build GRCh38
rs935256177 Chr.7: 100681160-100681163 on Build GRCh38
rs965771696 Chr.7: 100680909-100680909 on Build GRCh38
rs971954291 Chr.7: 100681302-100681302 on Build GRCh38
rs938747108 Chr.7: 100681233-100681233 on Build GRCh38
rs1001738891 Chr.7: 100680999-100680999 on Build GRCh38
rs368356220 Chr.7: 100681357-100681358 on Build GRCh38
rs905935947 Chr.7: 100681223-100681224 on Build GRCh38
rs369197183 Chr.7: 100681373-100681374 on Build GRCh38
rs556155032 Chr.7: 100681252-100681252 on Build GRCh38
rs746523933 Chr.7: 100680917-100680917 on Build GRCh38
rs968784390 Chr.7: 100681384-100681384 on Build GRCh38
rs983072741 Chr.7: 100680980-100680980 on Build GRCh38
rs537198685 Chr.7: 100681265-100681265 on Build GRCh38
rs1029132389 Chr.7: 100681247-100681247 on Build GRCh38
rs989370452 Chr.7: 100681168-100681168 on Build GRCh38
rs954616169 Chr.7: 100681298-100681299 on Build GRCh38
rs948638026 Chr.7: 100681153-100681153 on Build GRCh38
rs567471838 Chr.7: 100681295-100681296 on Build GRCh38
rs1003155276 Chr.7: 100680968-100680968 on Build GRCh38
rs947357578 Chr.7: 100681216-100681216 on Build GRCh38
rs961908815 Chr.7: 100681296-100681296 on Build GRCh38
rs221792 Chr.7: 100681034-100681034 on Build GRCh38
rs777501129 Chr.7: 100681335-100681359 on Build GRCh38

View Full Product Details