Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100681301-100681805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF1
Entrez Gene ID
64599
Gene Name
GRB10 interacting GYF protein 1
Gene Aliases
GYF1, PERQ1
UniGene
Hs.414396
Gene Chromosome Location
Chr.7: 100679507-100694282 on Build GRCh38
Transcripts
NM_022574.4 XM_005250532.1 XM_011516472.1 XM_011516477.1 XM_017012526.1 XM_017012527.1 XM_017012528.1 XM_017012529.1 XM_017012530.1 XM_017012531.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7249258 c.2213-8G>T p.? Chr.7: 100681779-100681779 on Build GRCh38
4637160 c.2227G>A p.G743R Chr.7: 100681757-100681757 on Build GRCh38
6670944 c.2251G>A p.V751M Chr.7: 100681733-100681733 on Build GRCh38
7305197 c.3056-8G>A p.? Chr.7: 100681779-100681779 on Build GRCh38
5556236 c.2226C>T p.H742H Chr.7: 100681758-100681758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184096804 Chr.7: 100681609-100681609 on Build GRCh38
rs994872503 Chr.7: 100681382-100681382 on Build GRCh38
rs535234900 Chr.7: 100681616-100681616 on Build GRCh38
rs1030852818 Chr.7: 100681329-100681329 on Build GRCh38
rs10713381 Chr.7: 100681505-100681505 on Build GRCh38
rs199837245 Chr.7: 100681688-100681688 on Build GRCh38
rs1050101720 Chr.7: 100681581-100681581 on Build GRCh38
rs554171139 Chr.7: 100681462-100681462 on Build GRCh38
rs11461467 Chr.7: 100681374-100681374 on Build GRCh38
rs981484538 Chr.7: 100681457-100681457 on Build GRCh38
rs937450490 Chr.7: 100681472-100681472 on Build GRCh38
rs371623872 Chr.7: 100681671-100681671 on Build GRCh38
rs958999790 Chr.7: 100681639-100681639 on Build GRCh38
rs372578124 Chr.7: 100681705-100681705 on Build GRCh38
rs906398885 Chr.7: 100681760-100681760 on Build GRCh38
rs956513606 Chr.7: 100681367-100681367 on Build GRCh38
rs1005966162 Chr.7: 100681603-100681603 on Build GRCh38
rs762980608 Chr.7: 100681708-100681708 on Build GRCh38
rs980519244 Chr.7: 100681437-100681437 on Build GRCh38
rs543954884 Chr.7: 100681542-100681542 on Build GRCh38
rs776501684 Chr.7: 100681700-100681700 on Build GRCh38
rs376852672 Chr.7: 100681713-100681713 on Build GRCh38
rs201424895 Chr.7: 100681516-100681516 on Build GRCh38
rs750431732 Chr.7: 100681737-100681737 on Build GRCh38
rs13246855 Chr.7: 100681376-100681376 on Build GRCh38
rs1057129093 Chr.7: 100681510-100681516 on Build GRCh38
rs560235248 Chr.7: 100681611-100681611 on Build GRCh38
rs917868065 Chr.7: 100681416-100681416 on Build GRCh38
rs989421245 Chr.7: 100681368-100681369 on Build GRCh38
rs200959228 Chr.7: 100681740-100681740 on Build GRCh38
rs527474262 Chr.7: 100681614-100681614 on Build GRCh38
rs764411848 Chr.7: 100681714-100681714 on Build GRCh38
rs4729603 Chr.7: 100681540-100681540 on Build GRCh38
rs997117228 Chr.7: 100681557-100681557 on Build GRCh38
rs144072251 Chr.7: 100681625-100681625 on Build GRCh38
rs907957884 Chr.7: 100681633-100681633 on Build GRCh38
rs760695915 Chr.7: 100681634-100681634 on Build GRCh38
rs768819516 Chr.7: 100681657-100681657 on Build GRCh38
rs1002124808 Chr.7: 100681777-100681777 on Build GRCh38
rs543856808 Chr.7: 100681736-100681736 on Build GRCh38
rs555173229 Chr.7: 100681712-100681712 on Build GRCh38
rs899130419 Chr.7: 100681375-100681375 on Build GRCh38
rs190509510 Chr.7: 100681516-100681516 on Build GRCh38
rs774118098 Chr.7: 100681757-100681757 on Build GRCh38
rs751544948 Chr.7: 100681374-100681375 on Build GRCh38
rs1026397379 Chr.7: 100681604-100681604 on Build GRCh38
rs201843233 Chr.7: 100681753-100681753 on Build GRCh38
rs899420464 Chr.7: 100681527-100681527 on Build GRCh38
rs186195280 Chr.7: 100681478-100681478 on Build GRCh38
rs933577830 Chr.7: 100681722-100681722 on Build GRCh38
rs755377225 Chr.7: 100681620-100681620 on Build GRCh38
rs199607107 Chr.7: 100681654-100681655 on Build GRCh38
rs193253445 Chr.7: 100681636-100681636 on Build GRCh38
rs182731336 Chr.7: 100681668-100681668 on Build GRCh38
rs779439784 Chr.7: 100681670-100681670 on Build GRCh38
rs536816759 Chr.7: 100681711-100681711 on Build GRCh38
rs758918093 Chr.7: 100681739-100681739 on Build GRCh38
rs771963367 Chr.7: 100681556-100681556 on Build GRCh38
rs375529824 Chr.7: 100681515-100681515 on Build GRCh38
rs1037990622 Chr.7: 100681522-100681522 on Build GRCh38
rs768375119 Chr.7: 100681687-100681687 on Build GRCh38
rs576637946 Chr.7: 100681521-100681521 on Build GRCh38
rs770926718 Chr.7: 100681754-100681754 on Build GRCh38
rs398095260 Chr.7: 100681515-100681515 on Build GRCh38
rs146231586 Chr.7: 100681327-100681327 on Build GRCh38
rs548800481 Chr.7: 100681615-100681615 on Build GRCh38
rs199929208 Chr.7: 100681781-100681781 on Build GRCh38
rs543102219 Chr.7: 100681580-100681580 on Build GRCh38
rs188600231 Chr.7: 100681626-100681626 on Build GRCh38
rs914741152 Chr.7: 100681538-100681538 on Build GRCh38
rs771745865 Chr.7: 100681762-100681762 on Build GRCh38
rs182609831 Chr.7: 100681435-100681435 on Build GRCh38
rs190393880 Chr.7: 100681505-100681505 on Build GRCh38
rs1016448649 Chr.7: 100681606-100681606 on Build GRCh38
rs747790602 Chr.7: 100681747-100681747 on Build GRCh38
rs765383035 Chr.7: 100681780-100681780 on Build GRCh38
rs532409036 Chr.7: 100681585-100681585 on Build GRCh38
rs755265221 Chr.7: 100681742-100681742 on Build GRCh38
rs116169359 Chr.7: 100681779-100681779 on Build GRCh38
rs758826136 Chr.7: 100681681-100681681 on Build GRCh38
rs915142933 Chr.7: 100681371-100681372 on Build GRCh38
rs753980145 Chr.7: 100681717-100681717 on Build GRCh38
rs373650190 Chr.7: 100681783-100681783 on Build GRCh38
rs112138994 Chr.7: 100681338-100681338 on Build GRCh38
rs747700910 Chr.7: 100681691-100681691 on Build GRCh38
rs745647111 Chr.7: 100681485-100681487 on Build GRCh38
rs1030738581 Chr.7: 100681613-100681613 on Build GRCh38
rs1010739223 Chr.7: 100681624-100681624 on Build GRCh38
rs368356220 Chr.7: 100681357-100681358 on Build GRCh38
rs576725260 Chr.7: 100681733-100681733 on Build GRCh38
rs369197183 Chr.7: 100681373-100681374 on Build GRCh38
rs370112665 Chr.7: 100681725-100681725 on Build GRCh38
rs112886587 Chr.7: 100681758-100681758 on Build GRCh38
rs968784390 Chr.7: 100681384-100681384 on Build GRCh38
rs747275752 Chr.7: 100681684-100681684 on Build GRCh38
rs981815733 Chr.7: 100681627-100681627 on Build GRCh38
rs375013469 Chr.7: 100681678-100681678 on Build GRCh38
rs780946442 Chr.7: 100681743-100681743 on Build GRCh38
rs908071780 Chr.7: 100681515-100681516 on Build GRCh38
rs777501129 Chr.7: 100681335-100681359 on Build GRCh38

View Full Product Details