Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100682928-100683431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF1
Entrez Gene ID
64599
Gene Name
GRB10 interacting GYF protein 1
Gene Aliases
GYF1, PERQ1
UniGene
Hs.414396
Gene Chromosome Location
Chr.7: 100679507-100694282 on Build GRCh38
Transcripts
NM_022574.4 XM_005250532.1 XM_011516472.1 XM_011516477.1 XM_017012526.1 XM_017012527.1 XM_017012528.1 XM_017012529.1 XM_017012530.1 XM_017012531.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
54536 c.2178G>A p.L726L Chr.7: 100683319-100683319 on Build GRCh38
3729391 c.1505G>A p.R502Q Chr.7: 100683076-100683076 on Build GRCh38
3737297 c.2114G>T p.R705L Chr.7: 100683383-100683383 on Build GRCh38
7120958 c.2090G>T p.R697L Chr.7: 100683407-100683407 on Build GRCh38
6670903 c.1240G>A p.A414T Chr.7: 100683414-100683414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757429798 Chr.7: 100683310-100683310 on Build GRCh38
rs372612503 Chr.7: 100683245-100683245 on Build GRCh38
rs374893437 Chr.7: 100683383-100683383 on Build GRCh38
rs143878382 Chr.7: 100683304-100683304 on Build GRCh38
rs367822871 Chr.7: 100683390-100683390 on Build GRCh38
rs746966202 Chr.7: 100683355-100683357 on Build GRCh38
rs750871793 Chr.7: 100683236-100683236 on Build GRCh38
rs771883212 Chr.7: 100683253-100683253 on Build GRCh38
rs745766057 Chr.7: 100683173-100683173 on Build GRCh38
rs763839483 Chr.7: 100683105-100683105 on Build GRCh38
rs761593309 Chr.7: 100683205-100683205 on Build GRCh38
rs752363021 Chr.7: 100683000-100683000 on Build GRCh38
rs202101717 Chr.7: 100683371-100683371 on Build GRCh38
rs780782148 Chr.7: 100683325-100683325 on Build GRCh38
rs376344485 Chr.7: 100683261-100683261 on Build GRCh38
rs781264689 Chr.7: 100683385-100683385 on Build GRCh38
rs568247705 Chr.7: 100683311-100683311 on Build GRCh38
rs759072400 Chr.7: 100682994-100682994 on Build GRCh38
rs556905326 Chr.7: 100683342-100683342 on Build GRCh38
rs772985821 Chr.7: 100683274-100683274 on Build GRCh38
rs563987336 Chr.7: 100683178-100683178 on Build GRCh38
rs778603866 Chr.7: 100683172-100683172 on Build GRCh38
rs780327495 Chr.7: 100683397-100683397 on Build GRCh38
rs569247361 Chr.7: 100683351-100683351 on Build GRCh38
rs757629443 Chr.7: 100683185-100683186 on Build GRCh38
rs755046743 Chr.7: 100683281-100683284 on Build GRCh38
rs766223390 Chr.7: 100683381-100683381 on Build GRCh38
rs780031087 Chr.7: 100683316-100683316 on Build GRCh38
rs1033819639 Chr.7: 100682980-100682980 on Build GRCh38
rs777957103 Chr.7: 100683161-100683161 on Build GRCh38
rs148785085 Chr.7: 100683315-100683315 on Build GRCh38
rs749362923 Chr.7: 100683270-100683271 on Build GRCh38
rs200156528 Chr.7: 100683016-100683016 on Build GRCh38
rs911032111 Chr.7: 100683144-100683144 on Build GRCh38
rs767972301 Chr.7: 100683276-100683276 on Build GRCh38
rs903516169 Chr.7: 100683367-100683367 on Build GRCh38
rs748321952 Chr.7: 100683038-100683038 on Build GRCh38
rs753925214 Chr.7: 100683307-100683307 on Build GRCh38
rs1001433829 Chr.7: 100682977-100682977 on Build GRCh38
rs369031943 Chr.7: 100683241-100683241 on Build GRCh38
rs775943625 Chr.7: 100683284-100683284 on Build GRCh38
rs752141510 Chr.7: 100683167-100683168 on Build GRCh38
rs374629274 Chr.7: 100683269-100683269 on Build GRCh38
rs752058031 Chr.7: 100683022-100683022 on Build GRCh38
rs551528395 Chr.7: 100683407-100683407 on Build GRCh38
rs770770471 Chr.7: 100683058-100683058 on Build GRCh38
rs746898061 Chr.7: 100683319-100683319 on Build GRCh38
rs370906568 Chr.7: 100683314-100683314 on Build GRCh38
rs752780672 Chr.7: 100683153-100683153 on Build GRCh38
rs772384938 Chr.7: 100683341-100683341 on Build GRCh38
rs754888392 Chr.7: 100683384-100683384 on Build GRCh38
rs771568466 Chr.7: 100683077-100683077 on Build GRCh38
rs776396510 Chr.7: 100683204-100683204 on Build GRCh38
rs748843458 Chr.7: 100683392-100683392 on Build GRCh38
rs752069062 Chr.7: 100683148-100683148 on Build GRCh38
rs531083829 Chr.7: 100683198-100683198 on Build GRCh38
rs535433466 Chr.7: 100683328-100683328 on Build GRCh38
rs756162193 Chr.7: 100683236-100683236 on Build GRCh38
rs780029270 Chr.7: 100683264-100683264 on Build GRCh38
rs769405131 Chr.7: 100683330-100683330 on Build GRCh38
rs144810973 Chr.7: 100682952-100682952 on Build GRCh38
rs774883276 Chr.7: 100683080-100683080 on Build GRCh38
rs764646476 Chr.7: 100683020-100683025 on Build GRCh38
rs761782860 Chr.7: 100683356-100683356 on Build GRCh38
rs770582603 Chr.7: 100683393-100683393 on Build GRCh38
rs766718677 Chr.7: 100683216-100683216 on Build GRCh38
rs541882700 Chr.7: 100683135-100683135 on Build GRCh38
rs943948097 Chr.7: 100683175-100683175 on Build GRCh38
rs945356104 Chr.7: 100683207-100683207 on Build GRCh38
rs747779027 Chr.7: 100683271-100683271 on Build GRCh38
rs977792634 Chr.7: 100683213-100683213 on Build GRCh38
rs148635376 Chr.7: 100683262-100683262 on Build GRCh38
rs759808299 Chr.7: 100683275-100683275 on Build GRCh38
rs774321331 Chr.7: 100683064-100683064 on Build GRCh38
rs777623530 Chr.7: 100683327-100683328 on Build GRCh38
rs768006572 Chr.7: 100683234-100683235 on Build GRCh38
rs770860493 Chr.7: 100682990-100682990 on Build GRCh38
rs221793 Chr.7: 100683273-100683273 on Build GRCh38
rs758903065 Chr.7: 100683015-100683015 on Build GRCh38
rs756254194 Chr.7: 100683154-100683154 on Build GRCh38
rs773063004 Chr.7: 100683254-100683254 on Build GRCh38
rs943342145 Chr.7: 100683021-100683021 on Build GRCh38
rs770917522 Chr.7: 100683366-100683367 on Build GRCh38
rs376603210 Chr.7: 100683106-100683106 on Build GRCh38
rs939474720 Chr.7: 100683282-100683282 on Build GRCh38
rs776772139 Chr.7: 100683369-100683369 on Build GRCh38
rs760343954 Chr.7: 100683099-100683099 on Build GRCh38
rs781162981 Chr.7: 100683037-100683037 on Build GRCh38
rs571580972 Chr.7: 100682989-100682989 on Build GRCh38
rs368127773 Chr.7: 100683338-100683338 on Build GRCh38
rs923772801 Chr.7: 100682968-100682968 on Build GRCh38
rs143900305 Chr.7: 100683321-100683321 on Build GRCh38
rs529705193 Chr.7: 100683076-100683076 on Build GRCh38
rs767907287 Chr.7: 100683227-100683227 on Build GRCh38
rs374593762 Chr.7: 100683331-100683331 on Build GRCh38
rs760024412 Chr.7: 100683226-100683226 on Build GRCh38
rs749007990 Chr.7: 100683339-100683339 on Build GRCh38
rs967726034 Chr.7: 100683195-100683195 on Build GRCh38
rs746509851 Chr.7: 100683192-100683192 on Build GRCh38
rs1012289943 Chr.7: 100683184-100683184 on Build GRCh38
rs769151234 Chr.7: 100683349-100683349 on Build GRCh38
rs770017575 Chr.7: 100683040-100683040 on Build GRCh38
rs566533404 Chr.7: 100683408-100683408 on Build GRCh38
rs201771840 Chr.7: 100683252-100683252 on Build GRCh38
rs765402186 Chr.7: 100683358-100683358 on Build GRCh38
rs766983114 Chr.7: 100682998-100682998 on Build GRCh38
rs774593198 Chr.7: 100683222-100683222 on Build GRCh38
rs749558916 Chr.7: 100683168-100683168 on Build GRCh38
rs774221587 Chr.7: 100682992-100682992 on Build GRCh38
rs765645729 Chr.7: 100683014-100683014 on Build GRCh38
rs12531792 Chr.7: 100683001-100683001 on Build GRCh38
rs1021907570 Chr.7: 100683185-100683185 on Build GRCh38
rs777478408 Chr.7: 100683050-100683050 on Build GRCh38
rs752731026 Chr.7: 100683386-100683386 on Build GRCh38
rs373297103 Chr.7: 100683057-100683057 on Build GRCh38
rs761201481 Chr.7: 100683300-100683300 on Build GRCh38
rs774995550 Chr.7: 100683183-100683183 on Build GRCh38
rs377347579 Chr.7: 100683266-100683266 on Build GRCh38
rs770728430 Chr.7: 100683169-100683169 on Build GRCh38
rs756351073 Chr.7: 100683243-100683244 on Build GRCh38
rs977843545 Chr.7: 100682964-100682964 on Build GRCh38
rs781275978 Chr.7: 100683150-100683150 on Build GRCh38
rs573370693 Chr.7: 100683269-100683270 on Build GRCh38
rs753259209 Chr.7: 100683230-100683230 on Build GRCh38
rs936730908 Chr.7: 100683235-100683235 on Build GRCh38
rs755057651 Chr.7: 100683031-100683031 on Build GRCh38

View Full Product Details