Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159307954-159308459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER1A
Entrez Gene ID
2205
Gene Name
Fc fragment of IgE receptor Ia
Gene Aliases
FCE1A, FcERI
UniGene
Hs.897
Gene Chromosome Location
Chr.1: 159283888-159308224 on Build GRCh38
Transcripts
NM_002001.3

SNP Detail

SNP ID Chromosome Location
rs182293417 Chr.1: 159308231-159308231 on Build GRCh38
rs898522031 Chr.1: 159308015-159308015 on Build GRCh38
rs902715351 Chr.1: 159308338-159308338 on Build GRCh38
rs145953038 Chr.1: 159308364-159308364 on Build GRCh38
rs79965525 Chr.1: 159308014-159308014 on Build GRCh38
rs996878087 Chr.1: 159308039-159308039 on Build GRCh38
rs979737478 Chr.1: 159308170-159308170 on Build GRCh38
rs781389683 Chr.1: 159308035-159308035 on Build GRCh38
rs545659327 Chr.1: 159308063-159308063 on Build GRCh38
rs190576386 Chr.1: 159307994-159307994 on Build GRCh38
rs888618178 Chr.1: 159308265-159308265 on Build GRCh38
rs761590395 Chr.1: 159308376-159308376 on Build GRCh38
rs961351521 Chr.1: 159308006-159308006 on Build GRCh38
rs1032598847 Chr.1: 159308340-159308340 on Build GRCh38
rs567723574 Chr.1: 159308141-159308141 on Build GRCh38
rs898486625 Chr.1: 159308283-159308283 on Build GRCh38
rs925793324 Chr.1: 159308115-159308115 on Build GRCh38
rs549636785 Chr.1: 159308128-159308128 on Build GRCh38
rs762173630 Chr.1: 159308431-159308431 on Build GRCh38
rs1001522165 Chr.1: 159308339-159308339 on Build GRCh38
rs774519329 Chr.1: 159308238-159308239 on Build GRCh38
rs932655419 Chr.1: 159308319-159308319 on Build GRCh38
rs1037656085 Chr.1: 159308269-159308269 on Build GRCh38
rs1021530214 Chr.1: 159308335-159308335 on Build GRCh38
rs959109889 Chr.1: 159308199-159308199 on Build GRCh38
rs559208548 Chr.1: 159308327-159308327 on Build GRCh38
rs371523345 Chr.1: 159308041-159308041 on Build GRCh38
rs967348811 Chr.1: 159308118-159308118 on Build GRCh38
rs1053043154 Chr.1: 159308157-159308157 on Build GRCh38
rs7549785 Chr.1: 159308078-159308078 on Build GRCh38
rs1049773465 Chr.1: 159308320-159308320 on Build GRCh38
rs990497222 Chr.1: 159308363-159308363 on Build GRCh38
rs1008365386 Chr.1: 159308083-159308083 on Build GRCh38
rs560372743 Chr.1: 159308008-159308008 on Build GRCh38
rs1024228673 Chr.1: 159308359-159308359 on Build GRCh38
rs1015853833 Chr.1: 159308330-159308330 on Build GRCh38
rs773807098 Chr.1: 159308325-159308325 on Build GRCh38
rs148254525 Chr.1: 159308436-159308436 on Build GRCh38
rs900272094 Chr.1: 159308239-159308242 on Build GRCh38
rs980353811 Chr.1: 159308089-159308089 on Build GRCh38
rs768506743 Chr.1: 159308312-159308312 on Build GRCh38
rs868534170 Chr.1: 159308028-159308028 on Build GRCh38
rs917679877 Chr.1: 159308220-159308220 on Build GRCh38
rs958994417 Chr.1: 159308354-159308354 on Build GRCh38
rs565637560 Chr.1: 159308239-159308239 on Build GRCh38
rs961566880 Chr.1: 159308343-159308343 on Build GRCh38
rs564075841 Chr.1: 159308065-159308065 on Build GRCh38
rs185718654 Chr.1: 159307989-159307989 on Build GRCh38
rs1014225338 Chr.1: 159308357-159308357 on Build GRCh38
rs1021185820 Chr.1: 159308107-159308107 on Build GRCh38
rs762784423 Chr.1: 159308294-159308294 on Build GRCh38
rs187354752 Chr.1: 159308242-159308242 on Build GRCh38
rs935942615 Chr.1: 159308152-159308152 on Build GRCh38
rs1032415857 Chr.1: 159308175-159308175 on Build GRCh38
rs925820338 Chr.1: 159308390-159308390 on Build GRCh38
rs1028401858 Chr.1: 159308056-159308056 on Build GRCh38
rs983302161 Chr.1: 159308226-159308226 on Build GRCh38
rs528845630 Chr.1: 159308182-159308182 on Build GRCh38
rs990529903 Chr.1: 159308205-159308205 on Build GRCh38
rs949169717 Chr.1: 159308225-159308225 on Build GRCh38
rs76352112 Chr.1: 159308387-159308387 on Build GRCh38

View Full Product Details